Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 125719790-125720298 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EDRF1
Entrez Gene ID
26098
Gene Name
erythroid differentiation regulatory factor 1
Gene Aliases
C10orf137
UniGene
Hs.468688
Gene Chromosome Location
Chr.10: 125719515-125764143 on Build GRCh38
Transcripts
NM_001202438.1 NM_015608.2 NR_110857.1 NR_110858.1 NR_110859.1 XM_017016066.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4633132 c.78C>A p.S26S Chr.10: 125719885-125719885 on Build GRCh38
3367944 c.80A>T p.Q27L Chr.10: 125719887-125719887 on Build GRCh38
1185120 c.85G>A p.E29K Chr.10: 125719892-125719892 on Build GRCh38
3367943 c.80A>T p.Q27L Chr.10: 125719887-125719887 on Build GRCh38
4422289 c.61G>C p.G21R Chr.10: 125719868-125719868 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769152104 Chr.10: 125719889-125719889 on Build GRCh38
rs899040247 Chr.10: 125720184-125720184 on Build GRCh38
rs148211721 Chr.10: 125719935-125719935 on Build GRCh38
rs772303462 Chr.10: 125719916-125719916 on Build GRCh38
rs941340133 Chr.10: 125720110-125720110 on Build GRCh38
rs749742052 Chr.10: 125719898-125719898 on Build GRCh38
rs774700737 Chr.10: 125719912-125719912 on Build GRCh38
rs547076699 Chr.10: 125720137-125720137 on Build GRCh38
rs746372531 Chr.10: 125719832-125719852 on Build GRCh38
rs771120801 Chr.10: 125719967-125719967 on Build GRCh38
rs768788107 Chr.10: 125719953-125719974 on Build GRCh38
rs748552701 Chr.10: 125719964-125719964 on Build GRCh38
rs751426250 Chr.10: 125719944-125719944 on Build GRCh38
rs970074794 Chr.10: 125720101-125720101 on Build GRCh38
rs930523986 Chr.10: 125720152-125720152 on Build GRCh38
rs773542299 Chr.10: 125719965-125719965 on Build GRCh38
rs751336062 Chr.10: 125719884-125719884 on Build GRCh38
rs770352516 Chr.10: 125719818-125719818 on Build GRCh38
rs766357819 Chr.10: 125719934-125719934 on Build GRCh38
rs202048511 Chr.10: 125719825-125719825 on Build GRCh38
rs758272677 Chr.10: 125719873-125719873 on Build GRCh38
rs749319293 Chr.10: 125719931-125719932 on Build GRCh38
rs762016897 Chr.10: 125719844-125719844 on Build GRCh38
rs369793672 Chr.10: 125719900-125719900 on Build GRCh38
rs760859400 Chr.10: 125719922-125719922 on Build GRCh38
rs532950476 Chr.10: 125719988-125719988 on Build GRCh38
rs145949068 Chr.10: 125719862-125719862 on Build GRCh38
rs944485700 Chr.10: 125720269-125720269 on Build GRCh38
rs200830388 Chr.10: 125719914-125719914 on Build GRCh38
rs780293524 Chr.10: 125719932-125719933 on Build GRCh38
rs774580688 Chr.10: 125719958-125719960 on Build GRCh38
rs1050285598 Chr.10: 125720245-125720245 on Build GRCh38
rs926198236 Chr.10: 125719991-125719991 on Build GRCh38
rs555027542 Chr.10: 125719888-125719888 on Build GRCh38
rs61742141 Chr.10: 125719863-125719863 on Build GRCh38
rs773275817 Chr.10: 125719921-125719921 on Build GRCh38
rs376638772 Chr.10: 125719945-125719945 on Build GRCh38
rs979453074 Chr.10: 125719990-125719990 on Build GRCh38
rs983261443 Chr.10: 125720165-125720165 on Build GRCh38
rs180763014 Chr.10: 125720002-125720002 on Build GRCh38
rs141217167 Chr.10: 125720060-125720060 on Build GRCh38
rs886196826 Chr.10: 125719969-125719969 on Build GRCh38
rs189227775 Chr.10: 125719983-125719983 on Build GRCh38
rs1008645773 Chr.10: 125719970-125719970 on Build GRCh38
rs767849331 Chr.10: 125720070-125720070 on Build GRCh38
rs756565802 Chr.10: 125720180-125720180 on Build GRCh38
rs80307570 Chr.10: 125720066-125720066 on Build GRCh38
rs917228729 Chr.10: 125719947-125719947 on Build GRCh38
rs774612696 Chr.10: 125719837-125719837 on Build GRCh38
rs755489280 Chr.10: 125719963-125719963 on Build GRCh38
rs767659888 Chr.10: 125719847-125719847 on Build GRCh38
rs764069992 Chr.10: 125719939-125719939 on Build GRCh38
rs756318027 Chr.10: 125719917-125719918 on Build GRCh38
rs768983485 Chr.10: 125719830-125719830 on Build GRCh38
rs944431721 Chr.10: 125719989-125719989 on Build GRCh38
rs887113113 Chr.10: 125719839-125719839 on Build GRCh38
rs113389269 Chr.10: 125719846-125719846 on Build GRCh38
rs901447393 Chr.10: 125719885-125719885 on Build GRCh38
rs1028569046 Chr.10: 125720025-125720025 on Build GRCh38
rs745312721 Chr.10: 125719814-125719814 on Build GRCh38
rs904732174 Chr.10: 125720035-125720035 on Build GRCh38
rs571513500 Chr.10: 125720179-125720179 on Build GRCh38
rs958884069 Chr.10: 125719996-125719996 on Build GRCh38
rs1020411684 Chr.10: 125719981-125719981 on Build GRCh38
rs777708821 Chr.10: 125719875-125719875 on Build GRCh38
rs748441009 Chr.10: 125720162-125720162 on Build GRCh38
rs528646066 Chr.10: 125720111-125720111 on Build GRCh38
rs1000375607 Chr.10: 125720039-125720039 on Build GRCh38
rs373256950 Chr.10: 125719962-125719962 on Build GRCh38
rs908564038 Chr.10: 125720095-125720095 on Build GRCh38
rs948668990 Chr.10: 125719950-125719950 on Build GRCh38
rs780857748 Chr.10: 125719811-125719811 on Build GRCh38
rs898979600 Chr.10: 125719819-125719819 on Build GRCh38
rs890293312 Chr.10: 125720266-125720266 on Build GRCh38
rs549633631 Chr.10: 125720063-125720063 on Build GRCh38
rs74162846 Chr.10: 125719958-125719958 on Build GRCh38
rs780947520 Chr.10: 125719887-125719887 on Build GRCh38
rs558045330 Chr.10: 125719860-125719860 on Build GRCh38
rs996019813 Chr.10: 125720207-125720207 on Build GRCh38
rs767235923 Chr.10: 125719955-125719955 on Build GRCh38
rs1047465294 Chr.10: 125719827-125719827 on Build GRCh38
rs1005230251 Chr.10: 125719840-125719840 on Build GRCh38
rs867119106 Chr.10: 125719824-125719824 on Build GRCh38
rs997537885 Chr.10: 125720005-125720005 on Build GRCh38
rs780656352 Chr.10: 125719895-125719895 on Build GRCh38
rs750529608 Chr.10: 125719852-125719852 on Build GRCh38
rs555854317 Chr.10: 125720129-125720129 on Build GRCh38
rs763412690 Chr.10: 125719826-125719826 on Build GRCh38
rs74162847 Chr.10: 125720045-125720045 on Build GRCh38
rs112911176 Chr.10: 125720033-125720033 on Build GRCh38
rs762992802 Chr.10: 125719936-125719936 on Build GRCh38
rs750018773 Chr.10: 125719957-125719957 on Build GRCh38

View Full Product Details