Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 132785669-132786179 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NKX6-2
Entrez Gene ID
84504
Gene Name
NK6 homeobox 2
Gene Aliases
GTX, NKX6.2, NKX6B
UniGene
Hs.134013
Gene Chromosome Location
Chr.10: 132783179-132786033 on Build GRCh38
Transcripts
NM_177400.2 XM_017016789.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6382476 c.131C>T p.A44V Chr.10: 132785818-132785818 on Build GRCh38
4707102 c.80C>T p.T27M Chr.10: 132785869-132785869 on Build GRCh38
6370340 c.77A>G p.K26R Chr.10: 132785872-132785872 on Build GRCh38
7429805 c.226_227insC p.L76fs*>203 Chr.10: 132785722-132785723 on Build GRCh38
6865680 c.181A>T p.S61C Chr.10: 132785768-132785768 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750104258 Chr.10: 132785775-132785775 on Build GRCh38
rs1053942302 Chr.10: 132785737-132785737 on Build GRCh38
rs754619745 Chr.10: 132785934-132785934 on Build GRCh38
rs987572720 Chr.10: 132786132-132786132 on Build GRCh38
rs760194097 Chr.10: 132785925-132785925 on Build GRCh38
rs771929441 Chr.10: 132785886-132785886 on Build GRCh38
rs765977892 Chr.10: 132785986-132785986 on Build GRCh38
rs995958158 Chr.10: 132786109-132786109 on Build GRCh38
rs769936637 Chr.10: 132785893-132785893 on Build GRCh38
rs765019422 Chr.10: 132785935-132785935 on Build GRCh38
rs754809922 Chr.10: 132785877-132785877 on Build GRCh38
rs902759547 Chr.10: 132785721-132785721 on Build GRCh38
rs755735425 Chr.10: 132785859-132785859 on Build GRCh38
rs867959783 Chr.10: 132786094-132786094 on Build GRCh38
rs749886753 Chr.10: 132785853-132785853 on Build GRCh38
rs762419092 Chr.10: 132785905-132785905 on Build GRCh38
rs769996722 Chr.10: 132785829-132785829 on Build GRCh38
rs777724818 Chr.10: 132785942-132785942 on Build GRCh38
rs774855370 Chr.10: 132785904-132785904 on Build GRCh38
rs1012349735 Chr.10: 132785751-132785751 on Build GRCh38
rs763247598 Chr.10: 132785898-132785898 on Build GRCh38
rs906608194 Chr.10: 132786040-132786040 on Build GRCh38
rs544054070 Chr.10: 132785705-132785705 on Build GRCh38
rs1046096610 Chr.10: 132785733-132785733 on Build GRCh38
rs768933878 Chr.10: 132785960-132785960 on Build GRCh38
rs768010945 Chr.10: 132785907-132785907 on Build GRCh38
rs201425867 Chr.10: 132785885-132785885 on Build GRCh38
rs774869718 Chr.10: 132785964-132785964 on Build GRCh38
rs779987195 Chr.10: 132785952-132785952 on Build GRCh38
rs762400550 Chr.10: 132785849-132785849 on Build GRCh38
rs199513396 Chr.10: 132785835-132785835 on Build GRCh38
rs893994455 Chr.10: 132785948-132785948 on Build GRCh38
rs923645893 Chr.10: 132786137-132786137 on Build GRCh38
rs775819490 Chr.10: 132785895-132785895 on Build GRCh38
rs1003888707 Chr.10: 132786079-132786079 on Build GRCh38
rs771054702 Chr.10: 132785813-132785813 on Build GRCh38
rs768605115 Chr.10: 132785792-132785792 on Build GRCh38
rs765650727 Chr.10: 132785715-132785715 on Build GRCh38
rs748478335 Chr.10: 132785965-132785965 on Build GRCh38
rs778783634 Chr.10: 132785799-132785799 on Build GRCh38
rs1045178618 Chr.10: 132785986-132785987 on Build GRCh38
rs778664745 Chr.10: 132785878-132785878 on Build GRCh38
rs749590861 Chr.10: 132785954-132785954 on Build GRCh38
rs563679091 Chr.10: 132785774-132785774 on Build GRCh38
rs752427216 Chr.10: 132785936-132785936 on Build GRCh38
rs752163900 Chr.10: 132785838-132785838 on Build GRCh38
rs1003991676 Chr.10: 132786054-132786060 on Build GRCh38
rs753582584 Chr.10: 132785932-132785932 on Build GRCh38
rs758415459 Chr.10: 132785951-132785952 on Build GRCh38
rs529443979 Chr.10: 132785827-132785827 on Build GRCh38
rs758354577 Chr.10: 132785939-132785939 on Build GRCh38
rs745907425 Chr.10: 132785890-132785890 on Build GRCh38
rs755790219 Chr.10: 132785787-132785787 on Build GRCh38
rs775736296 Chr.10: 132785830-132785830 on Build GRCh38
rs963191281 Chr.10: 132786105-132786105 on Build GRCh38
rs779717740 Chr.10: 132785790-132785790 on Build GRCh38
rs779708930 Chr.10: 132785863-132785863 on Build GRCh38
rs776766682 Chr.10: 132785817-132785817 on Build GRCh38
rs935539429 Chr.10: 132785722-132785722 on Build GRCh38
rs773812061 Chr.10: 132785971-132785971 on Build GRCh38
rs528160259 Chr.10: 132785966-132785966 on Build GRCh38
rs753192869 Chr.10: 132785751-132785752 on Build GRCh38
rs772490947 Chr.10: 132785969-132785969 on Build GRCh38
rs748053858 Chr.10: 132785810-132785810 on Build GRCh38
rs1052455820 Chr.10: 132785689-132785689 on Build GRCh38
rs1016662110 Chr.10: 132786139-132786139 on Build GRCh38
rs756869781 Chr.10: 132785740-132785740 on Build GRCh38
rs765777422 Chr.10: 132785928-132785928 on Build GRCh38
rs767206865 Chr.10: 132785851-132785851 on Build GRCh38
rs921652736 Chr.10: 132785866-132785866 on Build GRCh38
rs749924638 Chr.10: 132785912-132785912 on Build GRCh38
rs1028740709 Chr.10: 132786111-132786112 on Build GRCh38
rs935585851 Chr.10: 132785933-132785933 on Build GRCh38
rs1047039949 Chr.10: 132786061-132786061 on Build GRCh38
rs1016096099 Chr.10: 132786089-132786089 on Build GRCh38
rs769203805 Chr.10: 132785903-132785903 on Build GRCh38
rs749054984 Chr.10: 132785791-132785791 on Build GRCh38
rs753608135 Chr.10: 132785869-132785869 on Build GRCh38
rs906533084 Chr.10: 132785780-132785780 on Build GRCh38
rs974468123 Chr.10: 132785856-132785856 on Build GRCh38
rs1016084801 Chr.10: 132785840-132785840 on Build GRCh38
rs886382958 Chr.10: 132786072-132786072 on Build GRCh38
rs1014160317 Chr.10: 132785727-132785727 on Build GRCh38
rs761067447 Chr.10: 132785976-132785976 on Build GRCh38
rs746889156 Chr.10: 132785951-132785951 on Build GRCh38
rs866699154 Chr.10: 132785927-132785927 on Build GRCh38
rs781144494 Chr.10: 132785887-132785887 on Build GRCh38
rs956548689 Chr.10: 132786138-132786138 on Build GRCh38
rs948160364 Chr.10: 132785962-132785962 on Build GRCh38

View Full Product Details