Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 133231428-133231929 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UTF1
Entrez Gene ID
8433
Gene Name
undifferentiated embryonic cell transcription factor 1
Gene Aliases
-
UniGene
Hs.458406
Gene Chromosome Location
Chr.10: 133230274-133231558 on Build GRCh38
Transcripts
NM_003577.2

SNP Detail

SNP ID Chromosome Location
rs991420548 Chr.10: 133231848-133231848 on Build GRCh38
rs999563097 Chr.10: 133231500-133231500 on Build GRCh38
rs770826709 Chr.10: 133231465-133231465 on Build GRCh38
rs936979236 Chr.10: 133231679-133231679 on Build GRCh38
rs976618840 Chr.10: 133231537-133231537 on Build GRCh38
rs117065227 Chr.10: 133231821-133231821 on Build GRCh38
rs191619361 Chr.10: 133231756-133231756 on Build GRCh38
rs934854742 Chr.10: 133231739-133231739 on Build GRCh38
rs756346828 Chr.10: 133231806-133231806 on Build GRCh38
rs916130205 Chr.10: 133231703-133231703 on Build GRCh38
rs555411574 Chr.10: 133231498-133231498 on Build GRCh38
rs1018200853 Chr.10: 133231638-133231638 on Build GRCh38
rs867938691 Chr.10: 133231751-133231751 on Build GRCh38
rs1039393713 Chr.10: 133231791-133231791 on Build GRCh38
rs946323831 Chr.10: 133231790-133231790 on Build GRCh38
rs922640153 Chr.10: 133231544-133231544 on Build GRCh38
rs911234622 Chr.10: 133231628-133231628 on Build GRCh38
rs182258967 Chr.10: 133231516-133231516 on Build GRCh38
rs924788207 Chr.10: 133231736-133231736 on Build GRCh38
rs1055452085 Chr.10: 133231683-133231683 on Build GRCh38
rs765182848 Chr.10: 133231471-133231471 on Build GRCh38
rs997982940 Chr.10: 133231651-133231651 on Build GRCh38
rs184247842 Chr.10: 133231819-133231819 on Build GRCh38
rs551343590 Chr.10: 133231464-133231464 on Build GRCh38
rs867912963 Chr.10: 133231576-133231576 on Build GRCh38
rs1045883454 Chr.10: 133231609-133231609 on Build GRCh38
rs767566015 Chr.10: 133231479-133231479 on Build GRCh38
rs1043164912 Chr.10: 133231667-133231667 on Build GRCh38
rs536992672 Chr.10: 133231496-133231496 on Build GRCh38
rs944257102 Chr.10: 133231630-133231630 on Build GRCh38
rs991453151 Chr.10: 133231687-133231687 on Build GRCh38
rs773039581 Chr.10: 133231463-133231463 on Build GRCh38
rs1034652051 Chr.10: 133231564-133231564 on Build GRCh38
rs759318119 Chr.10: 133231468-133231468 on Build GRCh38
rs1055965307 Chr.10: 133231587-133231587 on Build GRCh38
rs981554004 Chr.10: 133231721-133231721 on Build GRCh38
rs776295028 Chr.10: 133231466-133231466 on Build GRCh38
rs1036242036 Chr.10: 133231707-133231707 on Build GRCh38
rs950316747 Chr.10: 133231710-133231710 on Build GRCh38
rs963126515 Chr.10: 133231829-133231829 on Build GRCh38
rs375809262 Chr.10: 133231875-133231875 on Build GRCh38
rs904147946 Chr.10: 133231545-133231545 on Build GRCh38
rs2998111 Chr.10: 133231518-133231518 on Build GRCh38
rs543096791 Chr.10: 133231699-133231699 on Build GRCh38
rs971396734 Chr.10: 133231866-133231866 on Build GRCh38
rs780251799 Chr.10: 133231483-133231483 on Build GRCh38
rs910090899 Chr.10: 133231470-133231470 on Build GRCh38
rs995188546 Chr.10: 133231902-133231902 on Build GRCh38
rs946459725 Chr.10: 133231906-133231906 on Build GRCh38
rs951503084 Chr.10: 133231553-133231553 on Build GRCh38
rs74973131 Chr.10: 133231828-133231828 on Build GRCh38
rs945769533 Chr.10: 133231534-133231534 on Build GRCh38
rs779868428 Chr.10: 133231676-133231676 on Build GRCh38
rs1029515459 Chr.10: 133231653-133231653 on Build GRCh38
rs909901281 Chr.10: 133231566-133231566 on Build GRCh38
rs535849605 Chr.10: 133231853-133231853 on Build GRCh38
rs976818472 Chr.10: 133231461-133231461 on Build GRCh38
rs115053355 Chr.10: 133231568-133231568 on Build GRCh38
rs528617591 Chr.10: 133231763-133231763 on Build GRCh38
rs924891988 Chr.10: 133231886-133231886 on Build GRCh38
rs999303318 Chr.10: 133231620-133231620 on Build GRCh38
rs116741200 Chr.10: 133231607-133231607 on Build GRCh38
rs897631277 Chr.10: 133231738-133231738 on Build GRCh38
rs911077923 Chr.10: 133231448-133231448 on Build GRCh38
rs983159525 Chr.10: 133231555-133231555 on Build GRCh38
rs1014410144 Chr.10: 133231486-133231486 on Build GRCh38
rs537467460 Chr.10: 133231495-133231495 on Build GRCh38
rs186819723 Chr.10: 133231672-133231672 on Build GRCh38
rs553055444 Chr.10: 133231526-133231526 on Build GRCh38
rs951521818 Chr.10: 133231664-133231664 on Build GRCh38
rs12254139 Chr.10: 133231482-133231482 on Build GRCh38
rs1010608559 Chr.10: 133231635-133231635 on Build GRCh38
rs934842187 Chr.10: 133231890-133231890 on Build GRCh38
rs530702183 Chr.10: 133231820-133231820 on Build GRCh38
rs750409928 Chr.10: 133231650-133231650 on Build GRCh38
rs978970555 Chr.10: 133231823-133231823 on Build GRCh38
rs144254871 Chr.10: 133231622-133231622 on Build GRCh38
rs557478605 Chr.10: 133231789-133231789 on Build GRCh38
rs963897470 Chr.10: 133231454-133231454 on Build GRCh38
rs74777018 Chr.10: 133231535-133231535 on Build GRCh38
rs1018265360 Chr.10: 133231788-133231788 on Build GRCh38
rs1007413606 Chr.10: 133231781-133231782 on Build GRCh38
rs748715609 Chr.10: 133231513-133231513 on Build GRCh38
rs761924830 Chr.10: 133231476-133231476 on Build GRCh38
rs990724791 Chr.10: 133231900-133231900 on Build GRCh38
rs3008362 Chr.10: 133231531-133231531 on Build GRCh38
rs1004494448 Chr.10: 133231822-133231822 on Build GRCh38
rs1036211014 Chr.10: 133231876-133231876 on Build GRCh38
rs922714286 Chr.10: 133231462-133231462 on Build GRCh38
rs985498554 Chr.10: 133231626-133231626 on Build GRCh38
rs73390631 Chr.10: 133231614-133231614 on Build GRCh38
rs752592858 Chr.10: 133231475-133231475 on Build GRCh38
rs914959823 Chr.10: 133231905-133231905 on Build GRCh38
rs893461284 Chr.10: 133231624-133231624 on Build GRCh38
rs895520593 Chr.10: 133231693-133231693 on Build GRCh38
rs1052181885 Chr.10: 133231743-133231743 on Build GRCh38
rs998120371 Chr.10: 133231811-133231811 on Build GRCh38
rs962741394 Chr.10: 133231680-133231680 on Build GRCh38
rs556513839 Chr.10: 133231501-133231501 on Build GRCh38
rs760794780 Chr.10: 133231606-133231606 on Build GRCh38

View Full Product Details