Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 799683-800184 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIDD1
Entrez Gene ID
55367
Gene Name
p53-induced death domain protein 1
Gene Aliases
LRDD, PIDD
UniGene
Hs.592290
Gene Chromosome Location
Chr.11: 799179-809504 on Build GRCh38
Transcripts
NM_145886.3 NM_145887.3 XM_005253005.4 XM_005253006.4 XM_005253007.3 XM_005253008.4 XM_011520209.2 XM_011520210.2 XM_011520211.2 XM_011520212.1 XM_011520213.2 XM_017017993.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4746439 c.2308delG p.A770fs*8 Chr.11: 799981-799981 on Build GRCh38
4037021 c.2308G>C p.A770P Chr.11: 799981-799981 on Build GRCh38
4037019 c.2392C>T p.R798C Chr.11: 799897-799897 on Build GRCh38
5704203 c.2334T>A p.N778K Chr.11: 799955-799955 on Build GRCh38
3368580 c.2473C>T p.R825W Chr.11: 799816-799816 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772679398 Chr.11: 799796-799796 on Build GRCh38
rs368878936 Chr.11: 800089-800089 on Build GRCh38
rs779410209 Chr.11: 799787-799787 on Build GRCh38
rs753563318 Chr.11: 799905-799905 on Build GRCh38
rs764745475 Chr.11: 800035-800035 on Build GRCh38
rs771866822 Chr.11: 799948-799948 on Build GRCh38
rs766913787 Chr.11: 800055-800055 on Build GRCh38
rs748574022 Chr.11: 799792-799792 on Build GRCh38
rs765357677 Chr.11: 800149-800149 on Build GRCh38
rs201365934 Chr.11: 799936-799936 on Build GRCh38
rs72844793 Chr.11: 799778-799778 on Build GRCh38
rs1046329505 Chr.11: 799825-799825 on Build GRCh38
rs891849572 Chr.11: 800090-800090 on Build GRCh38
rs773536289 Chr.11: 800115-800115 on Build GRCh38
rs758773561 Chr.11: 799906-799906 on Build GRCh38
rs745996893 Chr.11: 799863-799863 on Build GRCh38
rs947639455 Chr.11: 799830-799830 on Build GRCh38
rs769349871 Chr.11: 800098-800098 on Build GRCh38
rs753073557 Chr.11: 799845-799845 on Build GRCh38
rs534903328 Chr.11: 799943-799943 on Build GRCh38
rs765994901 Chr.11: 799955-799955 on Build GRCh38
rs768464769 Chr.11: 799865-799865 on Build GRCh38
rs776336054 Chr.11: 799962-799962 on Build GRCh38
rs774270265 Chr.11: 799890-799890 on Build GRCh38
rs778370127 Chr.11: 799850-799850 on Build GRCh38
rs780838315 Chr.11: 799776-799776 on Build GRCh38
rs752001855 Chr.11: 799921-799921 on Build GRCh38
rs1028844466 Chr.11: 799731-799731 on Build GRCh38
rs377291947 Chr.11: 799959-799959 on Build GRCh38
rs549320058 Chr.11: 799903-799903 on Build GRCh38
rs749532519 Chr.11: 799896-799896 on Build GRCh38
rs758859772 Chr.11: 799846-799846 on Build GRCh38
rs777597968 Chr.11: 800084-800084 on Build GRCh38
rs550203539 Chr.11: 799815-799815 on Build GRCh38
rs891884675 Chr.11: 799851-799851 on Build GRCh38
rs763264127 Chr.11: 799981-799981 on Build GRCh38
rs755700751 Chr.11: 799842-799842 on Build GRCh38
rs761643890 Chr.11: 799894-799894 on Build GRCh38
rs749370608 Chr.11: 799926-799926 on Build GRCh38
rs933061610 Chr.11: 800046-800046 on Build GRCh38
rs570076242 Chr.11: 799827-799827 on Build GRCh38
rs776834189 Chr.11: 799809-799809 on Build GRCh38
rs7112652 Chr.11: 799879-799879 on Build GRCh38
rs756739509 Chr.11: 799773-799773 on Build GRCh38
rs745884349 Chr.11: 799930-799930 on Build GRCh38
rs201571144 Chr.11: 800000-800000 on Build GRCh38
rs762766546 Chr.11: 799817-799817 on Build GRCh38
rs774928541 Chr.11: 800100-800100 on Build GRCh38
rs547698891 Chr.11: 799742-799742 on Build GRCh38
rs536750035 Chr.11: 799990-799990 on Build GRCh38
rs377540452 Chr.11: 800086-800086 on Build GRCh38
rs954389965 Chr.11: 799726-799726 on Build GRCh38
rs753876461 Chr.11: 800064-800064 on Build GRCh38
rs577701934 Chr.11: 799989-799989 on Build GRCh38
rs773226675 Chr.11: 799952-799952 on Build GRCh38
rs746825323 Chr.11: 800104-800104 on Build GRCh38
rs755862613 Chr.11: 799783-799783 on Build GRCh38
rs748411024 Chr.11: 800075-800075 on Build GRCh38
rs762016135 Chr.11: 800042-800042 on Build GRCh38
rs779896253 Chr.11: 799843-799843 on Build GRCh38
rs748016739 Chr.11: 799944-799944 on Build GRCh38
rs763970972 Chr.11: 799822-799822 on Build GRCh38
rs764979154 Chr.11: 799980-799980 on Build GRCh38
rs779081787 Chr.11: 800062-800062 on Build GRCh38
rs556597386 Chr.11: 799999-799999 on Build GRCh38
rs749871545 Chr.11: 799897-799897 on Build GRCh38
rs375514556 Chr.11: 799834-799834 on Build GRCh38
rs764283771 Chr.11: 800007-800008 on Build GRCh38
rs759939358 Chr.11: 799842-799843 on Build GRCh38
rs761320735 Chr.11: 799828-799828 on Build GRCh38
rs768924293 Chr.11: 799942-799942 on Build GRCh38
rs780181772 Chr.11: 799937-799937 on Build GRCh38
rs916074389 Chr.11: 799984-799984 on Build GRCh38
rs953243872 Chr.11: 799733-799733 on Build GRCh38
rs760296825 Chr.11: 799898-799898 on Build GRCh38
rs767852155 Chr.11: 799983-799983 on Build GRCh38
rs750845540 Chr.11: 800048-800048 on Build GRCh38
rs756593078 Chr.11: 800053-800053 on Build GRCh38
rs557708824 Chr.11: 799966-799966 on Build GRCh38
rs113472855 Chr.11: 800105-800105 on Build GRCh38
rs778169509 Chr.11: 800165-800165 on Build GRCh38
rs750378507 Chr.11: 800164-800191 on Build GRCh38
rs777043913 Chr.11: 799833-799843 on Build GRCh38
rs535815286 Chr.11: 799858-799858 on Build GRCh38
rs766824154 Chr.11: 800131-800131 on Build GRCh38
rs370517270 Chr.11: 799987-799987 on Build GRCh38
rs758619369 Chr.11: 800077-800077 on Build GRCh38
rs781297327 Chr.11: 800094-800094 on Build GRCh38
rs747463685 Chr.11: 799855-799855 on Build GRCh38
rs746942999 Chr.11: 800003-800003 on Build GRCh38
rs201159021 Chr.11: 800093-800093 on Build GRCh38
rs767454364 Chr.11: 800086-800086 on Build GRCh38
rs61748608 Chr.11: 799988-799988 on Build GRCh38
rs754359487 Chr.11: 800132-800132 on Build GRCh38
rs759146615 Chr.11: 800013-800013 on Build GRCh38
rs372518532 Chr.11: 799832-799832 on Build GRCh38
rs367921253 Chr.11: 800007-800007 on Build GRCh38
rs756680940 Chr.11: 799986-799986 on Build GRCh38
rs757670955 Chr.11: 799925-799925 on Build GRCh38
rs900653366 Chr.11: 800163-800163 on Build GRCh38
rs778947606 Chr.11: 800074-800074 on Build GRCh38
rs762678665 Chr.11: 799873-799873 on Build GRCh38
rs761060413 Chr.11: 800118-800118 on Build GRCh38
rs758610886 Chr.11: 800164-800164 on Build GRCh38
rs983196928 Chr.11: 799760-799760 on Build GRCh38
rs1053602588 Chr.11: 799844-799844 on Build GRCh38
rs770916272 Chr.11: 799805-799805 on Build GRCh38
rs762652840 Chr.11: 800103-800103 on Build GRCh38
rs369802213 Chr.11: 799874-799874 on Build GRCh38
rs200073975 Chr.11: 799916-799916 on Build GRCh38
rs767919263 Chr.11: 800045-800045 on Build GRCh38
rs755110749 Chr.11: 800066-800066 on Build GRCh38
rs984707631 Chr.11: 799771-799771 on Build GRCh38
rs781107602 Chr.11: 799861-799861 on Build GRCh38
rs752781775 Chr.11: 800151-800151 on Build GRCh38
rs761842378 Chr.11: 800051-800052 on Build GRCh38
rs775788224 Chr.11: 799910-799912 on Build GRCh38
rs374765193 Chr.11: 800166-800166 on Build GRCh38
rs751640065 Chr.11: 800026-800026 on Build GRCh38
rs749507877 Chr.11: 799938-799938 on Build GRCh38
rs759661857 Chr.11: 799812-799812 on Build GRCh38
rs759469880 Chr.11: 800138-800138 on Build GRCh38
rs776246580 Chr.11: 800008-800008 on Build GRCh38
rs775710643 Chr.11: 799867-799867 on Build GRCh38
rs947987689 Chr.11: 799823-799823 on Build GRCh38
rs149669006 Chr.11: 799982-799982 on Build GRCh38
rs751826900 Chr.11: 799981-799981 on Build GRCh38
rs770295934 Chr.11: 799855-799857 on Build GRCh38
rs139616380 Chr.11: 799797-799797 on Build GRCh38
rs775220327 Chr.11: 800040-800040 on Build GRCh38
rs766932029 Chr.11: 799833-799833 on Build GRCh38

View Full Product Details