Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 3101511-3102010 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OSBPL5
Entrez Gene ID
114879
Gene Name
oxysterol binding protein like 5
Gene Aliases
OBPH1, ORP5
UniGene
Hs.436166
Gene Chromosome Location
Chr.11: 3087116-3165352 on Build GRCh38
Transcripts
NM_001144063.1 NM_020896.3 NM_145638.2 XM_011519873.2 XM_017017163.1 XM_017017164.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5014973 c.1444G>C p.V482L Chr.11: 3101681-3101681 on Build GRCh38
428897 c.1440G>A p.P480P Chr.11: 3101685-3101685 on Build GRCh38
3869078 c.1452C>T p.A484A Chr.11: 3101673-3101673 on Build GRCh38
4710391 c.1502C>T p.T501I Chr.11: 3101623-3101623 on Build GRCh38
4032334 c.1459G>A p.V487I Chr.11: 3101666-3101666 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs61736126 Chr.11: 3101685-3101685 on Build GRCh38
rs752121076 Chr.11: 3101586-3101586 on Build GRCh38
rs145914129 Chr.11: 3101610-3101610 on Build GRCh38
rs949258066 Chr.11: 3101535-3101535 on Build GRCh38
rs570993764 Chr.11: 3101733-3101733 on Build GRCh38
rs998951823 Chr.11: 3101858-3101858 on Build GRCh38
rs113414493 Chr.11: 3101918-3101918 on Build GRCh38
rs773101925 Chr.11: 3101648-3101648 on Build GRCh38
rs368447689 Chr.11: 3101716-3101716 on Build GRCh38
rs79593967 Chr.11: 3101925-3101925 on Build GRCh38
rs537293929 Chr.11: 3101704-3101704 on Build GRCh38
rs74587114 Chr.11: 3101558-3101558 on Build GRCh38
rs774126938 Chr.11: 3101554-3101554 on Build GRCh38
rs894619477 Chr.11: 3101937-3101937 on Build GRCh38
rs764769072 Chr.11: 3101583-3101583 on Build GRCh38
rs192288574 Chr.11: 3101581-3101581 on Build GRCh38
rs541467160 Chr.11: 3101546-3101546 on Build GRCh38
rs755002878 Chr.11: 3101695-3101695 on Build GRCh38
rs866511896 Chr.11: 3101806-3101806 on Build GRCh38
rs184584968 Chr.11: 3101861-3101861 on Build GRCh38
rs576381081 Chr.11: 3101866-3101866 on Build GRCh38
rs372869408 Chr.11: 3101703-3101703 on Build GRCh38
rs574177127 Chr.11: 3101757-3101757 on Build GRCh38
rs765777488 Chr.11: 3101579-3101579 on Build GRCh38
rs925035957 Chr.11: 3101756-3101756 on Build GRCh38
rs772098005 Chr.11: 3101707-3101707 on Build GRCh38
rs756406146 Chr.11: 3101596-3101596 on Build GRCh38
rs187410433 Chr.11: 3101926-3101926 on Build GRCh38
rs372476916 Chr.11: 3101730-3101730 on Build GRCh38
rs922267855 Chr.11: 3101916-3101916 on Build GRCh38
rs371626565 Chr.11: 3101575-3101575 on Build GRCh38
rs748106984 Chr.11: 3101630-3101630 on Build GRCh38
rs534983941 Chr.11: 3101734-3101734 on Build GRCh38
rs115492535 Chr.11: 3101594-3101594 on Build GRCh38
rs776026352 Chr.11: 3101751-3101751 on Build GRCh38
rs768135547 Chr.11: 3101735-3101735 on Build GRCh38
rs908284242 Chr.11: 3101921-3101921 on Build GRCh38
rs564664384 Chr.11: 3101640-3101640 on Build GRCh38
rs1038625855 Chr.11: 3101828-3101828 on Build GRCh38
rs1024272749 Chr.11: 3101852-3101852 on Build GRCh38
rs915191144 Chr.11: 3101873-3101873 on Build GRCh38
rs777420443 Chr.11: 3101709-3101709 on Build GRCh38
rs554610970 Chr.11: 3101815-3101815 on Build GRCh38
rs979693564 Chr.11: 3101874-3101874 on Build GRCh38
rs767358578 Chr.11: 3101572-3101572 on Build GRCh38
rs543246489 Chr.11: 3101857-3101857 on Build GRCh38
rs963971627 Chr.11: 3101891-3101891 on Build GRCh38
rs188171641 Chr.11: 3101540-3101540 on Build GRCh38
rs935274951 Chr.11: 3101557-3101557 on Build GRCh38
rs373377581 Chr.11: 3101816-3101816 on Build GRCh38
rs780183372 Chr.11: 3101694-3101694 on Build GRCh38
rs939161821 Chr.11: 3101544-3101544 on Build GRCh38
rs778764948 Chr.11: 3101618-3101618 on Build GRCh38
rs746467698 Chr.11: 3101558-3101580 on Build GRCh38
rs933775424 Chr.11: 3101790-3101790 on Build GRCh38
rs572203429 Chr.11: 3101793-3101793 on Build GRCh38
rs368035550 Chr.11: 3101600-3101600 on Build GRCh38
rs759182673 Chr.11: 3101884-3101884 on Build GRCh38
rs1051387167 Chr.11: 3101787-3101787 on Build GRCh38
rs766970695 Chr.11: 3101895-3101895 on Build GRCh38
rs1046843247 Chr.11: 3101919-3101919 on Build GRCh38
rs893484692 Chr.11: 3101559-3101559 on Build GRCh38
rs142720999 Chr.11: 3101539-3101539 on Build GRCh38
rs901539329 Chr.11: 3101778-3101778 on Build GRCh38
rs1024366107 Chr.11: 3101687-3101687 on Build GRCh38
rs374481137 Chr.11: 3101681-3101681 on Build GRCh38
rs922237046 Chr.11: 3101779-3101779 on Build GRCh38
rs749136249 Chr.11: 3101829-3101829 on Build GRCh38
rs770751041 Chr.11: 3101847-3101847 on Build GRCh38
rs542920014 Chr.11: 3101811-3101811 on Build GRCh38
rs1012898754 Chr.11: 3101851-3101851 on Build GRCh38
rs3832785 Chr.11: 3101719-3101719 on Build GRCh38
rs935063502 Chr.11: 3101896-3101896 on Build GRCh38
rs201884402 Chr.11: 3101715-3101715 on Build GRCh38
rs7121251 Chr.11: 3101771-3101771 on Build GRCh38
rs746890676 Chr.11: 3101710-3101710 on Build GRCh38
rs756284238 Chr.11: 3101690-3101690 on Build GRCh38
rs529697196 Chr.11: 3101924-3101924 on Build GRCh38
rs114586944 Chr.11: 3101573-3101573 on Build GRCh38
rs531963326 Chr.11: 3101649-3101649 on Build GRCh38
rs78184020 Chr.11: 3101682-3101682 on Build GRCh38
rs371872013 Chr.11: 3101551-3101551 on Build GRCh38
rs7106225 Chr.11: 3101587-3101587 on Build GRCh38
rs547003210 Chr.11: 3101656-3101656 on Build GRCh38
rs750856165 Chr.11: 3101686-3101686 on Build GRCh38
rs565848604 Chr.11: 3101670-3101670 on Build GRCh38
rs749436104 Chr.11: 3101609-3101609 on Build GRCh38
rs770677466 Chr.11: 3101652-3101652 on Build GRCh38
rs545403789 Chr.11: 3101898-3101898 on Build GRCh38
rs561142041 Chr.11: 3101582-3101582 on Build GRCh38
rs764857249 Chr.11: 3101667-3101667 on Build GRCh38
rs67090912 Chr.11: 3101817-3101817 on Build GRCh38
rs767137987 Chr.11: 3101798-3101798 on Build GRCh38
rs370324105 Chr.11: 3101666-3101666 on Build GRCh38
rs956606858 Chr.11: 3101864-3101864 on Build GRCh38
rs192558043 Chr.11: 3101956-3101956 on Build GRCh38
rs564601734 Chr.11: 3101860-3101860 on Build GRCh38
rs1006123841 Chr.11: 3101823-3101823 on Build GRCh38

View Full Product Details