Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 6169166-6169661 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR52B2
Entrez Gene ID
255725
Gene Name
olfactory receptor family 52 subfamily B member 2
Gene Aliases
OR11-70
UniGene
Hs.690457
Gene Chromosome Location
Chr.11: 6169355-6170326 on Build GRCh38
Transcripts
NM_001004052.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6734721 c.686G>T p.R229L Chr.11: 6169641-6169641 on Build GRCh38
4892553 c.811C>T p.P271S Chr.11: 6169516-6169516 on Build GRCh38
6551679 c.834G>A p.L278L Chr.11: 6169493-6169493 on Build GRCh38
6531052 c.874A>G p.I292V Chr.11: 6169453-6169453 on Build GRCh38
4710013 c.674G>A p.R225Q Chr.11: 6169653-6169653 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1006314395 Chr.11: 6169192-6169192 on Build GRCh38
rs751379420 Chr.11: 6169481-6169481 on Build GRCh38
rs200992980 Chr.11: 6169624-6169624 on Build GRCh38
rs747495626 Chr.11: 6169625-6169625 on Build GRCh38
rs989865688 Chr.11: 6169313-6169313 on Build GRCh38
rs891033280 Chr.11: 6169477-6169477 on Build GRCh38
rs986862752 Chr.11: 6169283-6169283 on Build GRCh38
rs777718049 Chr.11: 6169521-6169521 on Build GRCh38
rs369541312 Chr.11: 6169621-6169621 on Build GRCh38
rs796312611 Chr.11: 6169595-6169595 on Build GRCh38
rs752329635 Chr.11: 6169461-6169461 on Build GRCh38
rs776120152 Chr.11: 6169538-6169538 on Build GRCh38
rs745704728 Chr.11: 6169315-6169315 on Build GRCh38
rs761227088 Chr.11: 6169552-6169552 on Build GRCh38
rs748845579 Chr.11: 6169534-6169534 on Build GRCh38
rs890365683 Chr.11: 6169429-6169429 on Build GRCh38
rs753999259 Chr.11: 6169601-6169601 on Build GRCh38
rs879741470 Chr.11: 6169269-6169269 on Build GRCh38
rs760125194 Chr.11: 6169403-6169403 on Build GRCh38
rs769878658 Chr.11: 6169426-6169426 on Build GRCh38
rs763416747 Chr.11: 6169373-6169373 on Build GRCh38
rs552733421 Chr.11: 6169452-6169452 on Build GRCh38
rs566058261 Chr.11: 6169262-6169262 on Build GRCh38
rs750158940 Chr.11: 6169472-6169472 on Build GRCh38
rs372373798 Chr.11: 6169480-6169480 on Build GRCh38
rs1021317289 Chr.11: 6169545-6169545 on Build GRCh38
rs752612505 Chr.11: 6169590-6169590 on Build GRCh38
rs938039195 Chr.11: 6169194-6169194 on Build GRCh38
rs758234938 Chr.11: 6169317-6169317 on Build GRCh38
rs771354616 Chr.11: 6169455-6169455 on Build GRCh38
rs767344886 Chr.11: 6169582-6169582 on Build GRCh38
rs368388973 Chr.11: 6169306-6169306 on Build GRCh38
rs564653521 Chr.11: 6169381-6169381 on Build GRCh38
rs1027287559 Chr.11: 6169302-6169302 on Build GRCh38
rs893783876 Chr.11: 6169250-6169251 on Build GRCh38
rs770076737 Chr.11: 6169363-6169363 on Build GRCh38
rs748261016 Chr.11: 6169360-6169360 on Build GRCh38
rs756018080 Chr.11: 6169593-6169593 on Build GRCh38
rs368087230 Chr.11: 6169377-6169377 on Build GRCh38
rs376918037 Chr.11: 6169347-6169347 on Build GRCh38
rs762491581 Chr.11: 6169637-6169637 on Build GRCh38
rs753473618 Chr.11: 6169308-6169308 on Build GRCh38
rs528803353 Chr.11: 6169467-6169467 on Build GRCh38
rs531964849 Chr.11: 6169556-6169556 on Build GRCh38
rs576479650 Chr.11: 6169422-6169422 on Build GRCh38
rs776699674 Chr.11: 6169356-6169356 on Build GRCh38
rs574371085 Chr.11: 6169466-6169466 on Build GRCh38
rs765190402 Chr.11: 6169376-6169380 on Build GRCh38
rs750159482 Chr.11: 6169409-6169409 on Build GRCh38
rs760068021 Chr.11: 6169459-6169459 on Build GRCh38
rs977646835 Chr.11: 6169367-6169367 on Build GRCh38
rs1053795134 Chr.11: 6169210-6169210 on Build GRCh38
rs773410789 Chr.11: 6169365-6169365 on Build GRCh38
rs190537696 Chr.11: 6169598-6169598 on Build GRCh38
rs115058946 Chr.11: 6169573-6169573 on Build GRCh38
rs970960915 Chr.11: 6169341-6169341 on Build GRCh38
rs753254284 Chr.11: 6169235-6169235 on Build GRCh38
rs145347029 Chr.11: 6169263-6169263 on Build GRCh38
rs147668114 Chr.11: 6169404-6169404 on Build GRCh38
rs763646763 Chr.11: 6169596-6169596 on Build GRCh38
rs1017978449 Chr.11: 6169288-6169288 on Build GRCh38
rs372561472 Chr.11: 6169488-6169488 on Build GRCh38
rs765854149 Chr.11: 6169272-6169274 on Build GRCh38
rs993987651 Chr.11: 6169454-6169454 on Build GRCh38
rs189493020 Chr.11: 6169256-6169256 on Build GRCh38
rs114263793 Chr.11: 6169524-6169524 on Build GRCh38
rs180683313 Chr.11: 6169309-6169309 on Build GRCh38
rs989593054 Chr.11: 6169611-6169611 on Build GRCh38
rs771301186 Chr.11: 6169525-6169525 on Build GRCh38
rs1037806449 Chr.11: 6169433-6169433 on Build GRCh38
rs772681626 Chr.11: 6169528-6169528 on Build GRCh38
rs746886817 Chr.11: 6169420-6169420 on Build GRCh38
rs77344022 Chr.11: 6169620-6169620 on Build GRCh38
rs1033866524 Chr.11: 6169594-6169594 on Build GRCh38
rs758136385 Chr.11: 6169554-6169554 on Build GRCh38
rs926745744 Chr.11: 6169374-6169374 on Build GRCh38
rs774867876 Chr.11: 6169456-6169456 on Build GRCh38
rs762547194 Chr.11: 6169564-6169564 on Build GRCh38
rs994059756 Chr.11: 6169259-6169259 on Build GRCh38
rs532694878 Chr.11: 6169617-6169617 on Build GRCh38
rs751352867 Chr.11: 6169565-6169565 on Build GRCh38
rs747080555 Chr.11: 6169324-6169324 on Build GRCh38
rs866011026 Chr.11: 6169557-6169557 on Build GRCh38
rs772615448 Chr.11: 6169559-6169559 on Build GRCh38
rs368973280 Chr.11: 6169445-6169445 on Build GRCh38
rs186059999 Chr.11: 6169423-6169423 on Build GRCh38
rs754551600 Chr.11: 6169603-6169603 on Build GRCh38
rs764572397 Chr.11: 6169385-6169385 on Build GRCh38
rs778518510 Chr.11: 6169314-6169314 on Build GRCh38
rs558250641 Chr.11: 6169418-6169418 on Build GRCh38
rs372389388 Chr.11: 6169320-6169320 on Build GRCh38
rs764786355 Chr.11: 6169468-6169468 on Build GRCh38
rs756281772 Chr.11: 6169519-6169519 on Build GRCh38
rs746270760 Chr.11: 6169526-6169526 on Build GRCh38
rs377074219 Chr.11: 6169633-6169633 on Build GRCh38
rs376980001 Chr.11: 6169361-6169361 on Build GRCh38
rs758274575 Chr.11: 6169410-6169410 on Build GRCh38
rs768795331 Chr.11: 6169337-6169337 on Build GRCh38
rs201902095 Chr.11: 6169405-6169405 on Build GRCh38
rs762603066 Chr.11: 6169473-6169473 on Build GRCh38
rs757519505 Chr.11: 6169610-6169610 on Build GRCh38
rs1037732477 Chr.11: 6169634-6169634 on Build GRCh38
rs372147559 Chr.11: 6169257-6169257 on Build GRCh38
rs766050616 Chr.11: 6169566-6169566 on Build GRCh38
rs773539978 Chr.11: 6169446-6169446 on Build GRCh38
rs768165792 Chr.11: 6169465-6169465 on Build GRCh38
rs752699810 Chr.11: 6169511-6169511 on Build GRCh38

View Full Product Details