Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 33085754-33086290 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSTF3
Entrez Gene ID
1479
Gene Name
cleavage stimulation factor subunit 3
Gene Aliases
CSTF-77
UniGene
Hs.44402
Gene Chromosome Location
Chr.11: 33084584-33161491 on Build GRCh38
Transcripts
NM_001326.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6461770 c.1891-5A>T p.? Chr.11: 33085778-33085778 on Build GRCh38
428925 c.1840C>T p.P614S Chr.11: 33085945-33085945 on Build GRCh38
5017661 c.1876_1878delCCT p.P626delP Chr.11: 33085907-33085909 on Build GRCh38
7246227 c.1810C>G p.P604A Chr.11: 33085975-33085975 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs775155502 Chr.11: 33085961-33085961 on Build GRCh38
rs775081786 Chr.11: 33086180-33086180 on Build GRCh38
rs953665442 Chr.11: 33086241-33086242 on Build GRCh38
rs1055282446 Chr.11: 33086082-33086082 on Build GRCh38
rs776599477 Chr.11: 33085980-33085980 on Build GRCh38
rs1000963113 Chr.11: 33086204-33086204 on Build GRCh38
rs753189022 Chr.11: 33085875-33085875 on Build GRCh38
rs757985828 Chr.11: 33086029-33086029 on Build GRCh38
rs760546736 Chr.11: 33085814-33085814 on Build GRCh38
rs563376018 Chr.11: 33085975-33085975 on Build GRCh38
rs762990046 Chr.11: 33085994-33085994 on Build GRCh38
rs770702577 Chr.11: 33085785-33085785 on Build GRCh38
rs1007095295 Chr.11: 33085903-33085903 on Build GRCh38
rs114705753 Chr.11: 33086140-33086140 on Build GRCh38
rs909093698 Chr.11: 33086170-33086170 on Build GRCh38
rs759688592 Chr.11: 33086004-33086004 on Build GRCh38
rs750028112 Chr.11: 33086027-33086027 on Build GRCh38
rs771844785 Chr.11: 33085945-33085945 on Build GRCh38
rs938250323 Chr.11: 33086044-33086044 on Build GRCh38
rs755665819 Chr.11: 33086001-33086002 on Build GRCh38
rs397718693 Chr.11: 33086148-33086149 on Build GRCh38
rs373751924 Chr.11: 33085818-33085818 on Build GRCh38
rs764273677 Chr.11: 33085815-33085820 on Build GRCh38
rs1042566844 Chr.11: 33086185-33086185 on Build GRCh38
rs985123043 Chr.11: 33086162-33086162 on Build GRCh38
rs781338298 Chr.11: 33085877-33085878 on Build GRCh38
rs765280190 Chr.11: 33086161-33086161 on Build GRCh38
rs1051192650 Chr.11: 33085872-33085872 on Build GRCh38
rs536921512 Chr.11: 33086027-33086032 on Build GRCh38
rs773280400 Chr.11: 33085844-33085844 on Build GRCh38
rs10631653 Chr.11: 33086146-33086147 on Build GRCh38
rs902258217 Chr.11: 33086198-33086198 on Build GRCh38
rs1031994974 Chr.11: 33086238-33086238 on Build GRCh38
rs940371547 Chr.11: 33086252-33086252 on Build GRCh38
rs369652270 Chr.11: 33085836-33085836 on Build GRCh38
rs755331713 Chr.11: 33085865-33085865 on Build GRCh38
rs749954314 Chr.11: 33085999-33086002 on Build GRCh38
rs754432948 Chr.11: 33086020-33086020 on Build GRCh38
rs779783320 Chr.11: 33085998-33086002 on Build GRCh38
rs866854863 Chr.11: 33086126-33086126 on Build GRCh38
rs766478548 Chr.11: 33085995-33085995 on Build GRCh38
rs759497477 Chr.11: 33086010-33086010 on Build GRCh38
rs778674552 Chr.11: 33085920-33085920 on Build GRCh38
rs367945525 Chr.11: 33085940-33085940 on Build GRCh38
rs773457140 Chr.11: 33086007-33086007 on Build GRCh38
rs890061608 Chr.11: 33085889-33085889 on Build GRCh38
rs373168530 Chr.11: 33086025-33086025 on Build GRCh38
rs751648171 Chr.11: 33085820-33085822 on Build GRCh38
rs992019265 Chr.11: 33086096-33086096 on Build GRCh38
rs371781850 Chr.11: 33085888-33085888 on Build GRCh38
rs909476702 Chr.11: 33086035-33086035 on Build GRCh38
rs771801027 Chr.11: 33085807-33085807 on Build GRCh38
rs763078108 Chr.11: 33085848-33085848 on Build GRCh38
rs768469349 Chr.11: 33085976-33085976 on Build GRCh38
rs758441071 Chr.11: 33086257-33086257 on Build GRCh38
rs1006829917 Chr.11: 33086246-33086246 on Build GRCh38
rs145066590 Chr.11: 33086260-33086260 on Build GRCh38
rs374097467 Chr.11: 33085898-33085898 on Build GRCh38
rs375678885 Chr.11: 33086147-33086148 on Build GRCh38
rs745746853 Chr.11: 33085878-33085879 on Build GRCh38
rs116623504 Chr.11: 33086018-33086018 on Build GRCh38
rs545969010 Chr.11: 33086091-33086091 on Build GRCh38
rs960750766 Chr.11: 33086085-33086085 on Build GRCh38
rs200947426 Chr.11: 33086149-33086149 on Build GRCh38
rs751821541 Chr.11: 33085856-33085856 on Build GRCh38
rs774200981 Chr.11: 33085786-33085786 on Build GRCh38
rs750560129 Chr.11: 33086201-33086201 on Build GRCh38
rs767794182 Chr.11: 33085870-33085870 on Build GRCh38
rs756068003 Chr.11: 33085903-33085906 on Build GRCh38
rs767853941 Chr.11: 33086013-33086013 on Build GRCh38
rs762874129 Chr.11: 33086190-33086190 on Build GRCh38
rs748790656 Chr.11: 33085896-33085896 on Build GRCh38
rs757348319 Chr.11: 33085866-33085867 on Build GRCh38
rs869238886 Chr.11: 33086149-33086150 on Build GRCh38
rs756559528 Chr.11: 33086017-33086017 on Build GRCh38
rs778490694 Chr.11: 33085879-33085879 on Build GRCh38
rs1031521743 Chr.11: 33086067-33086067 on Build GRCh38
rs774510346 Chr.11: 33086002-33086002 on Build GRCh38
rs866638256 Chr.11: 33085943-33085943 on Build GRCh38
rs370927014 Chr.11: 33085799-33085799 on Build GRCh38
rs369970743 Chr.11: 33086023-33086023 on Build GRCh38
rs367546306 Chr.11: 33085850-33085850 on Build GRCh38

View Full Product Details