Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 34160577-34161067 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABTB2
Entrez Gene ID
25841
Gene Name
ankyrin repeat and BTB domain containing 2
Gene Aliases
ABTB2A, BTBD22
UniGene
Hs.23361
Gene Chromosome Location
Chr.11: 34150987-34358008 on Build GRCh38
Transcripts
NM_145804.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5838465 c.1727C>T p.S576L Chr.11: 34161015-34161015 on Build GRCh38
3710171 c.2248G>A p.E750K Chr.11: 34161052-34161052 on Build GRCh38
4886116 c.1798G>A p.E600K Chr.11: 34160944-34160944 on Build GRCh38
415619 c.1797C>T p.T599T Chr.11: 34160945-34160945 on Build GRCh38
1353652 c.1774_1776delGAG p.E592delE Chr.11: 34160966-34160968 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753286619 Chr.11: 34161021-34161021 on Build GRCh38
rs548995014 Chr.11: 34160659-34160660 on Build GRCh38
rs769791037 Chr.11: 34160869-34160869 on Build GRCh38
rs905364888 Chr.11: 34160722-34160722 on Build GRCh38
rs1017007638 Chr.11: 34160897-34160900 on Build GRCh38
rs773380868 Chr.11: 34160875-34160875 on Build GRCh38
rs529693430 Chr.11: 34160640-34160640 on Build GRCh38
rs935057566 Chr.11: 34160649-34160649 on Build GRCh38
rs761069466 Chr.11: 34161004-34161004 on Build GRCh38
rs757128957 Chr.11: 34160828-34160839 on Build GRCh38
rs772458230 Chr.11: 34160995-34160995 on Build GRCh38
rs763063010 Chr.11: 34160885-34160885 on Build GRCh38
rs756849874 Chr.11: 34160923-34160923 on Build GRCh38
rs372500990 Chr.11: 34160606-34160607 on Build GRCh38
rs371485631 Chr.11: 34160851-34160851 on Build GRCh38
rs544758403 Chr.11: 34160914-34160914 on Build GRCh38
rs1043936133 Chr.11: 34160713-34160713 on Build GRCh38
rs1019142675 Chr.11: 34160798-34160798 on Build GRCh38
rs200189695 Chr.11: 34160625-34160626 on Build GRCh38
rs759901270 Chr.11: 34160893-34160893 on Build GRCh38
rs373904135 Chr.11: 34161035-34161035 on Build GRCh38
rs537094006 Chr.11: 34160850-34160850 on Build GRCh38
rs995696030 Chr.11: 34160993-34160993 on Build GRCh38
rs1045810405 Chr.11: 34160924-34160924 on Build GRCh38
rs753440571 Chr.11: 34160919-34160919 on Build GRCh38
rs561058759 Chr.11: 34160977-34160977 on Build GRCh38
rs775876453 Chr.11: 34160999-34160999 on Build GRCh38
rs372294847 Chr.11: 34160985-34160985 on Build GRCh38
rs530583526 Chr.11: 34160996-34160996 on Build GRCh38
rs756259233 Chr.11: 34160669-34160670 on Build GRCh38
rs956401445 Chr.11: 34161010-34161010 on Build GRCh38
rs966151779 Chr.11: 34161022-34161022 on Build GRCh38
rs901063949 Chr.11: 34160809-34160812 on Build GRCh38
rs374518748 Chr.11: 34160960-34160960 on Build GRCh38
rs767544640 Chr.11: 34160966-34160968 on Build GRCh38
rs71037401 Chr.11: 34160706-34160707 on Build GRCh38
rs201712306 Chr.11: 34160785-34160785 on Build GRCh38
rs569816968 Chr.11: 34160661-34160661 on Build GRCh38
rs761720549 Chr.11: 34160960-34160962 on Build GRCh38
rs1025742582 Chr.11: 34160786-34160786 on Build GRCh38
rs552805477 Chr.11: 34160667-34160667 on Build GRCh38
rs370214314 Chr.11: 34160959-34160959 on Build GRCh38
rs774571777 Chr.11: 34160992-34160992 on Build GRCh38
rs1004574521 Chr.11: 34160796-34160796 on Build GRCh38
rs1001978300 Chr.11: 34160627-34160627 on Build GRCh38
rs942443702 Chr.11: 34161029-34161034 on Build GRCh38
rs965738454 Chr.11: 34160820-34160820 on Build GRCh38
rs549881982 Chr.11: 34160654-34160654 on Build GRCh38
rs566211081 Chr.11: 34160772-34160772 on Build GRCh38
rs373513180 Chr.11: 34160868-34160868 on Build GRCh38
rs368954407 Chr.11: 34161030-34161030 on Build GRCh38
rs370914114 Chr.11: 34160895-34160895 on Build GRCh38
rs942474715 Chr.11: 34160778-34160778 on Build GRCh38
rs764721975 Chr.11: 34161014-34161014 on Build GRCh38
rs868582683 Chr.11: 34160956-34160956 on Build GRCh38
rs74789123 Chr.11: 34161023-34161023 on Build GRCh38
rs114499809 Chr.11: 34160910-34160910 on Build GRCh38
rs923617372 Chr.11: 34160628-34160628 on Build GRCh38
rs749450917 Chr.11: 34160976-34160976 on Build GRCh38
rs375347114 Chr.11: 34160840-34160840 on Build GRCh38
rs563323500 Chr.11: 34161044-34161044 on Build GRCh38
rs898706166 Chr.11: 34160773-34160773 on Build GRCh38
rs1002285826 Chr.11: 34160743-34160743 on Build GRCh38
rs746912210 Chr.11: 34160951-34160951 on Build GRCh38
rs141845210 Chr.11: 34161029-34161029 on Build GRCh38
rs138074082 Chr.11: 34160777-34160777 on Build GRCh38
rs61016556 Chr.11: 34160706-34160706 on Build GRCh38
rs771241537 Chr.11: 34160889-34160889 on Build GRCh38
rs1030649434 Chr.11: 34161008-34161008 on Build GRCh38
rs554997980 Chr.11: 34160789-34160789 on Build GRCh38
rs558739112 Chr.11: 34160944-34160944 on Build GRCh38
rs151308415 Chr.11: 34160994-34160994 on Build GRCh38
rs142391398 Chr.11: 34160950-34160950 on Build GRCh38
rs532710740 Chr.11: 34160666-34160666 on Build GRCh38
rs774459656 Chr.11: 34160915-34160917 on Build GRCh38
rs758161018 Chr.11: 34161036-34161036 on Build GRCh38
rs755336228 Chr.11: 34160604-34160604 on Build GRCh38
rs768687496 Chr.11: 34160957-34160957 on Build GRCh38
rs148434413 Chr.11: 34160929-34160929 on Build GRCh38
rs753226333 Chr.11: 34160900-34160900 on Build GRCh38
rs866618948 Chr.11: 34160670-34160670 on Build GRCh38
rs981777527 Chr.11: 34160605-34160606 on Build GRCh38
rs763484799 Chr.11: 34160915-34160915 on Build GRCh38
rs774572893 Chr.11: 34160891-34160891 on Build GRCh38

View Full Product Details