Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 45914212-45914718 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PEX16
Entrez Gene ID
9409
Gene Name
peroxisomal biogenesis factor 16
Gene Aliases
PBD8A, PBD8B
UniGene
Hs.100915
Gene Chromosome Location
Chr.11: 45909669-45918123 on Build GRCh38
Transcripts
NM_004813.2 NM_057174.2 XM_011520474.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4425377 c.566G>A p.W189* Chr.11: 45914444-45914444 on Build GRCh38
5942428 c.461-8C>T p.? Chr.11: 45914692-45914692 on Build GRCh38
4032916 c.462T>C p.D154D Chr.11: 45914683-45914683 on Build GRCh38
927087 c.679C>T p.R227W Chr.11: 45914331-45914331 on Build GRCh38
4032911 c.592C>T p.R198W Chr.11: 45914418-45914418 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763001456 Chr.11: 45914303-45914303 on Build GRCh38
rs201706413 Chr.11: 45914358-45914358 on Build GRCh38
rs775473862 Chr.11: 45914636-45914636 on Build GRCh38
rs372347595 Chr.11: 45914356-45914356 on Build GRCh38
rs765466211 Chr.11: 45914441-45914441 on Build GRCh38
rs537046144 Chr.11: 45914401-45914401 on Build GRCh38
rs767299660 Chr.11: 45914424-45914424 on Build GRCh38
rs748963153 Chr.11: 45914488-45914488 on Build GRCh38
rs770685291 Chr.11: 45914490-45914490 on Build GRCh38
rs35874263 Chr.11: 45914407-45914407 on Build GRCh38
rs751814237 Chr.11: 45914456-45914456 on Build GRCh38
rs199552721 Chr.11: 45914254-45914254 on Build GRCh38
rs373361792 Chr.11: 45914562-45914562 on Build GRCh38
rs766753438 Chr.11: 45914449-45914449 on Build GRCh38
rs200522601 Chr.11: 45914353-45914353 on Build GRCh38
rs759501669 Chr.11: 45914422-45914422 on Build GRCh38
rs971604129 Chr.11: 45914253-45914254 on Build GRCh38
rs768523500 Chr.11: 45914652-45914652 on Build GRCh38
rs761839307 Chr.11: 45914432-45914432 on Build GRCh38
rs751488634 Chr.11: 45914378-45914378 on Build GRCh38
rs36084848 Chr.11: 45914449-45914449 on Build GRCh38
rs985796980 Chr.11: 45914517-45914517 on Build GRCh38
rs762887312 Chr.11: 45914412-45914412 on Build GRCh38
rs760707150 Chr.11: 45914380-45914380 on Build GRCh38
rs375515674 Chr.11: 45914394-45914394 on Build GRCh38
rs959280173 Chr.11: 45914426-45914426 on Build GRCh38
rs918637787 Chr.11: 45914647-45914647 on Build GRCh38
rs553575684 Chr.11: 45914530-45914530 on Build GRCh38
rs779040845 Chr.11: 45914392-45914392 on Build GRCh38
rs936507537 Chr.11: 45914620-45914620 on Build GRCh38
rs140695185 Chr.11: 45914684-45914684 on Build GRCh38
rs771831186 Chr.11: 45914618-45914618 on Build GRCh38
rs775355323 Chr.11: 45914495-45914495 on Build GRCh38
rs774875396 Chr.11: 45914246-45914246 on Build GRCh38
rs565706311 Chr.11: 45914381-45914381 on Build GRCh38
rs780865953 Chr.11: 45914402-45914402 on Build GRCh38
rs754024503 Chr.11: 45914330-45914330 on Build GRCh38
rs763840804 Chr.11: 45914305-45914305 on Build GRCh38
rs769516596 Chr.11: 45914365-45914365 on Build GRCh38
rs771211079 Chr.11: 45914660-45914660 on Build GRCh38
rs763683831 Chr.11: 45914278-45914278 on Build GRCh38
rs551455430 Chr.11: 45914569-45914569 on Build GRCh38
rs1024606739 Chr.11: 45914425-45914425 on Build GRCh38
rs3802758 Chr.11: 45914484-45914484 on Build GRCh38
rs147332976 Chr.11: 45914646-45914646 on Build GRCh38
rs755666423 Chr.11: 45914472-45914472 on Build GRCh38
rs753312075 Chr.11: 45914693-45914693 on Build GRCh38
rs758739455 Chr.11: 45914398-45914398 on Build GRCh38
rs756938526 Chr.11: 45914592-45914592 on Build GRCh38
rs762905186 Chr.11: 45914659-45914659 on Build GRCh38
rs778389525 Chr.11: 45914604-45914604 on Build GRCh38
rs754114296 Chr.11: 45914382-45914382 on Build GRCh38
rs771628786 Chr.11: 45914240-45914240 on Build GRCh38
rs773347735 Chr.11: 45914653-45914653 on Build GRCh38
rs201173264 Chr.11: 45914387-45914387 on Build GRCh38
rs775951837 Chr.11: 45914299-45914299 on Build GRCh38
rs753220876 Chr.11: 45914570-45914570 on Build GRCh38
rs769604713 Chr.11: 45914410-45914410 on Build GRCh38
rs1023213575 Chr.11: 45914243-45914243 on Build GRCh38
rs763344686 Chr.11: 45914372-45914372 on Build GRCh38
rs142611968 Chr.11: 45914283-45914283 on Build GRCh38
rs770726448 Chr.11: 45914415-45914415 on Build GRCh38
rs779235562 Chr.11: 45914337-45914337 on Build GRCh38
rs966472584 Chr.11: 45914446-45914446 on Build GRCh38
rs144063598 Chr.11: 45914467-45914467 on Build GRCh38
rs777404753 Chr.11: 45914579-45914579 on Build GRCh38
rs941343043 Chr.11: 45914605-45914605 on Build GRCh38
rs772953974 Chr.11: 45914411-45914411 on Build GRCh38
rs760163684 Chr.11: 45914273-45914273 on Build GRCh38
rs746173483 Chr.11: 45914338-45914338 on Build GRCh38
rs933338823 Chr.11: 45914523-45914523 on Build GRCh38
rs1026595479 Chr.11: 45914464-45914464 on Build GRCh38
rs746365663 Chr.11: 45914238-45914238 on Build GRCh38
rs368365098 Chr.11: 45914344-45914344 on Build GRCh38
rs745581860 Chr.11: 45914494-45914494 on Build GRCh38
rs766445843 Chr.11: 45914554-45914554 on Build GRCh38
rs766275337 Chr.11: 45914304-45914304 on Build GRCh38
rs766253527 Chr.11: 45914376-45914376 on Build GRCh38
rs368553807 Chr.11: 45914400-45914400 on Build GRCh38
rs886048327 Chr.11: 45914605-45914605 on Build GRCh38
rs750614956 Chr.11: 45914447-45914447 on Build GRCh38
rs201846257 Chr.11: 45914375-45914375 on Build GRCh38
rs111379584 Chr.11: 45914295-45914295 on Build GRCh38
rs755516862 Chr.11: 45914465-45914465 on Build GRCh38
rs750448293 Chr.11: 45914302-45914302 on Build GRCh38
rs756167148 Chr.11: 45914324-45914324 on Build GRCh38
rs773483384 Chr.11: 45914518-45914518 on Build GRCh38
rs371744103 Chr.11: 45914519-45914519 on Build GRCh38
rs751889301 Chr.11: 45914491-45914491 on Build GRCh38
rs1053995048 Chr.11: 45914637-45914637 on Build GRCh38
rs774416603 Chr.11: 45914670-45914670 on Build GRCh38
rs1038841857 Chr.11: 45914374-45914374 on Build GRCh38
rs199956751 Chr.11: 45914654-45914654 on Build GRCh38
rs943381202 Chr.11: 45914327-45914327 on Build GRCh38
rs993858392 Chr.11: 45914691-45914691 on Build GRCh38
rs761544516 Chr.11: 45914359-45914359 on Build GRCh38
rs761929034 Chr.11: 45914513-45914513 on Build GRCh38
rs772570363 Chr.11: 45914343-45914343 on Build GRCh38
rs61752117 Chr.11: 45914619-45914619 on Build GRCh38
rs573492839 Chr.11: 45914560-45914560 on Build GRCh38
rs748684198 Chr.11: 45914582-45914582 on Build GRCh38
rs757682196 Chr.11: 45914335-45914335 on Build GRCh38
rs909291081 Chr.11: 45914480-45914480 on Build GRCh38
rs773150003 Chr.11: 45914367-45914367 on Build GRCh38
rs768859616 Chr.11: 45914404-45914405 on Build GRCh38
rs777904359 Chr.11: 45914326-45914326 on Build GRCh38
rs375771098 Chr.11: 45914262-45914262 on Build GRCh38
rs768190501 Chr.11: 45914563-45914563 on Build GRCh38
rs141267636 Chr.11: 45914435-45914435 on Build GRCh38
rs375900051 Chr.11: 45914468-45914468 on Build GRCh38
rs557365860 Chr.11: 45914427-45914427 on Build GRCh38
rs774651831 Chr.11: 45914405-45914407 on Build GRCh38
rs755750023 Chr.11: 45914572-45914572 on Build GRCh38
rs765119035 Chr.11: 45914299-45914299 on Build GRCh38
rs759988037 Chr.11: 45914679-45914679 on Build GRCh38
rs747599205 Chr.11: 45914403-45914403 on Build GRCh38
rs774218187 Chr.11: 45914418-45914418 on Build GRCh38
rs61752117 Chr.11: 45914619-45914619 on Build GRCh38
rs765298949 Chr.11: 45914514-45914514 on Build GRCh38
rs745696605 Chr.11: 45914616-45914616 on Build GRCh38

View Full Product Details