Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 60070791-60071299 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MS4A3
Entrez Gene ID
932
Gene Name
membrane spanning 4-domains A3
Gene Aliases
CD20L, HTM4
UniGene
Hs.99960
Gene Chromosome Location
Chr.11: 60056628-60071116 on Build GRCh38
Transcripts
NM_001031666.1 NM_001031809.1 NM_006138.4 XM_011545363.2

SNP Detail

SNP ID Chromosome Location
rs1053922897 Chr.11: 60071053-60071055 on Build GRCh38
rs182078408 Chr.11: 60070830-60070830 on Build GRCh38
rs1004542513 Chr.11: 60071115-60071115 on Build GRCh38
rs144568181 Chr.11: 60071008-60071008 on Build GRCh38
rs1055143581 Chr.11: 60070942-60070942 on Build GRCh38
rs78909904 Chr.11: 60071124-60071124 on Build GRCh38
rs907913843 Chr.11: 60071063-60071063 on Build GRCh38
rs537332083 Chr.11: 60071051-60071051 on Build GRCh38
rs963288838 Chr.11: 60071132-60071132 on Build GRCh38
rs963579869 Chr.11: 60070838-60070838 on Build GRCh38
rs1034158058 Chr.11: 60071161-60071161 on Build GRCh38
rs891522678 Chr.11: 60071271-60071271 on Build GRCh38
rs1002577022 Chr.11: 60070840-60070840 on Build GRCh38
rs969805652 Chr.11: 60070839-60070839 on Build GRCh38
rs1044241028 Chr.11: 60071138-60071138 on Build GRCh38
rs368642032 Chr.11: 60070868-60070868 on Build GRCh38
rs924195443 Chr.11: 60071256-60071256 on Build GRCh38
rs1037680394 Chr.11: 60071069-60071069 on Build GRCh38
rs114743176 Chr.11: 60070995-60070995 on Build GRCh38
rs988469551 Chr.11: 60070860-60070860 on Build GRCh38
rs868744724 Chr.11: 60071226-60071226 on Build GRCh38
rs1030017789 Chr.11: 60071067-60071067 on Build GRCh38
rs547246948 Chr.11: 60071004-60071004 on Build GRCh38
rs536659138 Chr.11: 60070991-60070991 on Build GRCh38
rs968982184 Chr.11: 60070891-60070891 on Build GRCh38
rs900273232 Chr.11: 60071055-60071055 on Build GRCh38
rs959463546 Chr.11: 60071177-60071177 on Build GRCh38
rs474951 Chr.11: 60071148-60071148 on Build GRCh38
rs368836552 Chr.11: 60071269-60071269 on Build GRCh38
rs955909026 Chr.11: 60070854-60070854 on Build GRCh38
rs79224552 Chr.11: 60070909-60070909 on Build GRCh38
rs890146682 Chr.11: 60071081-60071081 on Build GRCh38
rs757496647 Chr.11: 60071068-60071068 on Build GRCh38
rs938045604 Chr.11: 60070877-60070877 on Build GRCh38
rs186843464 Chr.11: 60071249-60071249 on Build GRCh38
rs974930025 Chr.11: 60070950-60070950 on Build GRCh38
rs902761344 Chr.11: 60071040-60071040 on Build GRCh38
rs990927492 Chr.11: 60071188-60071188 on Build GRCh38
rs534487623 Chr.11: 60071220-60071220 on Build GRCh38
rs752183628 Chr.11: 60070853-60070853 on Build GRCh38
rs1002620854 Chr.11: 60071235-60071235 on Build GRCh38
rs781614384 Chr.11: 60071116-60071116 on Build GRCh38
rs764469682 Chr.11: 60070989-60070989 on Build GRCh38

View Full Product Details