Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62242140-62242634 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCGB1D2
Entrez Gene ID
10647
Gene Name
secretoglobin family 1D member 2
Gene Aliases
LIPB, LPHB, LPNB
UniGene
Hs.204096
Gene Chromosome Location
Chr.11: 62242252-62244808 on Build GRCh38
Transcripts
NM_006551.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4477979 c.21C>T p.L7L Chr.11: 62242328-62242328 on Build GRCh38
5926803 c.55G>A p.A19T Chr.11: 62242362-62242362 on Build GRCh38
5572407 c.49T>A p.Y17N Chr.11: 62242356-62242356 on Build GRCh38
429430 c.42C>T p.L14L Chr.11: 62242349-62242349 on Build GRCh38
1561719 c.54delG p.A19fs*14 Chr.11: 62242361-62242361 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs936856335 Chr.11: 62242175-62242175 on Build GRCh38
rs923608728 Chr.11: 62242448-62242448 on Build GRCh38
rs748661408 Chr.11: 62242387-62242387 on Build GRCh38
rs549694739 Chr.11: 62242545-62242545 on Build GRCh38
rs755120089 Chr.11: 62242356-62242356 on Build GRCh38
rs770172788 Chr.11: 62242294-62242294 on Build GRCh38
rs774833628 Chr.11: 62242380-62242380 on Build GRCh38
rs945845954 Chr.11: 62242562-62242562 on Build GRCh38
rs139214011 Chr.11: 62242313-62242313 on Build GRCh38
rs756251405 Chr.11: 62242363-62242363 on Build GRCh38
rs776956717 Chr.11: 62242539-62242539 on Build GRCh38
rs765046837 Chr.11: 62242330-62242330 on Build GRCh38
rs1008992799 Chr.11: 62242546-62242546 on Build GRCh38
rs932125158 Chr.11: 62242483-62242483 on Build GRCh38
rs936805423 Chr.11: 62242544-62242544 on Build GRCh38
rs201001396 Chr.11: 62242329-62242329 on Build GRCh38
rs773414232 Chr.11: 62242349-62242349 on Build GRCh38
rs759156393 Chr.11: 62242327-62242327 on Build GRCh38
rs372512437 Chr.11: 62242270-62242270 on Build GRCh38
rs147775921 Chr.11: 62242598-62242598 on Build GRCh38
rs145793664 Chr.11: 62242269-62242269 on Build GRCh38
rs557648816 Chr.11: 62242565-62242565 on Build GRCh38
rs1041417229 Chr.11: 62242255-62242255 on Build GRCh38
rs368641266 Chr.11: 62242389-62242389 on Build GRCh38
rs1050635016 Chr.11: 62242514-62242514 on Build GRCh38
rs568644477 Chr.11: 62242512-62242512 on Build GRCh38
rs201846552 Chr.11: 62242314-62242314 on Build GRCh38
rs758287827 Chr.11: 62242335-62242335 on Build GRCh38
rs2232947 Chr.11: 62242319-62242319 on Build GRCh38
rs775289118 Chr.11: 62242414-62242414 on Build GRCh38
rs754070972 Chr.11: 62242315-62242315 on Build GRCh38
rs1054038124 Chr.11: 62242477-62242477 on Build GRCh38
rs749685773 Chr.11: 62242306-62242306 on Build GRCh38
rs780278667 Chr.11: 62242370-62242370 on Build GRCh38
rs892892792 Chr.11: 62242194-62242194 on Build GRCh38
rs545872221 Chr.11: 62242610-62242610 on Build GRCh38
rs746139518 Chr.11: 62242277-62242277 on Build GRCh38
rs759114892 Chr.11: 62242400-62242400 on Build GRCh38
rs769012037 Chr.11: 62242379-62242379 on Build GRCh38
rs945702280 Chr.11: 62242223-62242223 on Build GRCh38
rs764058216 Chr.11: 62242339-62242339 on Build GRCh38
rs761756312 Chr.11: 62242604-62242604 on Build GRCh38
rs3737534 Chr.11: 62242453-62242453 on Build GRCh38
rs752497174 Chr.11: 62242333-62242333 on Build GRCh38
rs1620121 Chr.11: 62242481-62242481 on Build GRCh38
rs1054100875 Chr.11: 62242193-62242193 on Build GRCh38
rs565617857 Chr.11: 62242422-62242422 on Build GRCh38
rs1041759373 Chr.11: 62242571-62242571 on Build GRCh38
rs935529041 Chr.11: 62242466-62242466 on Build GRCh38
rs777882485 Chr.11: 62242257-62242257 on Build GRCh38
rs112301264 Chr.11: 62242296-62242296 on Build GRCh38
rs769104183 Chr.11: 62242307-62242307 on Build GRCh38
rs755825022 Chr.11: 62242259-62242259 on Build GRCh38
rs139406960 Chr.11: 62242341-62242341 on Build GRCh38
rs955110143 Chr.11: 62242447-62242447 on Build GRCh38
rs914299124 Chr.11: 62242554-62242554 on Build GRCh38
rs776041012 Chr.11: 62242395-62242395 on Build GRCh38
rs1030358274 Chr.11: 62242435-62242435 on Build GRCh38
rs141510840 Chr.11: 62242318-62242318 on Build GRCh38
rs376107340 Chr.11: 62242371-62242371 on Build GRCh38
rs570221087 Chr.11: 62242394-62242394 on Build GRCh38
rs370395063 Chr.11: 62242340-62242340 on Build GRCh38
rs2232948 Chr.11: 62242411-62242411 on Build GRCh38

View Full Product Details