Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62270791-62271307 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCGB2A2
Entrez Gene ID
4250
Gene Name
secretoglobin family 2A member 2
Gene Aliases
MGB1, UGB2
UniGene
Hs.46452
Gene Chromosome Location
Chr.11: 62270130-62273160 on Build GRCh38
Transcripts
NM_002411.3 XM_005274005.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
339580 c.224G>A p.S75N Chr.11: 62271049-62271049 on Build GRCh38
5699719 c.225C>A p.S75R Chr.11: 62271050-62271050 on Build GRCh38
5821338 c.152A>T p.D51V Chr.11: 62270977-62270977 on Build GRCh38
5373054 c.181G>A p.E61K Chr.11: 62271006-62271006 on Build GRCh38
5543419 c.78G>A p.E26E Chr.11: 62270903-62270903 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752656752 Chr.11: 62271006-62271006 on Build GRCh38
rs1010865158 Chr.11: 62271091-62271091 on Build GRCh38
rs201000878 Chr.11: 62271034-62271034 on Build GRCh38
rs771543066 Chr.11: 62270979-62270979 on Build GRCh38
rs748746430 Chr.11: 62271184-62271184 on Build GRCh38
rs372541368 Chr.11: 62271067-62271067 on Build GRCh38
rs368089516 Chr.11: 62271084-62271084 on Build GRCh38
rs745401378 Chr.11: 62271209-62271209 on Build GRCh38
rs144639322 Chr.11: 62271103-62271103 on Build GRCh38
rs143763665 Chr.11: 62270885-62270885 on Build GRCh38
rs114921537 Chr.11: 62271035-62271035 on Build GRCh38
rs756292616 Chr.11: 62271127-62271127 on Build GRCh38
rs568889883 Chr.11: 62271124-62271124 on Build GRCh38
rs143019268 Chr.11: 62270889-62270889 on Build GRCh38
rs774916813 Chr.11: 62271203-62271203 on Build GRCh38
rs79158868 Chr.11: 62271029-62271029 on Build GRCh38
rs770538286 Chr.11: 62271088-62271088 on Build GRCh38
rs746268853 Chr.11: 62270871-62270871 on Build GRCh38
rs761789194 Chr.11: 62271121-62271121 on Build GRCh38
rs745560666 Chr.11: 62271207-62271208 on Build GRCh38
rs748177714 Chr.11: 62270855-62270855 on Build GRCh38
rs755370468 Chr.11: 62271163-62271163 on Build GRCh38
rs371996054 Chr.11: 62270830-62270830 on Build GRCh38
rs146064984 Chr.11: 62270976-62270976 on Build GRCh38
rs559678942 Chr.11: 62271066-62271066 on Build GRCh38
rs972988896 Chr.11: 62271257-62271257 on Build GRCh38
rs1007242941 Chr.11: 62270868-62270868 on Build GRCh38
rs749558588 Chr.11: 62271108-62271108 on Build GRCh38
rs528899216 Chr.11: 62271090-62271090 on Build GRCh38
rs867552370 Chr.11: 62271147-62271147 on Build GRCh38
rs927936896 Chr.11: 62270922-62270922 on Build GRCh38
rs115359496 Chr.11: 62271065-62271065 on Build GRCh38
rs754829394 Chr.11: 62270909-62270909 on Build GRCh38
rs758537527 Chr.11: 62270924-62270924 on Build GRCh38
rs991220656 Chr.11: 62271201-62271201 on Build GRCh38
rs376630772 Chr.11: 62270962-62270962 on Build GRCh38
rs760287496 Chr.11: 62270897-62270897 on Build GRCh38
rs777623420 Chr.11: 62270842-62270846 on Build GRCh38
rs777763012 Chr.11: 62270933-62270933 on Build GRCh38
rs747146202 Chr.11: 62270950-62270950 on Build GRCh38
rs200101717 Chr.11: 62270839-62270839 on Build GRCh38
rs941656689 Chr.11: 62270986-62270986 on Build GRCh38
rs148168628 Chr.11: 62270886-62270886 on Build GRCh38
rs770214480 Chr.11: 62270970-62270970 on Build GRCh38
rs758555967 Chr.11: 62270857-62270857 on Build GRCh38
rs769782640 Chr.11: 62271024-62271024 on Build GRCh38
rs752748582 Chr.11: 62270916-62270916 on Build GRCh38
rs781489958 Chr.11: 62270958-62270958 on Build GRCh38
rs766084370 Chr.11: 62270898-62270898 on Build GRCh38
rs769134446 Chr.11: 62271115-62271115 on Build GRCh38
rs780274420 Chr.11: 62271156-62271156 on Build GRCh38
rs1055841591 Chr.11: 62271157-62271157 on Build GRCh38
rs982213987 Chr.11: 62270918-62270918 on Build GRCh38
rs376092456 Chr.11: 62271068-62271068 on Build GRCh38
rs775289055 Chr.11: 62271238-62271238 on Build GRCh38
rs753525603 Chr.11: 62270908-62270908 on Build GRCh38
rs531920676 Chr.11: 62271211-62271211 on Build GRCh38
rs895047652 Chr.11: 62271181-62271181 on Build GRCh38
rs752579301 Chr.11: 62271169-62271169 on Build GRCh38
rs751582771 Chr.11: 62271120-62271120 on Build GRCh38
rs772554381 Chr.11: 62270983-62270983 on Build GRCh38
rs1024786075 Chr.11: 62271216-62271216 on Build GRCh38
rs754924227 Chr.11: 62270842-62270842 on Build GRCh38
rs756999292 Chr.11: 62270891-62270891 on Build GRCh38
rs759400983 Chr.11: 62271001-62271001 on Build GRCh38
rs536192644 Chr.11: 62271275-62271276 on Build GRCh38
rs771303036 Chr.11: 62270864-62270864 on Build GRCh38
rs75491361 Chr.11: 62271032-62271032 on Build GRCh38
rs747251146 Chr.11: 62270863-62270863 on Build GRCh38
rs757362582 Chr.11: 62271049-62271049 on Build GRCh38
rs371658204 Chr.11: 62271102-62271102 on Build GRCh38
rs776408906 Chr.11: 62271092-62271092 on Build GRCh38
rs780855305 Chr.11: 62271171-62271171 on Build GRCh38
rs749846065 Chr.11: 62270978-62270978 on Build GRCh38
rs776348342 Chr.11: 62270999-62270999 on Build GRCh38
rs770427836 Chr.11: 62270997-62270997 on Build GRCh38
rs1024839856 Chr.11: 62270920-62270920 on Build GRCh38
rs993246516 Chr.11: 62270844-62270844 on Build GRCh38
rs746823326 Chr.11: 62270859-62270859 on Build GRCh38
rs913768790 Chr.11: 62270895-62270895 on Build GRCh38
rs757374187 Chr.11: 62270956-62270956 on Build GRCh38
rs369145886 Chr.11: 62270870-62270870 on Build GRCh38
rs372699744 Chr.11: 62270875-62270875 on Build GRCh38
rs372833801 Chr.11: 62270985-62270985 on Build GRCh38
rs754141112 Chr.11: 62271158-62271158 on Build GRCh38
rs766139374 Chr.11: 62270835-62270835 on Build GRCh38
rs756698777 Chr.11: 62271200-62271200 on Build GRCh38
rs779485575 Chr.11: 62271069-62271069 on Build GRCh38
rs1051434369 Chr.11: 62271058-62271058 on Build GRCh38
rs772900941 Chr.11: 62270891-62270891 on Build GRCh38
rs7129637 Chr.11: 62271280-62271280 on Build GRCh38
rs779011931 Chr.11: 62270853-62270853 on Build GRCh38
rs567079945 Chr.11: 62271262-62271262 on Build GRCh38
rs750615102 Chr.11: 62271126-62271126 on Build GRCh38
rs765181973 Chr.11: 62271005-62271005 on Build GRCh38
rs966779520 Chr.11: 62270899-62270899 on Build GRCh38
rs959486204 Chr.11: 62271177-62271177 on Build GRCh38
rs778052515 Chr.11: 62270861-62270861 on Build GRCh38
rs918854592 Chr.11: 62271258-62271258 on Build GRCh38
rs138798780 Chr.11: 62271059-62271059 on Build GRCh38
rs376342209 Chr.11: 62271174-62271174 on Build GRCh38
rs763870683 Chr.11: 62271118-62271118 on Build GRCh38
rs966588719 Chr.11: 62271274-62271274 on Build GRCh38
rs762310027 Chr.11: 62271073-62271073 on Build GRCh38
rs189723654 Chr.11: 62270911-62270911 on Build GRCh38

View Full Product Details