Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62574893-62575381 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TUT1
Entrez Gene ID
64852
Gene Name
terminal uridylyl transferase 1, U6 snRNA-specific
Gene Aliases
PAPD2, RBM21, STARPAP, TUTase, URLC6
UniGene
Hs.728983
Gene Chromosome Location
Chr.11: 62575045-62591637 on Build GRCh38
Transcripts
NM_022830.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4848508 c.2556G>T p.L852L Chr.11: 62575277-62575277 on Build GRCh38
6814408 c.2655G>A p.P885P Chr.11: 62575178-62575178 on Build GRCh38
6814412 c.2567G>A p.S856N Chr.11: 62575266-62575266 on Build GRCh38
3791698 c.2632G>A p.D878N Chr.11: 62575201-62575201 on Build GRCh38
5442247 c.2375_2384del10 p.G792fs*20 Chr.11: 62575335-62575344 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777252459 Chr.11: 62575197-62575197 on Build GRCh38
rs761135793 Chr.11: 62575233-62575233 on Build GRCh38
rs773778058 Chr.11: 62575224-62575224 on Build GRCh38
rs536442629 Chr.11: 62575141-62575141 on Build GRCh38
rs745309998 Chr.11: 62574941-62574941 on Build GRCh38
rs573829216 Chr.11: 62575285-62575285 on Build GRCh38
rs762376439 Chr.11: 62575240-62575241 on Build GRCh38
rs572447780 Chr.11: 62575209-62575209 on Build GRCh38
rs779811606 Chr.11: 62575306-62575306 on Build GRCh38
rs769854710 Chr.11: 62575120-62575120 on Build GRCh38
rs562813679 Chr.11: 62575343-62575343 on Build GRCh38
rs900998160 Chr.11: 62575247-62575247 on Build GRCh38
rs1031315047 Chr.11: 62575318-62575318 on Build GRCh38
rs763713118 Chr.11: 62575251-62575251 on Build GRCh38
rs754989393 Chr.11: 62575340-62575340 on Build GRCh38
rs146742342 Chr.11: 62575220-62575220 on Build GRCh38
rs757949582 Chr.11: 62575194-62575194 on Build GRCh38
rs149939399 Chr.11: 62575159-62575159 on Build GRCh38
rs774674793 Chr.11: 62575072-62575072 on Build GRCh38
rs781081708 Chr.11: 62575102-62575102 on Build GRCh38
rs376433999 Chr.11: 62574917-62574919 on Build GRCh38
rs986416238 Chr.11: 62575218-62575218 on Build GRCh38
rs372140568 Chr.11: 62575089-62575089 on Build GRCh38
rs764797013 Chr.11: 62575181-62575181 on Build GRCh38
rs763145000 Chr.11: 62574958-62574958 on Build GRCh38
rs921213782 Chr.11: 62574995-62574995 on Build GRCh38
rs768700337 Chr.11: 62575142-62575142 on Build GRCh38
rs751203709 Chr.11: 62575260-62575260 on Build GRCh38
rs147146917 Chr.11: 62575154-62575154 on Build GRCh38
rs571025236 Chr.11: 62575038-62575038 on Build GRCh38
rs772352087 Chr.11: 62575348-62575348 on Build GRCh38
rs756973153 Chr.11: 62575266-62575266 on Build GRCh38
rs768041389 Chr.11: 62575357-62575357 on Build GRCh38
rs778613415 Chr.11: 62575081-62575081 on Build GRCh38
rs754506259 Chr.11: 62575072-62575072 on Build GRCh38
rs978420528 Chr.11: 62575309-62575309 on Build GRCh38
rs777495837 Chr.11: 62575090-62575090 on Build GRCh38
rs1056044 Chr.11: 62574921-62574921 on Build GRCh38
rs763243913 Chr.11: 62575357-62575357 on Build GRCh38
rs143498027 Chr.11: 62575214-62575214 on Build GRCh38
rs769221161 Chr.11: 62575049-62575049 on Build GRCh38
rs183285818 Chr.11: 62575100-62575100 on Build GRCh38
rs750997568 Chr.11: 62575071-62575071 on Build GRCh38
rs570212302 Chr.11: 62575155-62575155 on Build GRCh38
rs376557209 Chr.11: 62575132-62575132 on Build GRCh38
rs34169545 Chr.11: 62575291-62575291 on Build GRCh38
rs36080058 Chr.11: 62575346-62575346 on Build GRCh38
rs752092769 Chr.11: 62575191-62575191 on Build GRCh38
rs768808132 Chr.11: 62575213-62575213 on Build GRCh38
rs751802009 Chr.11: 62575048-62575048 on Build GRCh38
rs547069441 Chr.11: 62575092-62575092 on Build GRCh38
rs768084128 Chr.11: 62575062-62575062 on Build GRCh38
rs757027935 Chr.11: 62575201-62575201 on Build GRCh38
rs201318162 Chr.11: 62575217-62575217 on Build GRCh38
rs762339006 Chr.11: 62575250-62575250 on Build GRCh38
rs781026577 Chr.11: 62575203-62575203 on Build GRCh38
rs138638609 Chr.11: 62575211-62575211 on Build GRCh38
rs775762407 Chr.11: 62574953-62574953 on Build GRCh38
rs777226986 Chr.11: 62575242-62575242 on Build GRCh38
rs752379553 Chr.11: 62575086-62575086 on Build GRCh38
rs969068007 Chr.11: 62575304-62575304 on Build GRCh38
rs745859092 Chr.11: 62575111-62575111 on Build GRCh38
rs1036488397 Chr.11: 62575070-62575070 on Build GRCh38
rs767163500 Chr.11: 62575280-62575280 on Build GRCh38
rs769905791 Chr.11: 62574952-62574952 on Build GRCh38
rs766748841 Chr.11: 62575234-62575234 on Build GRCh38
rs200189280 Chr.11: 62575205-62575205 on Build GRCh38
rs771548950 Chr.11: 62575335-62575335 on Build GRCh38
rs944476638 Chr.11: 62575060-62575060 on Build GRCh38
rs751265667 Chr.11: 62575198-62575198 on Build GRCh38
rs1050261847 Chr.11: 62575007-62575007 on Build GRCh38
rs764512372 Chr.11: 62575045-62575045 on Build GRCh38
rs762224955 Chr.11: 62575153-62575153 on Build GRCh38
rs200684858 Chr.11: 62575049-62575049 on Build GRCh38
rs975364214 Chr.11: 62574972-62574972 on Build GRCh38
rs1019478061 Chr.11: 62574932-62574932 on Build GRCh38
rs1008987703 Chr.11: 62575230-62575230 on Build GRCh38
rs367814209 Chr.11: 62575144-62575144 on Build GRCh38
rs749284411 Chr.11: 62575339-62575339 on Build GRCh38
rs115926342 Chr.11: 62575206-62575206 on Build GRCh38

View Full Product Details