Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 62667095-62667592 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LBHD1
Entrez Gene ID
79081
Gene Name
LBH domain containing 1
Gene Aliases
C11orf48
UniGene
Hs.9061
Gene Chromosome Location
Chr.11: 62662817-62671769 on Build GRCh38
Transcripts
NM_024099.3 NM_001043229.1 XM_005274232.4

Gene Symbol
METTL12
Entrez Gene ID
751071
Gene Name
methyltransferase like 12
Gene Aliases
U99HG
UniGene
Hs.640726
Gene Chromosome Location
Chr.11: 62665252-62667451 on Build GRCh38
Transcripts
NM_024099.3 NM_001043229.1 XM_005274232.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5776544 c.516C>G p.L172L Chr.11: 62667545-62667545 on Build GRCh38
4666640 c.536A>G p.E179G Chr.11: 62667525-62667525 on Build GRCh38
6614024 c.538+2T>C p.? Chr.11: 62667521-62667521 on Build GRCh38
7078154 c.509C>T p.S170F Chr.11: 62667552-62667552 on Build GRCh38
929770 c.474C>T p.G158G Chr.11: 62667587-62667587 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs760065671 Chr.11: 62667500-62667500 on Build GRCh38
rs769745897 Chr.11: 62667505-62667505 on Build GRCh38
rs973719485 Chr.11: 62667256-62667256 on Build GRCh38
rs527424664 Chr.11: 62667531-62667531 on Build GRCh38
rs752236081 Chr.11: 62667458-62667458 on Build GRCh38
rs573649158 Chr.11: 62667266-62667266 on Build GRCh38
rs184318382 Chr.11: 62667254-62667254 on Build GRCh38
rs766009048 Chr.11: 62667447-62667447 on Build GRCh38
rs546870654 Chr.11: 62667523-62667523 on Build GRCh38
rs899871249 Chr.11: 62667488-62667492 on Build GRCh38
rs751392015 Chr.11: 62667569-62667569 on Build GRCh38
rs926273105 Chr.11: 62667549-62667549 on Build GRCh38
rs937011309 Chr.11: 62667222-62667222 on Build GRCh38
rs955589104 Chr.11: 62667151-62667151 on Build GRCh38
rs770090933 Chr.11: 62667396-62667396 on Build GRCh38
rs568048221 Chr.11: 62667259-62667259 on Build GRCh38
rs1049283552 Chr.11: 62667236-62667236 on Build GRCh38
rs778405450 Chr.11: 62667402-62667403 on Build GRCh38
rs764306346 Chr.11: 62667440-62667440 on Build GRCh38
rs940844320 Chr.11: 62667260-62667260 on Build GRCh38
rs564273174 Chr.11: 62667375-62667375 on Build GRCh38
rs1000900240 Chr.11: 62667298-62667298 on Build GRCh38
rs140170783 Chr.11: 62667524-62667524 on Build GRCh38
rs775000759 Chr.11: 62667443-62667443 on Build GRCh38
rs377120219 Chr.11: 62667267-62667267 on Build GRCh38
rs781071897 Chr.11: 62667502-62667502 on Build GRCh38
rs771499177 Chr.11: 62667452-62667453 on Build GRCh38
rs940986674 Chr.11: 62667245-62667245 on Build GRCh38
rs149450298 Chr.11: 62667535-62667535 on Build GRCh38
rs762370033 Chr.11: 62667446-62667446 on Build GRCh38
rs775568658 Chr.11: 62667404-62667404 on Build GRCh38
rs777042192 Chr.11: 62667459-62667460 on Build GRCh38
rs971213504 Chr.11: 62667451-62667451 on Build GRCh38
rs746011566 Chr.11: 62667503-62667503 on Build GRCh38
rs538056161 Chr.11: 62667124-62667124 on Build GRCh38
rs372883451 Chr.11: 62667555-62667555 on Build GRCh38
rs757230581 Chr.11: 62667496-62667496 on Build GRCh38
rs763430769 Chr.11: 62667437-62667437 on Build GRCh38
rs746974325 Chr.11: 62667478-62667478 on Build GRCh38
rs747606081 Chr.11: 62667447-62667455 on Build GRCh38
rs758219416 Chr.11: 62667463-62667463 on Build GRCh38
rs762467933 Chr.11: 62667554-62667554 on Build GRCh38
rs927253593 Chr.11: 62667509-62667509 on Build GRCh38
rs556083342 Chr.11: 62667453-62667453 on Build GRCh38
rs984504133 Chr.11: 62667550-62667550 on Build GRCh38
rs765650877 Chr.11: 62667419-62667419 on Build GRCh38
rs775079346 Chr.11: 62667526-62667526 on Build GRCh38
rs753440716 Chr.11: 62667450-62667450 on Build GRCh38
rs945243912 Chr.11: 62667271-62667271 on Build GRCh38
rs770112982 Chr.11: 62667553-62667553 on Build GRCh38
rs530779440 Chr.11: 62667281-62667281 on Build GRCh38
rs949946977 Chr.11: 62667136-62667136 on Build GRCh38
rs878911903 Chr.11: 62667149-62667149 on Build GRCh38
rs983424196 Chr.11: 62667163-62667163 on Build GRCh38
rs191417550 Chr.11: 62667193-62667193 on Build GRCh38
rs199532450 Chr.11: 62667499-62667499 on Build GRCh38
rs1020086542 Chr.11: 62667449-62667449 on Build GRCh38
rs200294500 Chr.11: 62667562-62667562 on Build GRCh38
rs117313894 Chr.11: 62667268-62667268 on Build GRCh38
rs889116453 Chr.11: 62667273-62667273 on Build GRCh38
rs192961319 Chr.11: 62667441-62667441 on Build GRCh38
rs749402374 Chr.11: 62667519-62667519 on Build GRCh38
rs187110095 Chr.11: 62667312-62667312 on Build GRCh38
rs775793113 Chr.11: 62667518-62667518 on Build GRCh38
rs578174201 Chr.11: 62667210-62667210 on Build GRCh38
rs777395137 Chr.11: 62667467-62667467 on Build GRCh38
rs956836160 Chr.11: 62667383-62667383 on Build GRCh38
rs1032828033 Chr.11: 62667321-62667321 on Build GRCh38
rs151251828 Chr.11: 62667530-62667530 on Build GRCh38
rs370358731 Chr.11: 62667540-62667540 on Build GRCh38
rs1036558810 Chr.11: 62667264-62667264 on Build GRCh38
rs752428625 Chr.11: 62667547-62667547 on Build GRCh38

View Full Product Details