Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 21564159-21564735 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESCO1
Entrez Gene ID
114799
Gene Name
establishment of sister chromatid cohesion N-acetyltransferase 1
Gene Aliases
A930014I12Rik, CTF, ECO1, EFO1, ESO1
UniGene
Hs.464733
Gene Chromosome Location
Chr.18: 21529281-21601015 on Build GRCh38
Transcripts
NM_052911.2 XM_011525798.1 XM_011525799.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4928896 c.1799C>A p.T600N Chr.18: 21564225-21564225 on Build GRCh38
986935 c.1770G>T p.K590N Chr.18: 21564254-21564254 on Build GRCh38
6465323 c.1769A>G p.K590R Chr.18: 21564255-21564255 on Build GRCh38
1387958 c.1747C>T p.H583Y Chr.18: 21564277-21564277 on Build GRCh38
181990 c.1718G>A p.R573Q Chr.18: 21564306-21564306 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762456232 Chr.18: 21564320-21564321 on Build GRCh38
rs754183001 Chr.18: 21564331-21564331 on Build GRCh38
rs201612317 Chr.18: 21564206-21564206 on Build GRCh38
rs189617054 Chr.18: 21564195-21564195 on Build GRCh38
rs1018847789 Chr.18: 21564678-21564678 on Build GRCh38
rs1053856163 Chr.18: 21564618-21564618 on Build GRCh38
rs551225772 Chr.18: 21564691-21564691 on Build GRCh38
rs1045381387 Chr.18: 21564433-21564433 on Build GRCh38
rs959551467 Chr.18: 21564533-21564533 on Build GRCh38
rs547409026 Chr.18: 21564516-21564516 on Build GRCh38
rs961390873 Chr.18: 21564486-21564486 on Build GRCh38
rs769761376 Chr.18: 21564286-21564286 on Build GRCh38
rs376720124 Chr.18: 21564213-21564213 on Build GRCh38
rs1028236281 Chr.18: 21564398-21564398 on Build GRCh38
rs766802575 Chr.18: 21564321-21564321 on Build GRCh38
rs1007342425 Chr.18: 21564657-21564657 on Build GRCh38
rs1042335172 Chr.18: 21564586-21564586 on Build GRCh38
rs541463384 Chr.18: 21564386-21564386 on Build GRCh38
rs1049593327 Chr.18: 21564639-21564639 on Build GRCh38
rs755317465 Chr.18: 21564332-21564332 on Build GRCh38
rs953278612 Chr.18: 21564655-21564655 on Build GRCh38
rs758618222 Chr.18: 21564358-21564358 on Build GRCh38
rs955314616 Chr.18: 21564401-21564401 on Build GRCh38
rs1038968379 Chr.18: 21564545-21564545 on Build GRCh38
rs973870479 Chr.18: 21564514-21564514 on Build GRCh38
rs900619527 Chr.18: 21564622-21564622 on Build GRCh38
rs755406299 Chr.18: 21564239-21564239 on Build GRCh38
rs748915800 Chr.18: 21564414-21564414 on Build GRCh38
rs556678584 Chr.18: 21564268-21564268 on Build GRCh38
rs995610880 Chr.18: 21564686-21564686 on Build GRCh38
rs949638677 Chr.18: 21564548-21564548 on Build GRCh38
rs765823724 Chr.18: 21564365-21564368 on Build GRCh38
rs149718555 Chr.18: 21564250-21564250 on Build GRCh38
rs948179559 Chr.18: 21564432-21564432 on Build GRCh38
rs1015269684 Chr.18: 21564476-21564476 on Build GRCh38
rs775106701 Chr.18: 21564321-21564321 on Build GRCh38
rs753186098 Chr.18: 21564340-21564340 on Build GRCh38
rs112451583 Chr.18: 21564671-21564671 on Build GRCh38
rs1015653180 Chr.18: 21564679-21564679 on Build GRCh38
rs923977308 Chr.18: 21564598-21564598 on Build GRCh38
rs398032086 Chr.18: 21564569-21564569 on Build GRCh38
rs930426493 Chr.18: 21564398-21564398 on Build GRCh38
rs376441317 Chr.18: 21564685-21564685 on Build GRCh38
rs772910319 Chr.18: 21564311-21564311 on Build GRCh38
rs200657231 Chr.18: 21564306-21564306 on Build GRCh38
rs375031583 Chr.18: 21564397-21564398 on Build GRCh38
rs765675134 Chr.18: 21564337-21564337 on Build GRCh38
rs755828169 Chr.18: 21564393-21564397 on Build GRCh38
rs754415293 Chr.18: 21564235-21564235 on Build GRCh38
rs866825796 Chr.18: 21564706-21564706 on Build GRCh38
rs77344817 Chr.18: 21564408-21564408 on Build GRCh38
rs775118502 Chr.18: 21564289-21564289 on Build GRCh38
rs906775390 Chr.18: 21564491-21564491 on Build GRCh38
rs756540306 Chr.18: 21564210-21564210 on Build GRCh38
rs1008162992 Chr.18: 21564676-21564676 on Build GRCh38
rs767012173 Chr.18: 21564310-21564310 on Build GRCh38
rs35032343 Chr.18: 21564558-21564558 on Build GRCh38
rs565593850 Chr.18: 21564309-21564309 on Build GRCh38
rs748195607 Chr.18: 21564262-21564262 on Build GRCh38
rs527670164 Chr.18: 21564312-21564312 on Build GRCh38
rs571238534 Chr.18: 21564692-21564692 on Build GRCh38
rs531426221 Chr.18: 21564687-21564687 on Build GRCh38
rs542388506 Chr.18: 21564330-21564330 on Build GRCh38
rs911809268 Chr.18: 21564245-21564245 on Build GRCh38
rs936728124 Chr.18: 21564617-21564617 on Build GRCh38
rs971930190 Chr.18: 21564694-21564694 on Build GRCh38
rs892433649 Chr.18: 21564560-21564560 on Build GRCh38
rs918165301 Chr.18: 21564547-21564547 on Build GRCh38
rs200328471 Chr.18: 21564297-21564297 on Build GRCh38
rs550013725 Chr.18: 21564650-21564650 on Build GRCh38
rs1007860153 Chr.18: 21564455-21564455 on Build GRCh38
rs181535271 Chr.18: 21564638-21564638 on Build GRCh38
rs146765045 Chr.18: 21564276-21564276 on Build GRCh38
rs990980285 Chr.18: 21564534-21564534 on Build GRCh38

View Full Product Details