Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 88090304-88090704 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCG2
Entrez Gene ID
9429
Gene Name
ATP binding cassette subfamily G member 2 (Junior blood group)
Gene Aliases
ABC15, ABCP, BCRP, BCRP1, BMDP, CD338, CDw338, EST157481, GOUT1, MRX, MXR, MXR-1, MXR1, UAQTL1
UniGene
Hs.480218
Gene Chromosome Location
Chr.4: 88090264-88231417 on Build GRCh38
Transcripts
NM_001257386.1 NM_004827.2 XM_005263354.3 XM_005263355.3 XM_005263356.3 XM_011532420.2 XM_017008852.1 XM_017008853.1

SNP Detail

SNP ID Chromosome Location
rs145123789 Chr.4: 88090648-88090648 on Build GRCh38
rs114469340 Chr.4: 88090483-88090483 on Build GRCh38
rs947297245 Chr.4: 88090665-88090665 on Build GRCh38
rs1019703204 Chr.4: 88090489-88090489 on Build GRCh38
rs115329310 Chr.4: 88090601-88090601 on Build GRCh38
rs1046062948 Chr.4: 88090341-88090341 on Build GRCh38
rs545996213 Chr.4: 88090576-88090576 on Build GRCh38
rs912309993 Chr.4: 88090635-88090635 on Build GRCh38
rs188930061 Chr.4: 88090357-88090357 on Build GRCh38
rs550834012 Chr.4: 88090355-88090355 on Build GRCh38
rs965731624 Chr.4: 88090493-88090493 on Build GRCh38
rs775461441 Chr.4: 88090529-88090529 on Build GRCh38
rs557935020 Chr.4: 88090365-88090365 on Build GRCh38
rs539917181 Chr.4: 88090645-88090645 on Build GRCh38
rs908751535 Chr.4: 88090546-88090546 on Build GRCh38
rs557650193 Chr.4: 88090509-88090509 on Build GRCh38
rs963577925 Chr.4: 88090516-88090516 on Build GRCh38
rs986525292 Chr.4: 88090634-88090634 on Build GRCh38
rs539639015 Chr.4: 88090360-88090360 on Build GRCh38
rs376176502 Chr.4: 88090606-88090606 on Build GRCh38
rs575820405 Chr.4: 88090566-88090566 on Build GRCh38
rs529201239 Chr.4: 88090349-88090349 on Build GRCh38
rs1002926942 Chr.4: 88090466-88090466 on Build GRCh38
rs535116458 Chr.4: 88090440-88090440 on Build GRCh38
rs538105525 Chr.4: 88090393-88090393 on Build GRCh38
rs1020775717 Chr.4: 88090427-88090427 on Build GRCh38
rs1010679141 Chr.4: 88090403-88090403 on Build GRCh38
rs879187137 Chr.4: 88090506-88090506 on Build GRCh38
rs115770495 Chr.4: 88090508-88090508 on Build GRCh38
rs906299817 Chr.4: 88090594-88090594 on Build GRCh38
rs988459393 Chr.4: 88090646-88090646 on Build GRCh38
rs931581938 Chr.4: 88090510-88090510 on Build GRCh38
rs905957278 Chr.4: 88090457-88090457 on Build GRCh38
rs553452752 Chr.4: 88090443-88090443 on Build GRCh38
rs934565116 Chr.4: 88090361-88090361 on Build GRCh38
rs984283210 Chr.4: 88090528-88090528 on Build GRCh38
rs748697417 Chr.4: 88090639-88090639 on Build GRCh38
rs768238016 Chr.4: 88090563-88090563 on Build GRCh38
rs1011441426 Chr.4: 88090436-88090436 on Build GRCh38

View Full Product Details