Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 116789283-116789782 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APOA5
Entrez Gene ID
116519
Gene Name
apolipoprotein A5
Gene Aliases
APOAV, RAP3
UniGene
Hs.283923
Gene Chromosome Location
Chr.11: 116789370-116792420 on Build GRCh38
Transcripts
NM_001166598.1 NM_052968.4 XM_017017169.1

SNP Detail

SNP ID Chromosome Location
rs1021561508 Chr.11: 116789395-116789395 on Build GRCh38
rs765775223 Chr.11: 116789582-116789582 on Build GRCh38
rs969042527 Chr.11: 116789560-116789560 on Build GRCh38
rs115102021 Chr.11: 116789356-116789356 on Build GRCh38
rs1053741927 Chr.11: 116789722-116789722 on Build GRCh38
rs536542142 Chr.11: 116789603-116789603 on Build GRCh38
rs754460898 Chr.11: 116789631-116789631 on Build GRCh38
rs991213407 Chr.11: 116789571-116789571 on Build GRCh38
rs1031826225 Chr.11: 116789457-116789457 on Build GRCh38
rs919198231 Chr.11: 116789307-116789307 on Build GRCh38
rs933220583 Chr.11: 116789700-116789700 on Build GRCh38
rs576576803 Chr.11: 116789709-116789709 on Build GRCh38
rs1052121148 Chr.11: 116789705-116789705 on Build GRCh38
rs958616459 Chr.11: 116789567-116789567 on Build GRCh38
rs759635244 Chr.11: 116789644-116789645 on Build GRCh38
rs534211586 Chr.11: 116789527-116789527 on Build GRCh38
rs1043110759 Chr.11: 116789359-116789359 on Build GRCh38
rs1052212185 Chr.11: 116789316-116789316 on Build GRCh38
rs35009896 Chr.11: 116789486-116789486 on Build GRCh38
rs1051765889 Chr.11: 116789358-116789358 on Build GRCh38
rs554471735 Chr.11: 116789491-116789491 on Build GRCh38
rs547584744 Chr.11: 116789355-116789355 on Build GRCh38
rs772948091 Chr.11: 116789394-116789394 on Build GRCh38
rs941784732 Chr.11: 116789584-116789584 on Build GRCh38
rs919119238 Chr.11: 116789680-116789680 on Build GRCh38
rs551919092 Chr.11: 116789393-116789393 on Build GRCh38
rs997882660 Chr.11: 116789357-116789357 on Build GRCh38
rs999065255 Chr.11: 116789455-116789455 on Build GRCh38
rs764436886 Chr.11: 116789585-116789585 on Build GRCh38
rs554191422 Chr.11: 116789533-116789533 on Build GRCh38
rs1016002610 Chr.11: 116789626-116789626 on Build GRCh38
rs566731687 Chr.11: 116789620-116789620 on Build GRCh38
rs908949810 Chr.11: 116789580-116789580 on Build GRCh38
rs866709526 Chr.11: 116789310-116789310 on Build GRCh38
rs1007560678 Chr.11: 116789376-116789376 on Build GRCh38
rs893872755 Chr.11: 116789733-116789733 on Build GRCh38
rs1025080446 Chr.11: 116789548-116789548 on Build GRCh38
rs889126068 Chr.11: 116789360-116789360 on Build GRCh38
rs35822632 Chr.11: 116789437-116789437 on Build GRCh38
rs182378073 Chr.11: 116789570-116789570 on Build GRCh38
rs1032409586 Chr.11: 116789385-116789385 on Build GRCh38
rs1021113748 Chr.11: 116789756-116789756 on Build GRCh38
rs760425054 Chr.11: 116789468-116789468 on Build GRCh38
rs33984246 Chr.11: 116789734-116789734 on Build GRCh38
rs779275499 Chr.11: 116789553-116789553 on Build GRCh38
rs963079561 Chr.11: 116789684-116789684 on Build GRCh38
rs146109377 Chr.11: 116789415-116789415 on Build GRCh38
rs1007529615 Chr.11: 116789717-116789717 on Build GRCh38
rs889100545 Chr.11: 116789710-116789710 on Build GRCh38
rs556776664 Chr.11: 116789707-116789707 on Build GRCh38
rs1014482648 Chr.11: 116789485-116789488 on Build GRCh38

View Full Product Details