Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118253636-118254183 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MPZL2
Entrez Gene ID
10205
Gene Name
myelin protein zero like 2
Gene Aliases
EVA, EVA1
UniGene
Hs.116651
Gene Chromosome Location
Chr.11: 118253416-118264536 on Build GRCh38
Transcripts
NM_005797.3

SNP Detail

SNP ID Chromosome Location
rs927245574 Chr.11: 118254010-118254023 on Build GRCh38
rs953203483 Chr.11: 118253781-118253781 on Build GRCh38
rs1014722897 Chr.11: 118253742-118253742 on Build GRCh38
rs1055675363 Chr.11: 118253711-118253711 on Build GRCh38
rs997704436 Chr.11: 118253772-118253772 on Build GRCh38
rs547050492 Chr.11: 118253817-118253817 on Build GRCh38
rs1031979428 Chr.11: 118253846-118253846 on Build GRCh38
rs565740307 Chr.11: 118253664-118253664 on Build GRCh38
rs941970201 Chr.11: 118254046-118254046 on Build GRCh38
rs897633486 Chr.11: 118253743-118253743 on Build GRCh38
rs41444446 Chr.11: 118253732-118253732 on Build GRCh38
rs571072289 Chr.11: 118253694-118253694 on Build GRCh38
rs748687354 Chr.11: 118253725-118253725 on Build GRCh38
rs778523010 Chr.11: 118253825-118253825 on Build GRCh38
rs1052397620 Chr.11: 118253716-118253716 on Build GRCh38
rs958291200 Chr.11: 118253862-118253862 on Build GRCh38
rs933357435 Chr.11: 118254108-118254108 on Build GRCh38
rs1036418891 Chr.11: 118253663-118253663 on Build GRCh38
rs997357168 Chr.11: 118254083-118254083 on Build GRCh38
rs968325444 Chr.11: 118253769-118253769 on Build GRCh38
rs949564659 Chr.11: 118253877-118253877 on Build GRCh38
rs887698563 Chr.11: 118253724-118253724 on Build GRCh38
rs1036387746 Chr.11: 118254024-118254024 on Build GRCh38
rs148512287 Chr.11: 118254146-118254146 on Build GRCh38
rs1030539467 Chr.11: 118254118-118254118 on Build GRCh38
rs941997398 Chr.11: 118254014-118254014 on Build GRCh38
rs540738549 Chr.11: 118254049-118254049 on Build GRCh38
rs578183962 Chr.11: 118253986-118253986 on Build GRCh38
rs536242662 Chr.11: 118253865-118253865 on Build GRCh38
rs867061647 Chr.11: 118253918-118253918 on Build GRCh38
rs938016612 Chr.11: 118253709-118253709 on Build GRCh38
rs897688000 Chr.11: 118254070-118254070 on Build GRCh38
rs575882076 Chr.11: 118253921-118253921 on Build GRCh38
rs1030075169 Chr.11: 118253778-118253778 on Build GRCh38
rs998504645 Chr.11: 118253773-118253773 on Build GRCh38
rs914094257 Chr.11: 118253866-118253866 on Build GRCh38
rs73596401 Chr.11: 118253886-118253886 on Build GRCh38
rs189327641 Chr.11: 118253960-118253960 on Build GRCh38
rs905219604 Chr.11: 118253684-118253684 on Build GRCh38
rs567192849 Chr.11: 118253834-118253834 on Build GRCh38
rs531325334 Chr.11: 118254033-118254033 on Build GRCh38
rs373910491 Chr.11: 118253968-118253968 on Build GRCh38
rs775016131 Chr.11: 118254115-118254115 on Build GRCh38
rs969921151 Chr.11: 118253910-118253910 on Build GRCh38

View Full Product Details