Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 118747684-118748180 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX6
Entrez Gene ID
1656
Gene Name
DEAD-box helicase 6
Gene Aliases
HLR2, P54, RCK
UniGene
Hs.408461
Gene Chromosome Location
Chr.11: 118747763-118791263 on Build GRCh38
Transcripts
NM_001257191.2 NM_004397.5 XM_011542644.1 XM_011542645.1 XM_017017251.1

SNP Detail

SNP ID Chromosome Location
rs193115497 Chr.11: 118748046-118748046 on Build GRCh38
rs374405269 Chr.11: 118747910-118747913 on Build GRCh38
rs890166044 Chr.11: 118747914-118747914 on Build GRCh38
rs938789319 Chr.11: 118747772-118747772 on Build GRCh38
rs188952095 Chr.11: 118747871-118747871 on Build GRCh38
rs1048033 Chr.11: 118747937-118747937 on Build GRCh38
rs371691727 Chr.11: 118748144-118748144 on Build GRCh38
rs79694906 Chr.11: 118747909-118747909 on Build GRCh38
rs572118329 Chr.11: 118747780-118747780 on Build GRCh38
rs1048040 Chr.11: 118747911-118747911 on Build GRCh38
rs782402184 Chr.11: 118747725-118747725 on Build GRCh38
rs960862646 Chr.11: 118748069-118748069 on Build GRCh38
rs781928713 Chr.11: 118747767-118747767 on Build GRCh38
rs640120 Chr.11: 118748128-118748128 on Build GRCh38
rs948696009 Chr.11: 118748109-118748109 on Build GRCh38
rs992908770 Chr.11: 118748082-118748082 on Build GRCh38
rs782049151 Chr.11: 118747797-118747797 on Build GRCh38
rs1048036 Chr.11: 118747926-118747926 on Build GRCh38
rs900150888 Chr.11: 118747963-118747963 on Build GRCh38
rs782154108 Chr.11: 118748141-118748141 on Build GRCh38
rs575757631 Chr.11: 118747909-118747910 on Build GRCh38
rs1008637774 Chr.11: 118747948-118747948 on Build GRCh38
rs1017808184 Chr.11: 118747957-118747957 on Build GRCh38
rs3177631 Chr.11: 118747776-118747776 on Build GRCh38
rs1048024 Chr.11: 118748067-118748067 on Build GRCh38
rs1022191335 Chr.11: 118747981-118747981 on Build GRCh38
rs572515270 Chr.11: 118748098-118748098 on Build GRCh38
rs183447257 Chr.11: 118747828-118747828 on Build GRCh38
rs571114193 Chr.11: 118748018-118748018 on Build GRCh38
rs948746638 Chr.11: 118747834-118747834 on Build GRCh38
rs548572595 Chr.11: 118747752-118747752 on Build GRCh38
rs782737606 Chr.11: 118747970-118747970 on Build GRCh38
rs977930672 Chr.11: 118747718-118747718 on Build GRCh38
rs993604048 Chr.11: 118747875-118747875 on Build GRCh38
rs782730469 Chr.11: 118748152-118748152 on Build GRCh38
rs546043156 Chr.11: 118747882-118747882 on Build GRCh38
rs915913146 Chr.11: 118747818-118747818 on Build GRCh38
rs544431701 Chr.11: 118748148-118748148 on Build GRCh38
rs556424545 Chr.11: 118747996-118747996 on Build GRCh38
rs1037904367 Chr.11: 118747857-118747857 on Build GRCh38
rs3741328 Chr.11: 118748117-118748117 on Build GRCh38
rs1048022 Chr.11: 118748076-118748076 on Build GRCh38
rs535768441 Chr.11: 118748031-118748031 on Build GRCh38
rs916011672 Chr.11: 118748101-118748101 on Build GRCh38
rs899438798 Chr.11: 118747866-118747866 on Build GRCh38
rs533995291 Chr.11: 118747849-118747849 on Build GRCh38

View Full Product Details