Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 42460426-42460870 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRICKLE1
Entrez Gene ID
144165
Gene Name
prickle planar cell polarity protein 1
Gene Aliases
EPM1B, RILP
UniGene
Hs.524348
Gene Chromosome Location
Chr.12: 42458338-42589770 on Build GRCh38
Transcripts
NM_001144881.1 NM_001144882.1 NM_001144883.1 NM_153026.2 XM_011537946.1 XM_011537947.2 XM_017018838.1 XM_017018839.1 XM_017018840.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1606099 c.1832A>G p.E611G Chr.12: 42460473-42460473 on Build GRCh38
5704426 c.1799C>T p.S600L Chr.12: 42460506-42460506 on Build GRCh38
1628587 c.1745C>A p.T582N Chr.12: 42460560-42460560 on Build GRCh38
139955 c.1722T>A p.C574* Chr.12: 42460583-42460583 on Build GRCh38
3460669 c.1702G>A p.E568K Chr.12: 42460603-42460603 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771584708 Chr.12: 42460629-42460629 on Build GRCh38
rs753059335 Chr.12: 42460696-42460696 on Build GRCh38
rs776696757 Chr.12: 42460511-42460512 on Build GRCh38
rs1007932940 Chr.12: 42460531-42460531 on Build GRCh38
rs772922391 Chr.12: 42460649-42460649 on Build GRCh38
rs757362404 Chr.12: 42460537-42460537 on Build GRCh38
rs747016163 Chr.12: 42460536-42460536 on Build GRCh38
rs959822383 Chr.12: 42460788-42460788 on Build GRCh38
rs752938677 Chr.12: 42460519-42460519 on Build GRCh38
rs955717976 Chr.12: 42460619-42460619 on Build GRCh38
rs1002543932 Chr.12: 42460617-42460617 on Build GRCh38
rs1046723969 Chr.12: 42460505-42460505 on Build GRCh38
rs999302949 Chr.12: 42460827-42460827 on Build GRCh38
rs368664984 Chr.12: 42460501-42460501 on Build GRCh38
rs533432460 Chr.12: 42460664-42460664 on Build GRCh38
rs996774066 Chr.12: 42460485-42460485 on Build GRCh38
rs778075325 Chr.12: 42460533-42460533 on Build GRCh38
rs767033548 Chr.12: 42460460-42460460 on Build GRCh38
rs1011226149 Chr.12: 42460627-42460627 on Build GRCh38
rs749854195 Chr.12: 42460472-42460472 on Build GRCh38
rs529965135 Chr.12: 42460792-42460792 on Build GRCh38
rs149518219 Chr.12: 42460658-42460658 on Build GRCh38
rs942056065 Chr.12: 42460806-42460806 on Build GRCh38
rs192905881 Chr.12: 42460593-42460593 on Build GRCh38
rs983554311 Chr.12: 42460791-42460791 on Build GRCh38
rs200319900 Chr.12: 42460714-42460714 on Build GRCh38
rs1051937466 Chr.12: 42460506-42460506 on Build GRCh38
rs754414301 Chr.12: 42460488-42460488 on Build GRCh38
rs370892022 Chr.12: 42460659-42460659 on Build GRCh38
rs749279543 Chr.12: 42460550-42460550 on Build GRCh38
rs760050261 Chr.12: 42460657-42460657 on Build GRCh38
rs747960166 Chr.12: 42460610-42460610 on Build GRCh38
rs766728682 Chr.12: 42460486-42460486 on Build GRCh38
rs138079613 Chr.12: 42460558-42460558 on Build GRCh38
rs748163360 Chr.12: 42460705-42460706 on Build GRCh38
rs765766362 Chr.12: 42460514-42460514 on Build GRCh38
rs907932475 Chr.12: 42460804-42460804 on Build GRCh38
rs774182139 Chr.12: 42460594-42460594 on Build GRCh38
rs890912439 Chr.12: 42460526-42460526 on Build GRCh38
rs1037691847 Chr.12: 42460846-42460846 on Build GRCh38
rs761518408 Chr.12: 42460449-42460449 on Build GRCh38
rs764151052 Chr.12: 42460715-42460715 on Build GRCh38
rs1055556905 Chr.12: 42460650-42460650 on Build GRCh38
rs758594837 Chr.12: 42460524-42460524 on Build GRCh38

View Full Product Details