Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 76815726-76816264 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZDHHC17
Entrez Gene ID
23390
Gene Name
zinc finger DHHC-type containing 17
Gene Aliases
HIP14, HIP3, HSPC294, HYPH
UniGene
Hs.4014
Gene Chromosome Location
Chr.12: 76764074-76853701 on Build GRCh38
Transcripts
NM_015336.2 XM_005268750.4 XM_011538083.1 XM_017019091.1 XM_017019092.1 XM_017019093.1 XM_017019094.1 XM_017019095.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5051937 c.638T>C p.F213S Chr.12: 76815886-76815886 on Build GRCh38
6265378 c.751G>T p.V251F Chr.12: 76815999-76815999 on Build GRCh38
3955161 c.664A>G p.K222E Chr.12: 76815912-76815912 on Build GRCh38
6366721 c.610G>T p.V204L Chr.12: 76815858-76815858 on Build GRCh38
1606649 c.609-10T>C p.? Chr.12: 76815847-76815847 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200668037 Chr.12: 76815833-76815833 on Build GRCh38
rs748985532 Chr.12: 76815880-76815880 on Build GRCh38
rs77238734 Chr.12: 76815846-76815847 on Build GRCh38
rs764278732 Chr.12: 76815935-76815935 on Build GRCh38
rs576406715 Chr.12: 76816033-76816033 on Build GRCh38
rs777270536 Chr.12: 76816002-76816002 on Build GRCh38
rs74104863 Chr.12: 76816216-76816216 on Build GRCh38
rs754946998 Chr.12: 76815902-76815902 on Build GRCh38
rs866976976 Chr.12: 76815876-76815876 on Build GRCh38
rs760785281 Chr.12: 76815933-76815933 on Build GRCh38
rs1054734032 Chr.12: 76815771-76815771 on Build GRCh38
rs756994156 Chr.12: 76816014-76816014 on Build GRCh38
rs982320786 Chr.12: 76816076-76816076 on Build GRCh38
rs758473499 Chr.12: 76816110-76816110 on Build GRCh38
rs1042203433 Chr.12: 76815802-76815802 on Build GRCh38
rs776316669 Chr.12: 76815833-76815834 on Build GRCh38
rs192442836 Chr.12: 76816063-76816063 on Build GRCh38
rs562178995 Chr.12: 76816127-76816127 on Build GRCh38
rs893712648 Chr.12: 76815780-76815780 on Build GRCh38
rs751418988 Chr.12: 76815965-76815965 on Build GRCh38
rs761492179 Chr.12: 76815834-76815834 on Build GRCh38
rs535267285 Chr.12: 76815765-76815765 on Build GRCh38
rs914949102 Chr.12: 76816133-76816133 on Build GRCh38
rs777871968 Chr.12: 76815817-76815819 on Build GRCh38
rs764655949 Chr.12: 76815834-76815836 on Build GRCh38
rs747695817 Chr.12: 76815848-76815848 on Build GRCh38
rs765473699 Chr.12: 76815792-76815792 on Build GRCh38
rs1026574514 Chr.12: 76816038-76816038 on Build GRCh38
rs777356275 Chr.12: 76815867-76815867 on Build GRCh38
rs781067803 Chr.12: 76815981-76815981 on Build GRCh38
rs963619418 Chr.12: 76815963-76815963 on Build GRCh38
rs200802830 Chr.12: 76815901-76815901 on Build GRCh38
rs531115647 Chr.12: 76815837-76815837 on Build GRCh38
rs759950035 Chr.12: 76815909-76815909 on Build GRCh38
rs774831158 Chr.12: 76815833-76815833 on Build GRCh38
rs752653960 Chr.12: 76815986-76815986 on Build GRCh38
rs762159922 Chr.12: 76815945-76815945 on Build GRCh38
rs748892285 Chr.12: 76816009-76816009 on Build GRCh38
rs780658728 Chr.12: 76815838-76815838 on Build GRCh38
rs1024209379 Chr.12: 76816032-76816032 on Build GRCh38
rs894304959 Chr.12: 76815955-76815955 on Build GRCh38
rs758067724 Chr.12: 76815819-76815819 on Build GRCh38
rs771135043 Chr.12: 76815889-76815889 on Build GRCh38
rs990429544 Chr.12: 76816114-76816114 on Build GRCh38
rs772688425 Chr.12: 76815927-76815927 on Build GRCh38
rs922066710 Chr.12: 76815786-76815786 on Build GRCh38
rs373436500 Chr.12: 76815966-76815966 on Build GRCh38
rs754044500 Chr.12: 76815938-76815938 on Build GRCh38
rs376192102 Chr.12: 76816039-76816039 on Build GRCh38
rs755564828 Chr.12: 76815994-76815994 on Build GRCh38
rs940677933 Chr.12: 76815822-76815822 on Build GRCh38
rs572647216 Chr.12: 76816143-76816143 on Build GRCh38
rs536981651 Chr.12: 76815836-76815836 on Build GRCh38
rs184421970 Chr.12: 76816148-76816148 on Build GRCh38
rs772395157 Chr.12: 76815834-76815835 on Build GRCh38
rs902381175 Chr.12: 76815817-76815817 on Build GRCh38
rs771415323 Chr.12: 76815827-76815830 on Build GRCh38
rs749766500 Chr.12: 76815826-76815826 on Build GRCh38
rs927675852 Chr.12: 76816097-76816097 on Build GRCh38
rs775683938 Chr.12: 76815930-76815930 on Build GRCh38
rs61930508 Chr.12: 76815849-76815849 on Build GRCh38
rs998029308 Chr.12: 76815820-76815820 on Build GRCh38
rs75525897 Chr.12: 76815847-76815847 on Build GRCh38
rs749889154 Chr.12: 76815848-76815849 on Build GRCh38
rs766072017 Chr.12: 76815951-76815951 on Build GRCh38
rs34301481 Chr.12: 76816122-76816123 on Build GRCh38

View Full Product Details