Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 77998252-77998752 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NAV3
Entrez Gene ID
89795
Gene Name
neuron navigator 3
Gene Aliases
POMFIL1, STEERIN3, unc53H3
UniGene
Hs.655301
Gene Chromosome Location
Chr.12: 77571862-78213012 on Build GRCh38
Transcripts
NM_001024383.1 NM_014903.5 XM_005269215.3 XM_011538940.2 XM_011538941.2 XM_011538944.2 XM_017020164.1 XM_017020165.1 XM_017020166.1 XM_017020167.1 XM_017020168.1 XM_017020169.1 XM_017020170.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5051946 c.796G>T p.A266S Chr.12: 77998392-77998392 on Build GRCh38
6032690 c.763C>T p.P255S Chr.12: 77998359-77998359 on Build GRCh38
5521767 c.874G>A p.E292K Chr.12: 77998470-77998470 on Build GRCh38
4747522 c.880+1_880+4delGTAA p.? Chr.12: 77998477-77998480 on Build GRCh38
6224041 c.741-3C>T p.? Chr.12: 77998334-77998334 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs903264974 Chr.12: 77998671-77998671 on Build GRCh38
rs746256657 Chr.12: 77998453-77998453 on Build GRCh38
rs959412535 Chr.12: 77998280-77998280 on Build GRCh38
rs774799435 Chr.12: 77998343-77998343 on Build GRCh38
rs760803030 Chr.12: 77998364-77998364 on Build GRCh38
rs771245212 Chr.12: 77998333-77998333 on Build GRCh38
rs995717104 Chr.12: 77998601-77998601 on Build GRCh38
rs997634760 Chr.12: 77998687-77998687 on Build GRCh38
rs937179708 Chr.12: 77998600-77998600 on Build GRCh38
rs781245455 Chr.12: 77998405-77998405 on Build GRCh38
rs768773187 Chr.12: 77998487-77998487 on Build GRCh38
rs200556761 Chr.12: 77998304-77998304 on Build GRCh38
rs775372605 Chr.12: 77998352-77998352 on Build GRCh38
rs754270000 Chr.12: 77998628-77998628 on Build GRCh38
rs928439733 Chr.12: 77998588-77998588 on Build GRCh38
rs776718075 Chr.12: 77998489-77998489 on Build GRCh38
rs747060237 Chr.12: 77998475-77998475 on Build GRCh38
rs766043128 Chr.12: 77998383-77998383 on Build GRCh38
rs770730449 Chr.12: 77998346-77998347 on Build GRCh38
rs755324163 Chr.12: 77998665-77998665 on Build GRCh38
rs1055612378 Chr.12: 77998616-77998616 on Build GRCh38
rs981234579 Chr.12: 77998586-77998586 on Build GRCh38
rs895180194 Chr.12: 77998620-77998620 on Build GRCh38
rs889247472 Chr.12: 77998704-77998704 on Build GRCh38
rs1045565501 Chr.12: 77998289-77998289 on Build GRCh38
rs966961778 Chr.12: 77998677-77998677 on Build GRCh38
rs929757183 Chr.12: 77998574-77998574 on Build GRCh38
rs752247909 Chr.12: 77998409-77998409 on Build GRCh38
rs555001037 Chr.12: 77998503-77998503 on Build GRCh38
rs865880453 Chr.12: 77998636-77998636 on Build GRCh38
rs765502810 Chr.12: 77998510-77998510 on Build GRCh38
rs755721519 Chr.12: 77998429-77998429 on Build GRCh38
rs756965261 Chr.12: 77998441-77998441 on Build GRCh38
rs953602073 Chr.12: 77998501-77998501 on Build GRCh38
rs779053825 Chr.12: 77998407-77998409 on Build GRCh38
rs758173215 Chr.12: 77998495-77998497 on Build GRCh38
rs920979756 Chr.12: 77998532-77998532 on Build GRCh38
rs751404552 Chr.12: 77998386-77998386 on Build GRCh38
rs947394504 Chr.12: 77998649-77998649 on Build GRCh38
rs577144695 Chr.12: 77998662-77998662 on Build GRCh38
rs764298501 Chr.12: 77998365-77998365 on Build GRCh38
rs569224647 Chr.12: 77998523-77998523 on Build GRCh38
rs759438743 Chr.12: 77998518-77998518 on Build GRCh38
rs530248152 Chr.12: 77998557-77998557 on Build GRCh38
rs79741620 Chr.12: 77998548-77998548 on Build GRCh38
rs898349230 Chr.12: 77998593-77998593 on Build GRCh38
rs772446522 Chr.12: 77998458-77998458 on Build GRCh38
rs759919600 Chr.12: 77998348-77998348 on Build GRCh38
rs749634996 Chr.12: 77998327-77998327 on Build GRCh38
rs780511446 Chr.12: 77998551-77998551 on Build GRCh38
rs1051856148 Chr.12: 77998688-77998688 on Build GRCh38
rs113203797 Chr.12: 77998297-77998297 on Build GRCh38
rs761905630 Chr.12: 77998491-77998491 on Build GRCh38
rs762715478 Chr.12: 77998378-77998378 on Build GRCh38
rs532173821 Chr.12: 77998494-77998494 on Build GRCh38
rs773962618 Chr.12: 77998515-77998515 on Build GRCh38
rs769553773 Chr.12: 77998317-77998317 on Build GRCh38
rs978294086 Chr.12: 77998728-77998728 on Build GRCh38
rs754969797 Chr.12: 77998305-77998305 on Build GRCh38
rs373504958 Chr.12: 77998359-77998359 on Build GRCh38
rs932338433 Chr.12: 77998535-77998535 on Build GRCh38
rs553732872 Chr.12: 77998617-77998617 on Build GRCh38
rs772635192 Chr.12: 77998349-77998349 on Build GRCh38
rs754808430 Chr.12: 77998398-77998398 on Build GRCh38
rs377652650 Chr.12: 77998446-77998446 on Build GRCh38
rs779280355 Chr.12: 77998710-77998710 on Build GRCh38
rs748899312 Chr.12: 77998438-77998438 on Build GRCh38
rs918324521 Chr.12: 77998552-77998552 on Build GRCh38
rs749903485 Chr.12: 77998469-77998469 on Build GRCh38
rs546082328 Chr.12: 77998666-77998666 on Build GRCh38
rs777541410 Chr.12: 77998436-77998436 on Build GRCh38
rs373170355 Chr.12: 77998288-77998288 on Build GRCh38
rs780530948 Chr.12: 77998306-77998306 on Build GRCh38
rs747668795 Chr.12: 77998312-77998312 on Build GRCh38
rs777633141 Chr.12: 77998326-77998326 on Build GRCh38
rs796717278 Chr.12: 77998366-77998366 on Build GRCh38
rs1036891834 Chr.12: 77998573-77998573 on Build GRCh38
rs893100753 Chr.12: 77998417-77998417 on Build GRCh38
rs754100018 Chr.12: 77998375-77998375 on Build GRCh38

View Full Product Details