Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 80797640-80798147 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LIN7A
Entrez Gene ID
8825
Gene Name
lin-7 homolog A, crumbs cell polarity complex component
Gene Aliases
LIN-7A, LIN7, MALS-1, TIP-33, VELI1
UniGene
Hs.144333
Gene Chromosome Location
Chr.12: 80792520-80937915 on Build GRCh38
Transcripts
NM_001324423.1 NM_004664.3 NR_136887.1 NR_136888.1 XM_011538928.2

SNP Detail

SNP ID Chromosome Location
rs538285367 Chr.12: 80797745-80797745 on Build GRCh38
rs183009367 Chr.12: 80797841-80797841 on Build GRCh38
rs192007078 Chr.12: 80797811-80797811 on Build GRCh38
rs935900190 Chr.12: 80797747-80797747 on Build GRCh38
rs140221371 Chr.12: 80797683-80797683 on Build GRCh38
rs905911923 Chr.12: 80797807-80797807 on Build GRCh38
rs1003619371 Chr.12: 80798082-80798082 on Build GRCh38
rs1017101303 Chr.12: 80797842-80797842 on Build GRCh38
rs1032464706 Chr.12: 80798064-80798064 on Build GRCh38
rs35359139 Chr.12: 80797924-80797925 on Build GRCh38
rs962979910 Chr.12: 80797883-80797883 on Build GRCh38
rs1032743026 Chr.12: 80797719-80797719 on Build GRCh38
rs755292611 Chr.12: 80797903-80797903 on Build GRCh38
rs754007061 Chr.12: 80797833-80797833 on Build GRCh38
rs772101339 Chr.12: 80798054-80798054 on Build GRCh38
rs188353296 Chr.12: 80797787-80797787 on Build GRCh38
rs950383674 Chr.12: 80797985-80797985 on Build GRCh38
rs1038833183 Chr.12: 80797733-80797733 on Build GRCh38
rs2279664 Chr.12: 80798063-80798063 on Build GRCh38
rs751501811 Chr.12: 80797717-80797717 on Build GRCh38
rs932375863 Chr.12: 80797884-80797884 on Build GRCh38
rs192701347 Chr.12: 80797963-80797963 on Build GRCh38
rs569834716 Chr.12: 80797935-80797935 on Build GRCh38
rs185709785 Chr.12: 80797975-80797975 on Build GRCh38
rs187736292 Chr.12: 80797910-80797910 on Build GRCh38
rs34370362 Chr.12: 80797971-80797972 on Build GRCh38
rs943201530 Chr.12: 80797701-80797701 on Build GRCh38
rs757139159 Chr.12: 80797760-80797760 on Build GRCh38
rs778986378 Chr.12: 80797957-80797957 on Build GRCh38
rs764166084 Chr.12: 80797672-80797672 on Build GRCh38
rs114176129 Chr.12: 80797949-80797949 on Build GRCh38
rs920991809 Chr.12: 80797860-80797860 on Build GRCh38
rs1036982986 Chr.12: 80797940-80797940 on Build GRCh38
rs182832384 Chr.12: 80797694-80797694 on Build GRCh38
rs564619000 Chr.12: 80797693-80797693 on Build GRCh38
rs747461035 Chr.12: 80798073-80798073 on Build GRCh38
rs987117898 Chr.12: 80798026-80798026 on Build GRCh38
rs972967398 Chr.12: 80797922-80797922 on Build GRCh38
rs1002316124 Chr.12: 80797698-80797698 on Build GRCh38
rs866986302 Chr.12: 80798009-80798009 on Build GRCh38
rs189024993 Chr.12: 80798071-80798071 on Build GRCh38
rs913821328 Chr.12: 80797764-80797764 on Build GRCh38
rs77984139 Chr.12: 80798124-80798124 on Build GRCh38
rs530961287 Chr.12: 80797673-80797673 on Build GRCh38
rs35411465 Chr.12: 80797944-80797945 on Build GRCh38
rs1024813255 Chr.12: 80797798-80797798 on Build GRCh38
rs761805183 Chr.12: 80797859-80797859 on Build GRCh38
rs905388521 Chr.12: 80798023-80798023 on Build GRCh38
rs958097138 Chr.12: 80797742-80797742 on Build GRCh38
rs145314504 Chr.12: 80797917-80797917 on Build GRCh38

View Full Product Details