Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 120341668-120342170 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MSI1
Entrez Gene ID
4440
Gene Name
musashi RNA binding protein 1
Gene Aliases
-
UniGene
Hs.158311
Gene Chromosome Location
Chr.12: 120339655-120369180 on Build GRCh38
Transcripts
NM_002442.3 XM_006719403.3 XM_006719404.3 XM_011538361.2 XM_011538362.2 XM_011538363.2 XM_011538364.2 XM_011538365.2 XM_011538366.2 XM_011538368.2 XM_011538370.2 XM_011538371.2

SNP Detail

SNP ID Chromosome Location
rs949172645 Chr.12: 120341950-120341950 on Build GRCh38
rs926556124 Chr.12: 120342043-120342043 on Build GRCh38
rs1039269476 Chr.12: 120341883-120341883 on Build GRCh38
rs532548347 Chr.12: 120342110-120342110 on Build GRCh38
rs923443060 Chr.12: 120341782-120341782 on Build GRCh38
rs546397894 Chr.12: 120342127-120342127 on Build GRCh38
rs992178947 Chr.12: 120342059-120342059 on Build GRCh38
rs971888208 Chr.12: 120341851-120341851 on Build GRCh38
rs939347477 Chr.12: 120341735-120341735 on Build GRCh38
rs954803612 Chr.12: 120341835-120341835 on Build GRCh38
rs980682424 Chr.12: 120341687-120341687 on Build GRCh38
rs929845460 Chr.12: 120341906-120341906 on Build GRCh38
rs2522137 Chr.12: 120342128-120342128 on Build GRCh38
rs896309081 Chr.12: 120341947-120341947 on Build GRCh38
rs543695457 Chr.12: 120342100-120342100 on Build GRCh38
rs554919969 Chr.12: 120342042-120342042 on Build GRCh38
rs771132283 Chr.12: 120342004-120342004 on Build GRCh38
rs1009249713 Chr.12: 120341930-120341930 on Build GRCh38
rs1049261274 Chr.12: 120341865-120341865 on Build GRCh38
rs552766023 Chr.12: 120341829-120341829 on Build GRCh38
rs887920825 Chr.12: 120341869-120341869 on Build GRCh38
rs572911279 Chr.12: 120341893-120341893 on Build GRCh38
rs541593494 Chr.12: 120341955-120341955 on Build GRCh38
rs1056988143 Chr.12: 120341803-120341803 on Build GRCh38
rs919331632 Chr.12: 120341813-120341813 on Build GRCh38
rs778496922 Chr.12: 120341785-120341785 on Build GRCh38
rs919219809 Chr.12: 120342078-120342078 on Build GRCh38
rs967840389 Chr.12: 120342022-120342022 on Build GRCh38
rs372261703 Chr.12: 120342096-120342096 on Build GRCh38
rs1029252512 Chr.12: 120341903-120341903 on Build GRCh38
rs777615026 Chr.12: 120342011-120342011 on Build GRCh38
rs929394462 Chr.12: 120342083-120342083 on Build GRCh38
rs528566152 Chr.12: 120342138-120342138 on Build GRCh38
rs981118948 Chr.12: 120342039-120342039 on Build GRCh38
rs1022099969 Chr.12: 120341946-120341946 on Build GRCh38
rs1008255271 Chr.12: 120341878-120341878 on Build GRCh38
rs184276230 Chr.12: 120342104-120342104 on Build GRCh38
rs919810865 Chr.12: 120341904-120341904 on Build GRCh38
rs1000459343 Chr.12: 120342040-120342040 on Build GRCh38
rs1044753719 Chr.12: 120341961-120341962 on Build GRCh38
rs926643451 Chr.12: 120341732-120341732 on Build GRCh38
rs961024535 Chr.12: 120342052-120342052 on Build GRCh38
rs534089226 Chr.12: 120341918-120341918 on Build GRCh38
rs899538113 Chr.12: 120341887-120341887 on Build GRCh38
rs929340198 Chr.12: 120341845-120341845 on Build GRCh38
rs1032971638 Chr.12: 120341694-120341694 on Build GRCh38
rs1049161219 Chr.12: 120342101-120342101 on Build GRCh38

View Full Product Details