Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 120738938-120739443 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACADS
Entrez Gene ID
35
Gene Name
acyl-CoA dehydrogenase, C-2 to C-3 short chain
Gene Aliases
ACAD3, SCAD
UniGene
Hs.507076
Gene Chromosome Location
Chr.12: 120725738-120740008 on Build GRCh38
Transcripts
NM_000017.3 NM_001302554.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1639055 c.1067C>T p.A356V Chr.12: 120739177-120739177 on Build GRCh38
3457074 c.1129C>G p.P377A Chr.12: 120739338-120739338 on Build GRCh38
5913318 c.1105G>A p.G369S Chr.12: 120739314-120739314 on Build GRCh38
7235654 c.1066G>A p.A356T Chr.12: 120739176-120739176 on Build GRCh38
5840325 c.1211G>A p.G404E Chr.12: 120739420-120739420 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs555045670 Chr.12: 120739056-120739056 on Build GRCh38
rs895841347 Chr.12: 120739385-120739385 on Build GRCh38
rs775999272 Chr.12: 120739391-120739391 on Build GRCh38
rs773946131 Chr.12: 120739268-120739268 on Build GRCh38
rs776623770 Chr.12: 120739201-120739201 on Build GRCh38
rs979058757 Chr.12: 120738978-120738979 on Build GRCh38
rs377559470 Chr.12: 120739178-120739178 on Build GRCh38
rs766183395 Chr.12: 120739366-120739366 on Build GRCh38
rs953436518 Chr.12: 120739344-120739344 on Build GRCh38
rs762443848 Chr.12: 120739290-120739290 on Build GRCh38
rs57852948 Chr.12: 120739000-120739001 on Build GRCh38
rs145466253 Chr.12: 120739314-120739314 on Build GRCh38
rs931944863 Chr.12: 120738968-120738968 on Build GRCh38
rs369840561 Chr.12: 120739407-120739407 on Build GRCh38
rs375931905 Chr.12: 120739404-120739404 on Build GRCh38
rs566325901 Chr.12: 120739317-120739317 on Build GRCh38
rs926187308 Chr.12: 120739405-120739405 on Build GRCh38
rs894829104 Chr.12: 120739034-120739034 on Build GRCh38
rs768426621 Chr.12: 120738958-120738958 on Build GRCh38
rs368572195 Chr.12: 120739227-120739227 on Build GRCh38
rs751801455 Chr.12: 120739217-120739217 on Build GRCh38
rs1057516421 Chr.12: 120739373-120739374 on Build GRCh38
rs757766697 Chr.12: 120739106-120739106 on Build GRCh38
rs866546417 Chr.12: 120739410-120739410 on Build GRCh38
rs1049568378 Chr.12: 120739055-120739055 on Build GRCh38
rs145594828 Chr.12: 120739163-120739163 on Build GRCh38
rs781172419 Chr.12: 120739162-120739162 on Build GRCh38
rs377069922 Chr.12: 120739418-120739418 on Build GRCh38
rs948685149 Chr.12: 120739392-120739392 on Build GRCh38
rs765011205 Chr.12: 120738965-120738965 on Build GRCh38
rs769675520 Chr.12: 120739254-120739254 on Build GRCh38
rs28940875 Chr.12: 120739347-120739347 on Build GRCh38
rs374867417 Chr.12: 120739340-120739340 on Build GRCh38
rs545737847 Chr.12: 120739419-120739419 on Build GRCh38
rs936141551 Chr.12: 120739244-120739244 on Build GRCh38
rs759774887 Chr.12: 120739394-120739394 on Build GRCh38
rs556890619 Chr.12: 120739416-120739416 on Build GRCh38
rs763455552 Chr.12: 120739381-120739381 on Build GRCh38
rs766657084 Chr.12: 120739382-120739382 on Build GRCh38
rs759217705 Chr.12: 120739274-120739274 on Build GRCh38
rs796051903 Chr.12: 120739149-120739149 on Build GRCh38
rs387906950 Chr.12: 120739141-120739141 on Build GRCh38
rs1015232355 Chr.12: 120739325-120739325 on Build GRCh38
rs762451430 Chr.12: 120739092-120739092 on Build GRCh38
rs770439736 Chr.12: 120739174-120739174 on Build GRCh38
rs759011410 Chr.12: 120739181-120739181 on Build GRCh38
rs750672257 Chr.12: 120739127-120739127 on Build GRCh38
rs371604145 Chr.12: 120739230-120739230 on Build GRCh38
rs140372512 Chr.12: 120739099-120739099 on Build GRCh38
rs953695669 Chr.12: 120739307-120739307 on Build GRCh38
rs202078273 Chr.12: 120739164-120739164 on Build GRCh38
rs926163707 Chr.12: 120739213-120739213 on Build GRCh38
rs561806153 Chr.12: 120739207-120739207 on Build GRCh38
rs796051905 Chr.12: 120739321-120739321 on Build GRCh38
rs368064268 Chr.12: 120739348-120739348 on Build GRCh38
rs776136316 Chr.12: 120739422-120739422 on Build GRCh38
rs770730868 Chr.12: 120739261-120739261 on Build GRCh38
rs566185314 Chr.12: 120738976-120738976 on Build GRCh38
rs545598561 Chr.12: 120739143-120739143 on Build GRCh38
rs777643097 Chr.12: 120739257-120739257 on Build GRCh38
rs987468270 Chr.12: 120739170-120739170 on Build GRCh38
rs758765936 Chr.12: 120739208-120739208 on Build GRCh38
rs758484911 Chr.12: 120739129-120739129 on Build GRCh38
rs946248018 Chr.12: 120738991-120738991 on Build GRCh38
rs746266217 Chr.12: 120738984-120738984 on Build GRCh38
rs754459133 Chr.12: 120739182-120739182 on Build GRCh38
rs368469075 Chr.12: 120739304-120739304 on Build GRCh38
rs754369412 Chr.12: 120739105-120739105 on Build GRCh38
rs374043146 Chr.12: 120739415-120739415 on Build GRCh38
rs534848765 Chr.12: 120739005-120739005 on Build GRCh38
rs922358310 Chr.12: 120739029-120739029 on Build GRCh38
rs201218974 Chr.12: 120739279-120739279 on Build GRCh38
rs370077075 Chr.12: 120739221-120739221 on Build GRCh38
rs371468373 Chr.12: 120738983-120738983 on Build GRCh38
rs771814293 Chr.12: 120739361-120739361 on Build GRCh38
rs397744635 Chr.12: 120739004-120739005 on Build GRCh38
rs1031962576 Chr.12: 120739275-120739275 on Build GRCh38
rs995064724 Chr.12: 120739338-120739338 on Build GRCh38
rs202124189 Chr.12: 120739362-120739362 on Build GRCh38
rs35233375 Chr.12: 120739357-120739357 on Build GRCh38
rs764750642 Chr.12: 120739101-120739101 on Build GRCh38
rs904655362 Chr.12: 120739002-120739002 on Build GRCh38
rs757155952 Chr.12: 120739411-120739411 on Build GRCh38
rs775074247 Chr.12: 120739287-120739287 on Build GRCh38
rs768748973 Chr.12: 120739160-120739160 on Build GRCh38
rs796051904 Chr.12: 120739167-120739167 on Build GRCh38
rs779408255 Chr.12: 120739120-120739120 on Build GRCh38
rs371934737 Chr.12: 120739326-120739326 on Build GRCh38
rs528266975 Chr.12: 120739188-120739188 on Build GRCh38
rs190464662 Chr.12: 120739076-120739076 on Build GRCh38
rs377672631 Chr.12: 120738960-120738960 on Build GRCh38
rs371323002 Chr.12: 120739358-120739358 on Build GRCh38
rs183161718 Chr.12: 120739339-120739339 on Build GRCh38
rs970614348 Chr.12: 120739082-120739082 on Build GRCh38
rs28941773 Chr.12: 120739168-120739168 on Build GRCh38
rs778608148 Chr.12: 120739412-120739412 on Build GRCh38
rs751738987 Chr.12: 120739315-120739315 on Build GRCh38
rs28940872 Chr.12: 120739356-120739356 on Build GRCh38
rs750909218 Chr.12: 120739310-120739310 on Build GRCh38
rs541587321 Chr.12: 120739194-120739194 on Build GRCh38
rs765914571 Chr.12: 120739098-120739098 on Build GRCh38
rs374392992 Chr.12: 120739225-120739225 on Build GRCh38
rs752240673 Chr.12: 120739221-120739221 on Build GRCh38
rs942118123 Chr.12: 120738959-120738959 on Build GRCh38
rs1057517155 Chr.12: 120739141-120739141 on Build GRCh38
rs1011856639 Chr.12: 120739036-120739036 on Build GRCh38
rs773646063 Chr.12: 120739378-120739378 on Build GRCh38
rs1000626538 Chr.12: 120739004-120739004 on Build GRCh38
rs537072819 Chr.12: 120739365-120739365 on Build GRCh38
rs200165866 Chr.12: 120739313-120739313 on Build GRCh38
rs2071266 Chr.12: 120738967-120738967 on Build GRCh38
rs768733898 Chr.12: 120739176-120739176 on Build GRCh38
rs767774362 Chr.12: 120739401-120739401 on Build GRCh38
rs34289241 Chr.12: 120739131-120739132 on Build GRCh38
rs780178412 Chr.12: 120739211-120739211 on Build GRCh38
rs550519759 Chr.12: 120739258-120739258 on Build GRCh38
rs697853 Chr.12: 120739175-120739175 on Build GRCh38
rs1041882591 Chr.12: 120738995-120738995 on Build GRCh38

View Full Product Details