Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 121793020-121793523 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RHOF
Entrez Gene ID
54509
Gene Name
ras homolog family member F, filopodia associated
Gene Aliases
ARHF, RIF
UniGene
Hs.707579
Gene Chromosome Location
Chr.12: 121777754-121793688 on Build GRCh38
Transcripts
NM_019034.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6000846 c.139C>A p.H47N Chr.12: 121793239-121793239 on Build GRCh38
6206712 c.114G>A p.V38V Chr.12: 121793520-121793520 on Build GRCh38
6365928 c.223G>A p.A75T Chr.12: 121793155-121793155 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs913055643 Chr.12: 121793234-121793234 on Build GRCh38
rs572885734 Chr.12: 121793140-121793141 on Build GRCh38
rs557265714 Chr.12: 121793102-121793102 on Build GRCh38
rs772139635 Chr.12: 121793125-121793125 on Build GRCh38
rs1041890795 Chr.12: 121793487-121793487 on Build GRCh38
rs921889794 Chr.12: 121793151-121793151 on Build GRCh38
rs1024081959 Chr.12: 121793339-121793339 on Build GRCh38
rs1000363191 Chr.12: 121793293-121793293 on Build GRCh38
rs886607489 Chr.12: 121793345-121793345 on Build GRCh38
rs866780494 Chr.12: 121793275-121793275 on Build GRCh38
rs553552436 Chr.12: 121793124-121793136 on Build GRCh38
rs768028609 Chr.12: 121793210-121793210 on Build GRCh38
rs935876905 Chr.12: 121793170-121793170 on Build GRCh38
rs1017156869 Chr.12: 121793343-121793343 on Build GRCh38
rs890535814 Chr.12: 121793198-121793198 on Build GRCh38
rs559445038 Chr.12: 121793313-121793313 on Build GRCh38
rs532875831 Chr.12: 121793341-121793341 on Build GRCh38
rs575653547 Chr.12: 121793280-121793281 on Build GRCh38
rs776383493 Chr.12: 121793157-121793157 on Build GRCh38
rs760785782 Chr.12: 121793164-121793165 on Build GRCh38
rs745858833 Chr.12: 121793104-121793104 on Build GRCh38
rs762387430 Chr.12: 121793207-121793207 on Build GRCh38
rs113086400 Chr.12: 121793303-121793303 on Build GRCh38
rs957384959 Chr.12: 121793470-121793470 on Build GRCh38
rs1013872382 Chr.12: 121793308-121793308 on Build GRCh38
rs921177092 Chr.12: 121793429-121793429 on Build GRCh38
rs772229293 Chr.12: 121793472-121793472 on Build GRCh38
rs942472344 Chr.12: 121793333-121793333 on Build GRCh38
rs775509947 Chr.12: 121793132-121793132 on Build GRCh38
rs563473250 Chr.12: 121793366-121793366 on Build GRCh38
rs553640548 Chr.12: 121793337-121793338 on Build GRCh38
rs370554512 Chr.12: 121793084-121793084 on Build GRCh38
rs988707855 Chr.12: 121793489-121793489 on Build GRCh38
rs2707067 Chr.12: 121793462-121793462 on Build GRCh38
rs17522849 Chr.12: 121793138-121793138 on Build GRCh38
rs776112192 Chr.12: 121793128-121793141 on Build GRCh38
rs925980735 Chr.12: 121793168-121793168 on Build GRCh38
rs753156820 Chr.12: 121793243-121793243 on Build GRCh38
rs946097709 Chr.12: 121793478-121793478 on Build GRCh38
rs761347478 Chr.12: 121793175-121793175 on Build GRCh38
rs1042266983 Chr.12: 121793225-121793225 on Build GRCh38
rs975294020 Chr.12: 121793354-121793354 on Build GRCh38
rs932011868 Chr.12: 121793448-121793448 on Build GRCh38
rs932062667 Chr.12: 121793153-121793153 on Build GRCh38
rs545046391 Chr.12: 121793360-121793360 on Build GRCh38
rs749926378 Chr.12: 121793197-121793197 on Build GRCh38
rs575394762 Chr.12: 121793137-121793137 on Build GRCh38
rs368277328 Chr.12: 121793078-121793078 on Build GRCh38
rs750239440 Chr.12: 121793257-121793257 on Build GRCh38
rs988737599 Chr.12: 121793227-121793227 on Build GRCh38
rs1033982370 Chr.12: 121793295-121793295 on Build GRCh38
rs775591084 Chr.12: 121793427-121793427 on Build GRCh38
rs553464063 Chr.12: 121793052-121793052 on Build GRCh38
rs780170687 Chr.12: 121793501-121793501 on Build GRCh38
rs200285606 Chr.12: 121793459-121793459 on Build GRCh38
rs772426261 Chr.12: 121793124-121793124 on Build GRCh38
rs201021224 Chr.12: 121793277-121793277 on Build GRCh38
rs982650682 Chr.12: 121793342-121793342 on Build GRCh38
rs530694396 Chr.12: 121793436-121793436 on Build GRCh38
rs764856318 Chr.12: 121793194-121793194 on Build GRCh38
rs1051893052 Chr.12: 121793177-121793177 on Build GRCh38
rs962622828 Chr.12: 121793351-121793351 on Build GRCh38
rs567549599 Chr.12: 121793461-121793461 on Build GRCh38
rs777970145 Chr.12: 121793458-121793458 on Build GRCh38
rs1024486309 Chr.12: 121793416-121793416 on Build GRCh38
rs773704722 Chr.12: 121793465-121793465 on Build GRCh38

View Full Product Details