Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 122130766-122131274 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MLXIP
Entrez Gene ID
22877
Gene Name
MLX interacting protein
Gene Aliases
MIR, MONDOA, bHLHe36
UniGene
Hs.437153
Gene Chromosome Location
Chr.12: 122078728-122147347 on Build GRCh38
Transcripts
NM_014938.5 XM_006719290.3 XM_006719291.3 XM_006719292.3 XM_006719293.3 XM_006719294.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4039532 c.911-1G>A p.? Chr.12: 122130843-122130843 on Build GRCh38
936154 c.996C>T p.F332F Chr.12: 122130929-122130929 on Build GRCh38
6136081 c.972C>T p.D324D Chr.12: 122130905-122130905 on Build GRCh38
5534608 c.952C>T p.P318S Chr.12: 122130885-122130885 on Build GRCh38
7241505 c.999G>A p.Q333Q Chr.12: 122130932-122130932 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs187181749 Chr.12: 122130848-122130848 on Build GRCh38
rs991450215 Chr.12: 122130985-122130985 on Build GRCh38
rs1035918816 Chr.12: 122131180-122131180 on Build GRCh38
rs777069112 Chr.12: 122130926-122130926 on Build GRCh38
rs888713870 Chr.12: 122131094-122131094 on Build GRCh38
rs116674819 Chr.12: 122131044-122131044 on Build GRCh38
rs188795431 Chr.12: 122131237-122131237 on Build GRCh38
rs763412914 Chr.12: 122130943-122130943 on Build GRCh38
rs193059558 Chr.12: 122131248-122131248 on Build GRCh38
rs527766628 Chr.12: 122130978-122130978 on Build GRCh38
rs748749094 Chr.12: 122131125-122131125 on Build GRCh38
rs1010632922 Chr.12: 122131018-122131018 on Build GRCh38
rs747836377 Chr.12: 122131030-122131030 on Build GRCh38
rs374645537 Chr.12: 122131071-122131071 on Build GRCh38
rs962307212 Chr.12: 122131070-122131070 on Build GRCh38
rs772187741 Chr.12: 122130908-122130908 on Build GRCh38
rs755817957 Chr.12: 122130840-122130840 on Build GRCh38
rs767005501 Chr.12: 122130828-122130828 on Build GRCh38
rs1021511526 Chr.12: 122131211-122131211 on Build GRCh38
rs961892966 Chr.12: 122130865-122130865 on Build GRCh38
rs11061536 Chr.12: 122131111-122131111 on Build GRCh38
rs541623068 Chr.12: 122130953-122130953 on Build GRCh38
rs753661510 Chr.12: 122130854-122130854 on Build GRCh38
rs552604683 Chr.12: 122130998-122130998 on Build GRCh38
rs193003921 Chr.12: 122131176-122131176 on Build GRCh38
rs758316748 Chr.12: 122130972-122130972 on Build GRCh38
rs771299555 Chr.12: 122130922-122130922 on Build GRCh38
rs4758654 Chr.12: 122130968-122130968 on Build GRCh38
rs1014653130 Chr.12: 122130949-122130950 on Build GRCh38
rs558387407 Chr.12: 122130789-122130789 on Build GRCh38
rs761373942 Chr.12: 122130809-122130809 on Build GRCh38
rs951893833 Chr.12: 122131153-122131153 on Build GRCh38
rs564445294 Chr.12: 122131015-122131015 on Build GRCh38
rs982003069 Chr.12: 122130841-122130841 on Build GRCh38
rs765007357 Chr.12: 122130961-122130961 on Build GRCh38
rs576748074 Chr.12: 122130801-122130801 on Build GRCh38
rs746145643 Chr.12: 122130980-122130980 on Build GRCh38
rs376004273 Chr.12: 122130959-122130959 on Build GRCh38
rs749961143 Chr.12: 122130950-122130950 on Build GRCh38
rs920388168 Chr.12: 122131139-122131139 on Build GRCh38
rs747212632 Chr.12: 122130911-122130911 on Build GRCh38
rs921472680 Chr.12: 122131056-122131056 on Build GRCh38
rs368270656 Chr.12: 122130910-122130910 on Build GRCh38
rs897576091 Chr.12: 122131158-122131158 on Build GRCh38
rs777950065 Chr.12: 122130974-122130974 on Build GRCh38
rs547997700 Chr.12: 122131092-122131092 on Build GRCh38
rs778921300 Chr.12: 122130875-122130875 on Build GRCh38
rs750049009 Chr.12: 122130831-122130831 on Build GRCh38
rs1005911334 Chr.12: 122130948-122130948 on Build GRCh38
rs568760093 Chr.12: 122131055-122131055 on Build GRCh38
rs368999745 Chr.12: 122130960-122130960 on Build GRCh38
rs1013183021 Chr.12: 122131197-122131197 on Build GRCh38
rs911331042 Chr.12: 122131230-122131230 on Build GRCh38
rs185130260 Chr.12: 122131194-122131194 on Build GRCh38
rs765779065 Chr.12: 122130798-122130798 on Build GRCh38
rs754782825 Chr.12: 122130975-122130975 on Build GRCh38
rs1048618818 Chr.12: 122131076-122131076 on Build GRCh38
rs1023409660 Chr.12: 122131036-122131036 on Build GRCh38
rs774128447 Chr.12: 122131236-122131236 on Build GRCh38
rs550206037 Chr.12: 122131046-122131046 on Build GRCh38
rs1002659795 Chr.12: 122131179-122131179 on Build GRCh38
rs1048468850 Chr.12: 122130955-122130955 on Build GRCh38
rs1023273719 Chr.12: 122130820-122130820 on Build GRCh38
rs536185412 Chr.12: 122131060-122131060 on Build GRCh38
rs969057544 Chr.12: 122130824-122130824 on Build GRCh38
rs780909280 Chr.12: 122130984-122130984 on Build GRCh38
rs201047061 Chr.12: 122130864-122130864 on Build GRCh38
rs765560664 Chr.12: 122130936-122130936 on Build GRCh38
rs539669771 Chr.12: 122131186-122131186 on Build GRCh38
rs774168637 Chr.12: 122130973-122130974 on Build GRCh38
rs767035595 Chr.12: 122130946-122130946 on Build GRCh38
rs972007259 Chr.12: 122131131-122131131 on Build GRCh38
rs776063027 Chr.12: 122130942-122130942 on Build GRCh38
rs149214437 Chr.12: 122130940-122130940 on Build GRCh38
rs977403274 Chr.12: 122131019-122131019 on Build GRCh38

View Full Product Details