Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 122206627-122207117 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GNT4
Entrez Gene ID
79369
Gene Name
UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 4
Gene Aliases
B3GN-T4, beta3Gn-T4
UniGene
Hs.363315
Gene Chromosome Location
Chr.12: 122203681-122208135 on Build GRCh38
Transcripts
NM_030765.2 XM_011538732.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7314657 c.682G>T p.V228L Chr.12: 122206933-122206933 on Build GRCh38
1184318 c.725T>C p.L242P Chr.12: 122206976-122206976 on Build GRCh38
3968043 c.562G>A p.E188K Chr.12: 122206813-122206813 on Build GRCh38
1184319 c.679A>G p.N227D Chr.12: 122206930-122206930 on Build GRCh38
116246 c.538G>A p.D180N Chr.12: 122206789-122206789 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1023352894 Chr.12: 122206670-122206670 on Build GRCh38
rs757251464 Chr.12: 122206755-122206755 on Build GRCh38
rs768800797 Chr.12: 122206791-122206791 on Build GRCh38
rs771029308 Chr.12: 122206703-122206703 on Build GRCh38
rs765215253 Chr.12: 122207011-122207011 on Build GRCh38
rs878914918 Chr.12: 122206678-122206678 on Build GRCh38
rs781295138 Chr.12: 122206763-122206763 on Build GRCh38
rs924671629 Chr.12: 122206772-122206772 on Build GRCh38
rs187124215 Chr.12: 122206818-122206818 on Build GRCh38
rs572271254 Chr.12: 122207047-122207047 on Build GRCh38
rs966234662 Chr.12: 122206974-122206974 on Build GRCh38
rs540022301 Chr.12: 122206718-122206718 on Build GRCh38
rs897573945 Chr.12: 122206873-122206873 on Build GRCh38
rs760130596 Chr.12: 122206671-122206671 on Build GRCh38
rs758241880 Chr.12: 122206689-122206689 on Build GRCh38
rs145634810 Chr.12: 122206933-122206933 on Build GRCh38
rs199538224 Chr.12: 122206935-122206935 on Build GRCh38
rs369076213 Chr.12: 122206832-122206832 on Build GRCh38
rs762277340 Chr.12: 122206719-122206719 on Build GRCh38
rs750759224 Chr.12: 122207091-122207091 on Build GRCh38
rs757917533 Chr.12: 122206952-122206962 on Build GRCh38
rs1011006255 Chr.12: 122206667-122206667 on Build GRCh38
rs777519885 Chr.12: 122206691-122206691 on Build GRCh38
rs965409107 Chr.12: 122206937-122206937 on Build GRCh38
rs751755393 Chr.12: 122207025-122207025 on Build GRCh38
rs767399145 Chr.12: 122206880-122206880 on Build GRCh38
rs749505997 Chr.12: 122206778-122206778 on Build GRCh38
rs775048350 Chr.12: 122206861-122206861 on Build GRCh38
rs576192434 Chr.12: 122206969-122206969 on Build GRCh38
rs373034990 Chr.12: 122206662-122206662 on Build GRCh38
rs748525931 Chr.12: 122206805-122206805 on Build GRCh38
rs150959436 Chr.12: 122206767-122206767 on Build GRCh38
rs878962745 Chr.12: 122206696-122206696 on Build GRCh38
rs142538376 Chr.12: 122206717-122206717 on Build GRCh38
rs775486046 Chr.12: 122207012-122207012 on Build GRCh38
rs745313481 Chr.12: 122206771-122206772 on Build GRCh38
rs771404601 Chr.12: 122206858-122206858 on Build GRCh38
rs774634761 Chr.12: 122207057-122207057 on Build GRCh38
rs762658951 Chr.12: 122206820-122206820 on Build GRCh38
rs368416594 Chr.12: 122206712-122206712 on Build GRCh38
rs763020841 Chr.12: 122207021-122207021 on Build GRCh38
rs770087243 Chr.12: 122206710-122206710 on Build GRCh38
rs768079578 Chr.12: 122206720-122206720 on Build GRCh38
rs769648867 Chr.12: 122207060-122207060 on Build GRCh38
rs764060683 Chr.12: 122207064-122207064 on Build GRCh38
rs148849316 Chr.12: 122206702-122206702 on Build GRCh38
rs777391992 Chr.12: 122207046-122207046 on Build GRCh38
rs767112200 Chr.12: 122206955-122206955 on Build GRCh38
rs939477759 Chr.12: 122207078-122207078 on Build GRCh38
rs745507521 Chr.12: 122206937-122206938 on Build GRCh38
rs377393964 Chr.12: 122206899-122206899 on Build GRCh38
rs369992469 Chr.12: 122206884-122206884 on Build GRCh38
rs765798528 Chr.12: 122206689-122206689 on Build GRCh38
rs758995947 Chr.12: 122206815-122206815 on Build GRCh38
rs779112409 Chr.12: 122206853-122206853 on Build GRCh38
rs746769802 Chr.12: 122206993-122206993 on Build GRCh38
rs770153583 Chr.12: 122206911-122206928 on Build GRCh38
rs147657286 Chr.12: 122206881-122206881 on Build GRCh38
rs773738674 Chr.12: 122206811-122206811 on Build GRCh38
rs375511104 Chr.12: 122206744-122206744 on Build GRCh38
rs751346265 Chr.12: 122206829-122206829 on Build GRCh38
rs199695726 Chr.12: 122206857-122206857 on Build GRCh38
rs781558278 Chr.12: 122206923-122206923 on Build GRCh38
rs747386259 Chr.12: 122206918-122206918 on Build GRCh38
rs780720644 Chr.12: 122207039-122207039 on Build GRCh38
rs775335390 Chr.12: 122206866-122206866 on Build GRCh38
rs753756714 Chr.12: 122206760-122206761 on Build GRCh38
rs763914539 Chr.12: 122206748-122206748 on Build GRCh38
rs1056237275 Chr.12: 122207081-122207081 on Build GRCh38
rs780092537 Chr.12: 122206769-122206769 on Build GRCh38
rs758601645 Chr.12: 122206950-122206950 on Build GRCh38
rs977232099 Chr.12: 122206777-122206777 on Build GRCh38
rs926801135 Chr.12: 122207071-122207071 on Build GRCh38
rs1002552024 Chr.12: 122207000-122207000 on Build GRCh38
rs760059825 Chr.12: 122206813-122206815 on Build GRCh38
rs768355359 Chr.12: 122206874-122206875 on Build GRCh38
rs376839421 Chr.12: 122206968-122206968 on Build GRCh38
rs756677704 Chr.12: 122207033-122207033 on Build GRCh38
rs972014071 Chr.12: 122206739-122206739 on Build GRCh38
rs1057256645 Chr.12: 122207005-122207005 on Build GRCh38
rs772447869 Chr.12: 122206855-122206855 on Build GRCh38
rs774180580 Chr.12: 122206874-122206874 on Build GRCh38
rs746674091 Chr.12: 122206855-122206856 on Build GRCh38
rs764445156 Chr.12: 122206661-122206661 on Build GRCh38
rs778104699 Chr.12: 122206909-122206909 on Build GRCh38
rs761938290 Chr.12: 122207075-122207075 on Build GRCh38
rs752837963 Chr.12: 122206946-122206946 on Build GRCh38
rs763392167 Chr.12: 122206659-122206659 on Build GRCh38
rs773132378 Chr.12: 122206696-122206696 on Build GRCh38
rs763189832 Chr.12: 122206714-122206714 on Build GRCh38
rs947010499 Chr.12: 122207058-122207058 on Build GRCh38
rs776062180 Chr.12: 122206724-122206724 on Build GRCh38
rs200601197 Chr.12: 122206751-122206751 on Build GRCh38
rs553617934 Chr.12: 122206906-122206906 on Build GRCh38
rs529236293 Chr.12: 122207045-122207045 on Build GRCh38
rs774168455 Chr.12: 122206824-122206824 on Build GRCh38
rs748471148 Chr.12: 122206854-122206854 on Build GRCh38
rs779055570 Chr.12: 122206901-122206901 on Build GRCh38
rs767745310 Chr.12: 122207031-122207031 on Build GRCh38
rs753888870 Chr.12: 122206891-122206891 on Build GRCh38
rs956743677 Chr.12: 122207003-122207003 on Build GRCh38
rs372233552 Chr.12: 122206656-122206656 on Build GRCh38
rs867531607 Chr.12: 122206687-122206687 on Build GRCh38
rs148329384 Chr.12: 122207063-122207063 on Build GRCh38
rs766472116 Chr.12: 122206703-122206733 on Build GRCh38
rs746006136 Chr.12: 122206708-122206708 on Build GRCh38
rs756056900 Chr.12: 122206836-122206836 on Build GRCh38
rs754611702 Chr.12: 122206681-122206681 on Build GRCh38
rs977437202 Chr.12: 122206771-122206771 on Build GRCh38
rs765268501 Chr.12: 122206940-122206940 on Build GRCh38
rs760658090 Chr.12: 122206886-122206886 on Build GRCh38
rs777981592 Chr.12: 122206959-122206959 on Build GRCh38
rs759462178 Chr.12: 122207009-122207009 on Build GRCh38
rs778960401 Chr.12: 122206769-122206770 on Build GRCh38
rs761654592 Chr.12: 122206875-122206875 on Build GRCh38
rs762387783 Chr.12: 122206870-122206870 on Build GRCh38
rs776974249 Chr.12: 122206886-122206886 on Build GRCh38
rs756552864 Chr.12: 122207093-122207093 on Build GRCh38
rs747018135 Chr.12: 122206651-122206651 on Build GRCh38
rs375787732 Chr.12: 122206848-122206848 on Build GRCh38
rs773127810 Chr.12: 122206932-122206932 on Build GRCh38
rs376862803 Chr.12: 122206765-122206765 on Build GRCh38
rs777417308 Chr.12: 122206859-122206859 on Build GRCh38
rs767839496 Chr.12: 122207082-122207082 on Build GRCh38
rs138745438 Chr.12: 122206896-122206896 on Build GRCh38
rs147782078 Chr.12: 122206648-122206648 on Build GRCh38
rs781413400 Chr.12: 122206706-122206706 on Build GRCh38
rs751811961 Chr.12: 122206715-122206716 on Build GRCh38
rs766286373 Chr.12: 122206889-122206889 on Build GRCh38
rs757232979 Chr.12: 122206768-122206769 on Build GRCh38
rs182526849 Chr.12: 122206652-122206652 on Build GRCh38
rs752366862 Chr.12: 122206683-122206683 on Build GRCh38
rs370749355 Chr.12: 122206985-122206985 on Build GRCh38
rs777064164 Chr.12: 122206682-122206683 on Build GRCh38
rs868683322 Chr.12: 122207076-122207076 on Build GRCh38
rs764263985 Chr.12: 122207023-122207023 on Build GRCh38
rs201025149 Chr.12: 122206834-122206834 on Build GRCh38
rs753386412 Chr.12: 122206676-122206676 on Build GRCh38
rs747595280 Chr.12: 122207043-122207043 on Build GRCh38
rs773351083 Chr.12: 122206700-122206700 on Build GRCh38
rs774899917 Chr.12: 122206819-122206819 on Build GRCh38
rs373879514 Chr.12: 122206908-122206908 on Build GRCh38
rs944132769 Chr.12: 122206806-122206806 on Build GRCh38
rs1021497816 Chr.12: 122207019-122207020 on Build GRCh38
rs748550610 Chr.12: 122206908-122206909 on Build GRCh38
rs762293588 Chr.12: 122206683-122206684 on Build GRCh38
rs745673381 Chr.12: 122207055-122207055 on Build GRCh38
rs1056583190 Chr.12: 122206929-122206929 on Build GRCh38
rs543502597 Chr.12: 122206980-122206980 on Build GRCh38
rs755916031 Chr.12: 122206728-122206729 on Build GRCh38
rs765853732 Chr.12: 122206674-122206674 on Build GRCh38
rs140444368 Chr.12: 122206924-122206924 on Build GRCh38
rs59453864 Chr.12: 122206994-122206994 on Build GRCh38
rs942799770 Chr.12: 122206770-122206770 on Build GRCh38
rs763118566 Chr.12: 122206696-122206697 on Build GRCh38
rs888718833 Chr.12: 122206837-122206837 on Build GRCh38
rs757597567 Chr.12: 122206967-122206967 on Build GRCh38
rs774533386 Chr.12: 122207070-122207070 on Build GRCh38
rs866253298 Chr.12: 122206936-122206937 on Build GRCh38
rs373036509 Chr.12: 122207052-122207052 on Build GRCh38
rs772500588 Chr.12: 122206808-122206808 on Build GRCh38
rs771793938 Chr.12: 122206992-122206992 on Build GRCh38
rs750868441 Chr.12: 122207032-122207032 on Build GRCh38
rs374005009 Chr.12: 122207094-122207094 on Build GRCh38
rs988575747 Chr.12: 122207042-122207042 on Build GRCh38
rs749396322 Chr.12: 122206840-122206840 on Build GRCh38
rs569095788 Chr.12: 122206743-122206743 on Build GRCh38
rs201793683 Chr.12: 122206773-122206773 on Build GRCh38
rs769430362 Chr.12: 122206818-122206820 on Build GRCh38
rs775788859 Chr.12: 122206711-122206711 on Build GRCh38
rs759565227 Chr.12: 122206939-122206939 on Build GRCh38
rs1034731747 Chr.12: 122206704-122206704 on Build GRCh38
rs759796815 Chr.12: 122206655-122206655 on Build GRCh38
rs757550904 Chr.12: 122207026-122207026 on Build GRCh38
rs144265607 Chr.12: 122207053-122207053 on Build GRCh38
rs766494063 Chr.12: 122206955-122206983 on Build GRCh38
rs762937151 Chr.12: 122206753-122206753 on Build GRCh38
rs138266947 Chr.12: 122206986-122206986 on Build GRCh38
rs200448613 Chr.12: 122206921-122206921 on Build GRCh38
rs778641187 Chr.12: 122206682-122206682 on Build GRCh38
rs751887736 Chr.12: 122206963-122206963 on Build GRCh38
rs778158051 Chr.12: 122206856-122206856 on Build GRCh38
rs774774427 Chr.12: 122206792-122206792 on Build GRCh38
rs780153302 Chr.12: 122206839-122206839 on Build GRCh38
rs951972920 Chr.12: 122206957-122206957 on Build GRCh38
rs763972147 Chr.12: 122206758-122206759 on Build GRCh38
rs753741736 Chr.12: 122206938-122206938 on Build GRCh38
rs550148151 Chr.12: 122206650-122206650 on Build GRCh38
rs910089724 Chr.12: 122207084-122207084 on Build GRCh38
rs866905152 Chr.12: 122206716-122206716 on Build GRCh38
rs761527942 Chr.12: 122206927-122206927 on Build GRCh38

View Full Product Details