Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 113122578-113123092 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
F10
Entrez Gene ID
2159
Gene Name
coagulation factor X
Gene Aliases
FX, FXA
UniGene
Hs.361463
Gene Chromosome Location
Chr.13: 113122799-113149529 on Build GRCh38
Transcripts
NM_000504.3 NM_001312674.1 NM_001312675.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
695943 c.38C>T p.S13F Chr.13: 113122893-113122893 on Build GRCh38
6323545 c.17A>T p.H6L Chr.13: 113122872-113122872 on Build GRCh38
5578518 c.6G>A p.G2G Chr.13: 113122861-113122861 on Build GRCh38
5599491 c.64G>A p.E22K Chr.13: 113122919-113122919 on Build GRCh38
3813459 c.8G>A p.R3H Chr.13: 113122863-113122863 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs765916051 Chr.13: 113122812-113122812 on Build GRCh38
rs763935393 Chr.13: 113122976-113122976 on Build GRCh38
rs3212993 Chr.13: 113122796-113122796 on Build GRCh38
rs545351284 Chr.13: 113123020-113123020 on Build GRCh38
rs5963 Chr.13: 113122874-113122874 on Build GRCh38
rs941799314 Chr.13: 113122656-113122656 on Build GRCh38
rs3212994 Chr.13: 113122816-113122816 on Build GRCh38
rs3829390 Chr.13: 113122792-113122792 on Build GRCh38
rs142098501 Chr.13: 113122981-113122981 on Build GRCh38
rs147578943 Chr.13: 113122699-113122699 on Build GRCh38
rs749360337 Chr.13: 113122836-113122836 on Build GRCh38
rs78778503 Chr.13: 113122795-113122795 on Build GRCh38
rs566093476 Chr.13: 113122684-113122684 on Build GRCh38
rs757309231 Chr.13: 113122917-113122917 on Build GRCh38
rs1026200798 Chr.13: 113122863-113122863 on Build GRCh38
rs386775038 Chr.13: 113122633-113122635 on Build GRCh38
rs1057343243 Chr.13: 113122719-113122719 on Build GRCh38
rs763524697 Chr.13: 113122601-113122601 on Build GRCh38
rs778851791 Chr.13: 113122957-113122957 on Build GRCh38
rs747352480 Chr.13: 113122935-113122935 on Build GRCh38
rs777143885 Chr.13: 113122938-113122938 on Build GRCh38
rs780456249 Chr.13: 113122933-113122933 on Build GRCh38
rs1039641348 Chr.13: 113123054-113123054 on Build GRCh38
rs756166351 Chr.13: 113122914-113122914 on Build GRCh38
rs375367218 Chr.13: 113122864-113122864 on Build GRCh38
rs778053228 Chr.13: 113122835-113122835 on Build GRCh38
rs1001878601 Chr.13: 113122757-113122757 on Build GRCh38
rs778995263 Chr.13: 113122920-113122920 on Build GRCh38
rs1043072547 Chr.13: 113122752-113122752 on Build GRCh38
rs115234520 Chr.13: 113122650-113122650 on Build GRCh38
rs1034291906 Chr.13: 113122798-113122798 on Build GRCh38
rs575581833 Chr.13: 113122709-113122709 on Build GRCh38
rs375632041 Chr.13: 113122887-113122887 on Build GRCh38
rs757542867 Chr.13: 113122846-113122846 on Build GRCh38
rs979830268 Chr.13: 113122693-113122693 on Build GRCh38
rs775469294 Chr.13: 113122961-113122961 on Build GRCh38
rs756113308 Chr.13: 113122828-113122828 on Build GRCh38
rs765863253 Chr.13: 113122876-113122876 on Build GRCh38
rs866979778 Chr.13: 113122834-113122834 on Build GRCh38
rs372884946 Chr.13: 113122877-113122877 on Build GRCh38
rs1042880057 Chr.13: 113122714-113122714 on Build GRCh38
rs909384773 Chr.13: 113122839-113122839 on Build GRCh38
rs145576121 Chr.13: 113122654-113122654 on Build GRCh38
rs751183743 Chr.13: 113122813-113122813 on Build GRCh38
rs753790195 Chr.13: 113122916-113122916 on Build GRCh38
rs773904083 Chr.13: 113122959-113122959 on Build GRCh38
rs904412882 Chr.13: 113122731-113122731 on Build GRCh38
rs879138874 Chr.13: 113123019-113123019 on Build GRCh38
rs772338742 Chr.13: 113122860-113122860 on Build GRCh38
rs60349518 Chr.13: 113123036-113123036 on Build GRCh38
rs779037571 Chr.13: 113122847-113122847 on Build GRCh38
rs370154986 Chr.13: 113122912-113122912 on Build GRCh38
rs373304646 Chr.13: 113122740-113122740 on Build GRCh38
rs3211718 Chr.13: 113122915-113122915 on Build GRCh38
rs1051453286 Chr.13: 113122674-113122674 on Build GRCh38
rs904528323 Chr.13: 113122791-113122791 on Build GRCh38
rs185296189 Chr.13: 113122603-113122603 on Build GRCh38
rs200661303 Chr.13: 113122818-113122818 on Build GRCh38
rs550132930 Chr.13: 113123053-113123053 on Build GRCh38
rs945918779 Chr.13: 113122732-113122732 on Build GRCh38
rs564479219 Chr.13: 113123035-113123035 on Build GRCh38
rs752683143 Chr.13: 113122820-113122820 on Build GRCh38
rs1056107957 Chr.13: 113123008-113123008 on Build GRCh38
rs942724907 Chr.13: 113123022-113123022 on Build GRCh38
rs767484560 Chr.13: 113122899-113122899 on Build GRCh38
rs371824280 Chr.13: 113122651-113122651 on Build GRCh38
rs3211717 Chr.13: 113122635-113122635 on Build GRCh38
rs753698256 Chr.13: 113122831-113122831 on Build GRCh38
rs376192608 Chr.13: 113122974-113122974 on Build GRCh38
rs758550534 Chr.13: 113122864-113122864 on Build GRCh38
rs758643474 Chr.13: 113122929-113122929 on Build GRCh38
rs114136715 Chr.13: 113122602-113122602 on Build GRCh38
rs1015960957 Chr.13: 113122897-113122897 on Build GRCh38
rs374448450 Chr.13: 113122939-113122939 on Build GRCh38
rs150082798 Chr.13: 113122986-113122986 on Build GRCh38
rs769138821 Chr.13: 113122867-113122867 on Build GRCh38
rs1025058864 Chr.13: 113122825-113122825 on Build GRCh38
rs149972574 Chr.13: 113122862-113122862 on Build GRCh38
rs7319750 Chr.13: 113123018-113123018 on Build GRCh38
rs563964 Chr.13: 113122633-113122633 on Build GRCh38
rs913046913 Chr.13: 113122980-113122980 on Build GRCh38
rs541896513 Chr.13: 113122960-113122960 on Build GRCh38
rs1013552421 Chr.13: 113123026-113123026 on Build GRCh38
rs746084152 Chr.13: 113122926-113122926 on Build GRCh38
rs570761117 Chr.13: 113122632-113122632 on Build GRCh38
rs746056006 Chr.13: 113122852-113122852 on Build GRCh38
rs772748215 Chr.13: 113122870-113122870 on Build GRCh38
rs891713304 Chr.13: 113122730-113122730 on Build GRCh38
rs1042934015 Chr.13: 113122989-113122989 on Build GRCh38

View Full Product Details