Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 113503410-113503910 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMCO3
Entrez Gene ID
55002
Gene Name
transmembrane and coiled-coil domains 3
Gene Aliases
C13orf11
UniGene
Hs.317593
Gene Chromosome Location
Chr.13: 113490993-113550229 on Build GRCh38
Transcripts
NM_017905.4 XM_006719969.1 XM_011537498.2 XM_011537501.2 XM_011537502.2 XM_011537504.2 XM_011537506.2 XM_011537507.2 XM_011537509.2 XM_017020629.1 XM_017020630.1 XM_017020631.1 XM_017020632.1 XM_017020633.1 XM_017020634.1 XM_017020635.1 XM_017020636.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3711007 c.994C>G p.L332V Chr.13: 113503544-113503544 on Build GRCh38
2264401 c.998T>C p.F333S Chr.13: 113503548-113503548 on Build GRCh38
7109671 c.1000C>G p.L334V Chr.13: 113503550-113503550 on Build GRCh38
1365608 c.1016T>G p.F339C Chr.13: 113503566-113503566 on Build GRCh38
6989763 c.1000C>T p.L334F Chr.13: 113503550-113503550 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756357693 Chr.13: 113503535-113503535 on Build GRCh38
rs942498121 Chr.13: 113503563-113503563 on Build GRCh38
rs772924729 Chr.13: 113503451-113503451 on Build GRCh38
rs555488323 Chr.13: 113503558-113503558 on Build GRCh38
rs1034412618 Chr.13: 113503771-113503771 on Build GRCh38
rs751130803 Chr.13: 113503503-113503503 on Build GRCh38
rs9805651 Chr.13: 113503763-113503763 on Build GRCh38
rs969202395 Chr.13: 113503762-113503762 on Build GRCh38
rs892392877 Chr.13: 113503712-113503712 on Build GRCh38
rs35309069 Chr.13: 113503731-113503731 on Build GRCh38
rs956297375 Chr.13: 113503809-113503809 on Build GRCh38
rs747802271 Chr.13: 113503513-113503513 on Build GRCh38
rs914760666 Chr.13: 113503840-113503840 on Build GRCh38
rs776592818 Chr.13: 113503851-113503851 on Build GRCh38
rs762063252 Chr.13: 113503579-113503579 on Build GRCh38
rs772284807 Chr.13: 113503546-113503546 on Build GRCh38
rs764570900 Chr.13: 113503485-113503485 on Build GRCh38
rs529386315 Chr.13: 113503825-113503825 on Build GRCh38
rs959999986 Chr.13: 113503772-113503772 on Build GRCh38
rs372564522 Chr.13: 113503631-113503631 on Build GRCh38
rs111803871 Chr.13: 113503663-113503663 on Build GRCh38
rs188600546 Chr.13: 113503714-113503714 on Build GRCh38
rs769571068 Chr.13: 113503446-113503446 on Build GRCh38
rs543461125 Chr.13: 113503544-113503544 on Build GRCh38
rs778034390 Chr.13: 113503626-113503627 on Build GRCh38
rs879034481 Chr.13: 113503605-113503605 on Build GRCh38
rs148852405 Chr.13: 113503783-113503783 on Build GRCh38
rs780887663 Chr.13: 113503508-113503508 on Build GRCh38
rs1040716400 Chr.13: 113503595-113503595 on Build GRCh38
rs754432050 Chr.13: 113503504-113503504 on Build GRCh38
rs918851139 Chr.13: 113503528-113503528 on Build GRCh38
rs752965387 Chr.13: 113503479-113503479 on Build GRCh38
rs193191708 Chr.13: 113503784-113503784 on Build GRCh38
rs764358380 Chr.13: 113503851-113503852 on Build GRCh38
rs762957812 Chr.13: 113503452-113503452 on Build GRCh38
rs764372730 Chr.13: 113503571-113503571 on Build GRCh38
rs1022170717 Chr.13: 113503715-113503715 on Build GRCh38
rs568210899 Chr.13: 113503803-113503803 on Build GRCh38
rs774518977 Chr.13: 113503459-113503459 on Build GRCh38
rs754207836 Chr.13: 113503492-113503492 on Build GRCh38
rs759500380 Chr.13: 113503461-113503461 on Build GRCh38
rs374929654 Chr.13: 113503470-113503470 on Build GRCh38
rs541081213 Chr.13: 113503499-113503499 on Build GRCh38
rs541220305 Chr.13: 113503655-113503655 on Build GRCh38
rs775657353 Chr.13: 113503549-113503549 on Build GRCh38
rs774130442 Chr.13: 113503532-113503532 on Build GRCh38
rs9805588 Chr.13: 113503531-113503531 on Build GRCh38
rs113066198 Chr.13: 113503879-113503879 on Build GRCh38
rs951837567 Chr.13: 113503689-113503689 on Build GRCh38
rs757814504 Chr.13: 113503495-113503495 on Build GRCh38
rs774006937 Chr.13: 113503609-113503609 on Build GRCh38
rs770720108 Chr.13: 113503455-113503455 on Build GRCh38
rs1048557423 Chr.13: 113503534-113503534 on Build GRCh38
rs777564085 Chr.13: 113503518-113503518 on Build GRCh38
rs2260011 Chr.13: 113503637-113503637 on Build GRCh38
rs143472577 Chr.13: 113503789-113503789 on Build GRCh38
rs758872137 Chr.13: 113503626-113503626 on Build GRCh38
rs528275779 Chr.13: 113503527-113503527 on Build GRCh38
rs369368431 Chr.13: 113503591-113503591 on Build GRCh38
rs766332325 Chr.13: 113503835-113503835 on Build GRCh38
rs370509029 Chr.13: 113503441-113503441 on Build GRCh38
rs914607917 Chr.13: 113503454-113503454 on Build GRCh38
rs750889375 Chr.13: 113503617-113503617 on Build GRCh38
rs752153228 Chr.13: 113503632-113503632 on Build GRCh38
rs760918217 Chr.13: 113503482-113503482 on Build GRCh38
rs79532189 Chr.13: 113503779-113503779 on Build GRCh38

View Full Product Details