Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 114298705-114299247 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
UPF3A
Entrez Gene ID
65110
Gene Name
UPF3 regulator of nonsense transcripts homolog A (yeast)
Gene Aliases
HUPF3A, RENT3A, UPF3
UniGene
Hs.533855
Gene Chromosome Location
Chr.13: 114281584-114305817 on Build GRCh38
Transcripts
NM_023011.3 NM_080687.2 XM_005266202.4 XM_006719991.3 XM_011534844.1 XM_011534845.2 XM_011534847.2 XM_011534848.2 XM_017020709.1 XM_017020710.1 XM_017020711.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5470130 c.901A>G p.R301G Chr.13: 114298894-114298894 on Build GRCh38
3667262 c.976A>G p.M326V Chr.13: 114298969-114298969 on Build GRCh38
6565320 c.971G>T p.R324M Chr.13: 114298964-114298964 on Build GRCh38
4617433 c.931A>G p.K311E Chr.13: 114298924-114298924 on Build GRCh38
3467477 c.982G>A p.G328S Chr.13: 114298975-114298975 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759009873 Chr.13: 114299016-114299018 on Build GRCh38
rs1017449423 Chr.13: 114299206-114299206 on Build GRCh38
rs557490786 Chr.13: 114299117-114299117 on Build GRCh38
rs28539632 Chr.13: 114299207-114299207 on Build GRCh38
rs1002972951 Chr.13: 114299120-114299120 on Build GRCh38
rs116367223 Chr.13: 114299175-114299175 on Build GRCh38
rs1050361797 Chr.13: 114299143-114299143 on Build GRCh38
rs765321568 Chr.13: 114299031-114299031 on Build GRCh38
rs777308493 Chr.13: 114298910-114298910 on Build GRCh38
rs148163540 Chr.13: 114299051-114299051 on Build GRCh38
rs1046384978 Chr.13: 114298824-114298824 on Build GRCh38
rs370734483 Chr.13: 114298935-114298935 on Build GRCh38
rs770816155 Chr.13: 114298817-114298817 on Build GRCh38
rs531937279 Chr.13: 114299000-114299000 on Build GRCh38
rs940687838 Chr.13: 114298846-114298846 on Build GRCh38
rs377378655 Chr.13: 114298966-114298966 on Build GRCh38
rs775808542 Chr.13: 114298945-114298945 on Build GRCh38
rs746173465 Chr.13: 114298888-114298888 on Build GRCh38
rs370451114 Chr.13: 114298977-114298977 on Build GRCh38
rs377221810 Chr.13: 114298812-114298812 on Build GRCh38
rs368928675 Chr.13: 114299029-114299029 on Build GRCh38
rs3993409 Chr.13: 114298969-114298969 on Build GRCh38
rs3752107 Chr.13: 114298948-114298948 on Build GRCh38
rs756225660 Chr.13: 114298815-114298815 on Build GRCh38
rs560113415 Chr.13: 114298867-114298867 on Build GRCh38
rs764287690 Chr.13: 114298951-114298951 on Build GRCh38
rs111532344 Chr.13: 114298944-114298944 on Build GRCh38
rs372827451 Chr.13: 114298800-114298800 on Build GRCh38
rs979734641 Chr.13: 114299189-114299189 on Build GRCh38
rs958254001 Chr.13: 114298741-114298741 on Build GRCh38
rs757773931 Chr.13: 114298858-114298858 on Build GRCh38
rs143253585 Chr.13: 114298960-114298960 on Build GRCh38
rs999876656 Chr.13: 114299209-114299209 on Build GRCh38
rs779342510 Chr.13: 114298860-114298860 on Build GRCh38
rs775418164 Chr.13: 114299006-114299006 on Build GRCh38
rs536822514 Chr.13: 114299156-114299156 on Build GRCh38
rs558847333 Chr.13: 114299162-114299162 on Build GRCh38
rs770888169 Chr.13: 114298939-114298939 on Build GRCh38
rs199704939 Chr.13: 114299206-114299207 on Build GRCh38
rs577051580 Chr.13: 114299191-114299191 on Build GRCh38
rs761811674 Chr.13: 114298954-114298954 on Build GRCh38
rs183120558 Chr.13: 114299224-114299224 on Build GRCh38
rs901481458 Chr.13: 114298869-114298869 on Build GRCh38
rs943077313 Chr.13: 114298855-114298855 on Build GRCh38
rs570837264 Chr.13: 114298770-114298773 on Build GRCh38
rs112144346 Chr.13: 114299034-114299034 on Build GRCh38
rs141968422 Chr.13: 114299128-114299128 on Build GRCh38
rs774448479 Chr.13: 114298941-114298941 on Build GRCh38
rs2701318 Chr.13: 114299067-114299067 on Build GRCh38
rs3993410 Chr.13: 114298984-114298984 on Build GRCh38
rs750549688 Chr.13: 114299050-114299050 on Build GRCh38
rs750791778 Chr.13: 114298871-114298882 on Build GRCh38
rs916681990 Chr.13: 114298766-114298766 on Build GRCh38
rs1036163298 Chr.13: 114299127-114299127 on Build GRCh38
rs74116886 Chr.13: 114299064-114299064 on Build GRCh38
rs747818618 Chr.13: 114298908-114298908 on Build GRCh38
rs1006852452 Chr.13: 114299075-114299075 on Build GRCh38
rs958502923 Chr.13: 114298976-114298976 on Build GRCh38
rs778956714 Chr.13: 114298989-114298989 on Build GRCh38
rs143806139 Chr.13: 114298864-114298864 on Build GRCh38
rs922969648 Chr.13: 114299063-114299063 on Build GRCh38
rs748816983 Chr.13: 114298932-114298932 on Build GRCh38
rs7999630 Chr.13: 114299007-114299007 on Build GRCh38
rs745825993 Chr.13: 114298999-114298999 on Build GRCh38
rs117024307 Chr.13: 114298837-114298837 on Build GRCh38
rs913816164 Chr.13: 114299001-114299001 on Build GRCh38
rs944507545 Chr.13: 114299173-114299173 on Build GRCh38
rs371335717 Chr.13: 114298798-114298798 on Build GRCh38
rs991371174 Chr.13: 114298758-114298758 on Build GRCh38
rs1047954925 Chr.13: 114298929-114298929 on Build GRCh38
rs755787826 Chr.13: 114298970-114298970 on Build GRCh38
rs1010231519 Chr.13: 114299142-114299142 on Build GRCh38
rs765709351 Chr.13: 114298959-114298959 on Build GRCh38
rs376399019 Chr.13: 114298907-114298907 on Build GRCh38
rs139333511 Chr.13: 114298975-114298975 on Build GRCh38
rs768861518 Chr.13: 114299026-114299026 on Build GRCh38
rs374688050 Chr.13: 114298953-114298953 on Build GRCh38
rs368615697 Chr.13: 114299028-114299028 on Build GRCh38
rs759466583 Chr.13: 114298799-114298799 on Build GRCh38
rs7982070 Chr.13: 114299210-114299210 on Build GRCh38
rs758765255 Chr.13: 114298895-114298895 on Build GRCh38
rs190712941 Chr.13: 114299074-114299074 on Build GRCh38
rs564771391 Chr.13: 114298987-114298987 on Build GRCh38
rs991010360 Chr.13: 114298990-114298990 on Build GRCh38
rs1044575045 Chr.13: 114299083-114299083 on Build GRCh38
rs539384185 Chr.13: 114299092-114299092 on Build GRCh38
rs960617867 Chr.13: 114298744-114298744 on Build GRCh38
rs967905548 Chr.13: 114299003-114299003 on Build GRCh38

View Full Product Details