Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 20430091-20430603 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KLHL33
Entrez Gene ID
123103
Gene Name
kelch like family member 33
Gene Aliases
-
UniGene
Hs.556014
Gene Chromosome Location
Chr.14: 20425852-20436509 on Build GRCh38
Transcripts
NM_001109997.2 XM_005267347.4 XM_011536450.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6001350 c.501G>A p.M167I Chr.14: 20430175-20430175 on Build GRCh38
3814427 c.302A>C p.H101P Chr.14: 20430374-20430374 on Build GRCh38
6001092 c.297C>T p.L99L Chr.14: 20430379-20430379 on Build GRCh38
3936454 c.220C>T p.R74C Chr.14: 20430456-20430456 on Build GRCh38
3936452 c.245C>T p.A82V Chr.14: 20430431-20430431 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs558348355 Chr.14: 20430168-20430168 on Build GRCh38
rs1031642800 Chr.14: 20430438-20430438 on Build GRCh38
rs1027802948 Chr.14: 20430214-20430214 on Build GRCh38
rs781672639 Chr.14: 20430392-20430392 on Build GRCh38
rs533271523 Chr.14: 20430348-20430348 on Build GRCh38
rs989593459 Chr.14: 20430265-20430265 on Build GRCh38
rs543538009 Chr.14: 20430189-20430189 on Build GRCh38
rs758014312 Chr.14: 20430246-20430246 on Build GRCh38
rs970718721 Chr.14: 20430139-20430139 on Build GRCh38
rs111564971 Chr.14: 20430232-20430232 on Build GRCh38
rs762184552 Chr.14: 20430407-20430407 on Build GRCh38
rs774326348 Chr.14: 20430159-20430159 on Build GRCh38
rs777568216 Chr.14: 20430305-20430305 on Build GRCh38
rs569861281 Chr.14: 20430431-20430431 on Build GRCh38
rs766652642 Chr.14: 20430315-20430316 on Build GRCh38
rs898147740 Chr.14: 20430456-20430456 on Build GRCh38
rs375147055 Chr.14: 20430499-20430499 on Build GRCh38
rs75780368 Chr.14: 20430388-20430388 on Build GRCh38
rs561745125 Chr.14: 20430193-20430193 on Build GRCh38
rs1025583210 Chr.14: 20430269-20430269 on Build GRCh38
rs747014779 Chr.14: 20430578-20430578 on Build GRCh38
rs775687666 Chr.14: 20430369-20430369 on Build GRCh38
rs780416931 Chr.14: 20430521-20430521 on Build GRCh38
rs1038121405 Chr.14: 20430430-20430430 on Build GRCh38
rs1050579751 Chr.14: 20430410-20430410 on Build GRCh38
rs749594652 Chr.14: 20430566-20430566 on Build GRCh38
rs530616192 Chr.14: 20430473-20430473 on Build GRCh38
rs778540589 Chr.14: 20430531-20430531 on Build GRCh38
rs981915275 Chr.14: 20430122-20430122 on Build GRCh38
rs752152668 Chr.14: 20430461-20430461 on Build GRCh38
rs970989153 Chr.14: 20430245-20430245 on Build GRCh38
rs549137826 Chr.14: 20430491-20430491 on Build GRCh38
rs771071543 Chr.14: 20430330-20430330 on Build GRCh38
rs956749932 Chr.14: 20430508-20430508 on Build GRCh38
rs755418436 Chr.14: 20430534-20430535 on Build GRCh38
rs752898059 Chr.14: 20430266-20430266 on Build GRCh38
rs1041981177 Chr.14: 20430319-20430319 on Build GRCh38
rs45448391 Chr.14: 20430426-20430426 on Build GRCh38
rs756025049 Chr.14: 20430301-20430301 on Build GRCh38
rs377615460 Chr.14: 20430391-20430391 on Build GRCh38
rs540170430 Chr.14: 20430299-20430299 on Build GRCh38
rs774691085 Chr.14: 20430342-20430342 on Build GRCh38
rs1023843497 Chr.14: 20430240-20430240 on Build GRCh38
rs536143952 Chr.14: 20430121-20430121 on Build GRCh38
rs777136277 Chr.14: 20430393-20430393 on Build GRCh38
rs866492022 Chr.14: 20430180-20430180 on Build GRCh38
rs756077656 Chr.14: 20430520-20430520 on Build GRCh38
rs551271666 Chr.14: 20430363-20430363 on Build GRCh38
rs372834006 Chr.14: 20430379-20430379 on Build GRCh38
rs962384556 Chr.14: 20430255-20430255 on Build GRCh38
rs1051047167 Chr.14: 20430196-20430196 on Build GRCh38
rs766135630 Chr.14: 20430231-20430231 on Build GRCh38
rs749043200 Chr.14: 20430329-20430329 on Build GRCh38
rs879024720 Chr.14: 20430455-20430455 on Build GRCh38
rs754558879 Chr.14: 20430474-20430474 on Build GRCh38
rs1050354065 Chr.14: 20430171-20430171 on Build GRCh38
rs994039969 Chr.14: 20430262-20430262 on Build GRCh38
rs567375766 Chr.14: 20430510-20430510 on Build GRCh38
rs78666331 Chr.14: 20430577-20430577 on Build GRCh38
rs202130260 Chr.14: 20430275-20430275 on Build GRCh38
rs1054264893 Chr.14: 20430153-20430153 on Build GRCh38
rs1023799496 Chr.14: 20430424-20430424 on Build GRCh38
rs914573987 Chr.14: 20430293-20430293 on Build GRCh38
rs541038404 Chr.14: 20430280-20430280 on Build GRCh38
rs867906895 Chr.14: 20430525-20430525 on Build GRCh38
rs529289338 Chr.14: 20430197-20430197 on Build GRCh38
rs12587478 Chr.14: 20430188-20430188 on Build GRCh38
rs923931753 Chr.14: 20430142-20430142 on Build GRCh38
rs933587861 Chr.14: 20430373-20430373 on Build GRCh38
rs11160645 Chr.14: 20430283-20430283 on Build GRCh38
rs990269443 Chr.14: 20430292-20430292 on Build GRCh38
rs17242648 Chr.14: 20430149-20430149 on Build GRCh38
rs750860123 Chr.14: 20430417-20430417 on Build GRCh38
rs550721927 Chr.14: 20430276-20430276 on Build GRCh38
rs1036669865 Chr.14: 20430304-20430304 on Build GRCh38
rs758196679 Chr.14: 20430158-20430158 on Build GRCh38
rs147268492 Chr.14: 20430141-20430141 on Build GRCh38
rs544792167 Chr.14: 20430420-20430420 on Build GRCh38
rs534550227 Chr.14: 20430567-20430567 on Build GRCh38
rs546054192 Chr.14: 20430150-20430150 on Build GRCh38
rs767481488 Chr.14: 20430179-20430179 on Build GRCh38
rs377055513 Chr.14: 20430254-20430254 on Build GRCh38
rs374640394 Chr.14: 20430377-20430377 on Build GRCh38
rs573523834 Chr.14: 20430528-20430528 on Build GRCh38
rs1003212014 Chr.14: 20430576-20430576 on Build GRCh38
rs763280914 Chr.14: 20430273-20430273 on Build GRCh38
rs746148586 Chr.14: 20430554-20430554 on Build GRCh38
rs753761938 Chr.14: 20430522-20430522 on Build GRCh38
rs748125642 Chr.14: 20430294-20430294 on Build GRCh38

View Full Product Details