Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 21017451-21017959 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NDRG2
Entrez Gene ID
57447
Gene Name
NDRG family member 2
Gene Aliases
SYLD
UniGene
Hs.525205
Gene Chromosome Location
Chr.14: 21016763-21070872 on Build GRCh38
Transcripts
NM_001282211.1 NM_001282212.1 NM_001282213.1 NM_001282214.1 NM_001282215.1 NM_001282216.1 NM_001320329.1 NM_016250.2 NM_201535.1 NM_201536.1 NM_201537.1 NM_201538.1 NM_201539.1 NM_201540.1 NM_201541.1 XM_011536996.2 XM_011536997.1 XM_011536998.1 XM_011536999.1 XM_011537001.1 XM_011537002.1 XM_017021480.1 XM_017021481.1 XM_017021482.1 XM_017021483.1 XM_017021484.1 XM_017021485.1 XM_017021486.1 XM_017021487.1 XM_017021488.1 XM_017021489.1 XM_017021490.1 XM_017021491.1 XM_017021492.1 XM_017021493.1 XM_017021494.1 XM_017021495.1 XM_017021496.1 XM_017021497.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
361760 c.1045C>A p.L349M Chr.14: 21017667-21017667 on Build GRCh38
4885264 c.1010C>A p.A337E Chr.14: 21017702-21017702 on Build GRCh38
954322 c.993C>T p.A331A Chr.14: 21017719-21017719 on Build GRCh38
4888483 c.979C>A p.R327R Chr.14: 21017733-21017733 on Build GRCh38
3814443 c.970C>T p.R324C Chr.14: 21017742-21017742 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1028802204 Chr.14: 21017501-21017501 on Build GRCh38
rs774021864 Chr.14: 21017752-21017752 on Build GRCh38
rs756174307 Chr.14: 21017689-21017689 on Build GRCh38
rs372921661 Chr.14: 21017811-21017811 on Build GRCh38
rs772406837 Chr.14: 21017482-21017482 on Build GRCh38
rs563421269 Chr.14: 21017797-21017797 on Build GRCh38
rs1002308778 Chr.14: 21017886-21017886 on Build GRCh38
rs547770040 Chr.14: 21017707-21017707 on Build GRCh38
rs750366803 Chr.14: 21017672-21017672 on Build GRCh38
rs796096401 Chr.14: 21017642-21017642 on Build GRCh38
rs537717120 Chr.14: 21017541-21017541 on Build GRCh38
rs370131705 Chr.14: 21017612-21017612 on Build GRCh38
rs748170347 Chr.14: 21017623-21017623 on Build GRCh38
rs1006495051 Chr.14: 21017534-21017534 on Build GRCh38
rs768193625 Chr.14: 21017717-21017717 on Build GRCh38
rs755190591 Chr.14: 21017604-21017604 on Build GRCh38
rs34978161 Chr.14: 21017719-21017719 on Build GRCh38
rs555445417 Chr.14: 21017673-21017673 on Build GRCh38
rs370030188 Chr.14: 21017772-21017772 on Build GRCh38
rs751702197 Chr.14: 21017580-21017580 on Build GRCh38
rs551514767 Chr.14: 21017494-21017494 on Build GRCh38
rs747835932 Chr.14: 21017936-21017936 on Build GRCh38
rs535431489 Chr.14: 21017632-21017632 on Build GRCh38
rs779406767 Chr.14: 21017785-21017785 on Build GRCh38
rs765329321 Chr.14: 21017656-21017656 on Build GRCh38
rs190675907 Chr.14: 21017697-21017697 on Build GRCh38
rs1243444 Chr.14: 21017895-21017895 on Build GRCh38
rs572120906 Chr.14: 21017678-21017678 on Build GRCh38
rs765204563 Chr.14: 21017556-21017556 on Build GRCh38
rs970652540 Chr.14: 21017885-21017885 on Build GRCh38
rs772708014 Chr.14: 21017719-21017719 on Build GRCh38
rs763824234 Chr.14: 21017884-21017884 on Build GRCh38
rs752893153 Chr.14: 21017766-21017766 on Build GRCh38
rs936422444 Chr.14: 21017628-21017628 on Build GRCh38
rs577782837 Chr.14: 21017753-21017753 on Build GRCh38
rs760531482 Chr.14: 21017558-21017558 on Build GRCh38
rs755169312 Chr.14: 21017684-21017684 on Build GRCh38
rs762825214 Chr.14: 21017739-21017739 on Build GRCh38
rs565923295 Chr.14: 21017820-21017820 on Build GRCh38
rs571760760 Chr.14: 21017511-21017511 on Build GRCh38
rs752842250 Chr.14: 21017685-21017685 on Build GRCh38
rs756225232 Chr.14: 21017770-21017770 on Build GRCh38
rs918570386 Chr.14: 21017917-21017917 on Build GRCh38
rs1037452785 Chr.14: 21017583-21017585 on Build GRCh38
rs980566113 Chr.14: 21017610-21017610 on Build GRCh38
rs114643241 Chr.14: 21017925-21017925 on Build GRCh38
rs969826966 Chr.14: 21017827-21017827 on Build GRCh38
rs750689405 Chr.14: 21017889-21017889 on Build GRCh38
rs375709473 Chr.14: 21017569-21017569 on Build GRCh38
rs374144090 Chr.14: 21017912-21017912 on Build GRCh38
rs753865924 Chr.14: 21017777-21017777 on Build GRCh38
rs766283302 Chr.14: 21017750-21017750 on Build GRCh38
rs142730462 Chr.14: 21017625-21017625 on Build GRCh38
rs374457004 Chr.14: 21017624-21017624 on Build GRCh38
rs979915084 Chr.14: 21017840-21017840 on Build GRCh38
rs746181403 Chr.14: 21017716-21017716 on Build GRCh38
rs760275223 Chr.14: 21017633-21017633 on Build GRCh38
rs767786777 Chr.14: 21017754-21017754 on Build GRCh38
rs371650957 Chr.14: 21017550-21017550 on Build GRCh38
rs761861297 Chr.14: 21017551-21017551 on Build GRCh38
rs772812565 Chr.14: 21017727-21017727 on Build GRCh38
rs376924416 Chr.14: 21017725-21017725 on Build GRCh38
rs539035605 Chr.14: 21017652-21017652 on Build GRCh38
rs150196221 Chr.14: 21017695-21017695 on Build GRCh38
rs993388736 Chr.14: 21017828-21017828 on Build GRCh38
rs746199044 Chr.14: 21017661-21017678 on Build GRCh38
rs528610037 Chr.14: 21017485-21017485 on Build GRCh38
rs1024825617 Chr.14: 21017878-21017878 on Build GRCh38
rs1016042060 Chr.14: 21017535-21017535 on Build GRCh38
rs764700871 Chr.14: 21017726-21017726 on Build GRCh38
rs368614805 Chr.14: 21017655-21017655 on Build GRCh38
rs757715719 Chr.14: 21017778-21017778 on Build GRCh38
rs970771640 Chr.14: 21017571-21017571 on Build GRCh38
rs182911950 Chr.14: 21017924-21017924 on Build GRCh38
rs772867279 Chr.14: 21017510-21017510 on Build GRCh38
rs768301542 Chr.14: 21017636-21017636 on Build GRCh38
rs200874762 Chr.14: 21017793-21017793 on Build GRCh38
rs753348243 Chr.14: 21017546-21017546 on Build GRCh38
rs771427840 Chr.14: 21017698-21017698 on Build GRCh38
rs781154658 Chr.14: 21017620-21017620 on Build GRCh38

View Full Product Details