Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16027743-16028251 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCNKA
Entrez Gene ID
1187
Gene Name
chloride voltage-gated channel Ka
Gene Aliases
CLCK1, ClC-K1, hClC-Ka
UniGene
Hs.591533
Gene Chromosome Location
Chr.1: 16021991-16034050 on Build GRCh38
Transcripts
NM_001042704.1 NM_001257139.1 NM_004070.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5895466 c.867-4G>A p.? Chr.1: 16028014-16028014 on Build GRCh38
6305406 c.867-3C>A p.? Chr.1: 16028015-16028015 on Build GRCh38
5052767 c.942G>A p.R314R Chr.1: 16028093-16028093 on Build GRCh38
898841 c.846C>A p.I282I Chr.1: 16027885-16027885 on Build GRCh38
6305529 c.867-4G>C p.? Chr.1: 16028014-16028014 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs563048986 Chr.1: 16027931-16027931 on Build GRCh38
rs774126212 Chr.1: 16027862-16027862 on Build GRCh38
rs371179594 Chr.1: 16028027-16028027 on Build GRCh38
rs760519796 Chr.1: 16027968-16027968 on Build GRCh38
rs774234204 Chr.1: 16027969-16027969 on Build GRCh38
rs745876845 Chr.1: 16028128-16028129 on Build GRCh38
rs761058203 Chr.1: 16028079-16028079 on Build GRCh38
rs1016078455 Chr.1: 16028227-16028227 on Build GRCh38
rs143337074 Chr.1: 16027870-16027870 on Build GRCh38
rs781193194 Chr.1: 16028108-16028108 on Build GRCh38
rs1043494133 Chr.1: 16027784-16027784 on Build GRCh38
rs770812787 Chr.1: 16028034-16028034 on Build GRCh38
rs769372786 Chr.1: 16028111-16028111 on Build GRCh38
rs766854851 Chr.1: 16028157-16028157 on Build GRCh38
rs769867328 Chr.1: 16027852-16027852 on Build GRCh38
rs757652917 Chr.1: 16027914-16027914 on Build GRCh38
rs200145823 Chr.1: 16027830-16027830 on Build GRCh38
rs551638915 Chr.1: 16027900-16027900 on Build GRCh38
rs533672725 Chr.1: 16027967-16027967 on Build GRCh38
rs192722050 Chr.1: 16027948-16027948 on Build GRCh38
rs960055054 Chr.1: 16027834-16027834 on Build GRCh38
rs759910166 Chr.1: 16028169-16028169 on Build GRCh38
rs776429454 Chr.1: 16028038-16028038 on Build GRCh38
rs773659269 Chr.1: 16028066-16028066 on Build GRCh38
rs745654489 Chr.1: 16028109-16028109 on Build GRCh38
rs748241713 Chr.1: 16028028-16028028 on Build GRCh38
rs762809631 Chr.1: 16027777-16027777 on Build GRCh38
rs377492878 Chr.1: 16027982-16027982 on Build GRCh38
rs1023937139 Chr.1: 16028159-16028159 on Build GRCh38
rs201275966 Chr.1: 16027874-16027874 on Build GRCh38
rs757401433 Chr.1: 16027946-16027947 on Build GRCh38
rs200268763 Chr.1: 16027899-16027899 on Build GRCh38
rs748769879 Chr.1: 16028115-16028115 on Build GRCh38
rs865822656 Chr.1: 16028167-16028167 on Build GRCh38
rs768521208 Chr.1: 16028162-16028162 on Build GRCh38
rs768064323 Chr.1: 16028000-16028000 on Build GRCh38
rs761109638 Chr.1: 16028137-16028137 on Build GRCh38
rs759150905 Chr.1: 16027974-16027974 on Build GRCh38
rs138110172 Chr.1: 16028086-16028086 on Build GRCh38
rs781343628 Chr.1: 16027917-16027917 on Build GRCh38
rs377753454 Chr.1: 16027947-16027947 on Build GRCh38
rs746879594 Chr.1: 16028105-16028105 on Build GRCh38
rs200545230 Chr.1: 16027787-16027787 on Build GRCh38
rs12126269 Chr.1: 16028095-16028095 on Build GRCh38
rs201927569 Chr.1: 16028030-16028030 on Build GRCh38
rs745462501 Chr.1: 16028047-16028047 on Build GRCh38
rs1013433573 Chr.1: 16027839-16027839 on Build GRCh38
rs1043222032 Chr.1: 16028078-16028078 on Build GRCh38
rs753052635 Chr.1: 16027888-16027889 on Build GRCh38
rs769813998 Chr.1: 16027776-16027776 on Build GRCh38
rs758691642 Chr.1: 16027929-16027929 on Build GRCh38
rs768672657 Chr.1: 16027942-16027942 on Build GRCh38
rs571199355 Chr.1: 16027966-16027966 on Build GRCh38
rs755086597 Chr.1: 16028026-16028026 on Build GRCh38
rs759083193 Chr.1: 16027903-16027903 on Build GRCh38
rs184146945 Chr.1: 16027951-16027951 on Build GRCh38
rs768580283 Chr.1: 16027884-16027886 on Build GRCh38
rs536038955 Chr.1: 16028048-16028048 on Build GRCh38
rs771717536 Chr.1: 16027871-16027871 on Build GRCh38
rs754018221 Chr.1: 16028020-16028020 on Build GRCh38
rs371622512 Chr.1: 16027831-16027831 on Build GRCh38
rs756386646 Chr.1: 16027924-16027924 on Build GRCh38
rs776377990 Chr.1: 16027971-16027971 on Build GRCh38
rs754069601 Chr.1: 16028089-16028089 on Build GRCh38
rs749417866 Chr.1: 16027940-16027940 on Build GRCh38
rs760341738 Chr.1: 16027893-16027893 on Build GRCh38
rs78557747 Chr.1: 16028147-16028148 on Build GRCh38
rs781497766 Chr.1: 16027835-16027835 on Build GRCh38
rs775612930 Chr.1: 16027854-16027854 on Build GRCh38
rs539030239 Chr.1: 16028096-16028096 on Build GRCh38
rs201670619 Chr.1: 16027970-16027970 on Build GRCh38
rs773710484 Chr.1: 16028119-16028119 on Build GRCh38
rs764309129 Chr.1: 16027938-16027939 on Build GRCh38
rs756003544 Chr.1: 16028136-16028137 on Build GRCh38
rs759935969 Chr.1: 16027877-16027877 on Build GRCh38
rs574603424 Chr.1: 16028158-16028158 on Build GRCh38
rs768170598 Chr.1: 16028117-16028117 on Build GRCh38
rs750794982 Chr.1: 16027920-16027920 on Build GRCh38
rs200325856 Chr.1: 16028010-16028010 on Build GRCh38
rs139738448 Chr.1: 16028061-16028061 on Build GRCh38
rs371274486 Chr.1: 16028092-16028092 on Build GRCh38
rs10927889 Chr.1: 16027785-16027785 on Build GRCh38
rs376669628 Chr.1: 16028091-16028091 on Build GRCh38
rs777233269 Chr.1: 16027788-16027789 on Build GRCh38
rs780276412 Chr.1: 16028014-16028014 on Build GRCh38
rs764014110 Chr.1: 16027780-16027780 on Build GRCh38
rs762285802 Chr.1: 16027999-16027999 on Build GRCh38
rs757133087 Chr.1: 16028106-16028106 on Build GRCh38
rs746018490 Chr.1: 16027848-16027848 on Build GRCh38
rs777748952 Chr.1: 16028031-16028031 on Build GRCh38
rs201587330 Chr.1: 16028003-16028003 on Build GRCh38
rs777043611 Chr.1: 16028160-16028160 on Build GRCh38
rs764586512 Chr.1: 16027980-16027980 on Build GRCh38
rs781111805 Chr.1: 16028077-16028079 on Build GRCh38
rs755711209 Chr.1: 16027800-16027800 on Build GRCh38
rs398089145 Chr.1: 16028148-16028149 on Build GRCh38
rs567160606 Chr.1: 16028196-16028196 on Build GRCh38
rs564498247 Chr.1: 16027803-16027803 on Build GRCh38
rs368824021 Chr.1: 16027859-16027859 on Build GRCh38
rs745988725 Chr.1: 16027775-16027775 on Build GRCh38
rs764690903 Chr.1: 16027910-16027910 on Build GRCh38
rs753265556 Chr.1: 16027808-16027808 on Build GRCh38
rs386628867 Chr.1: 16027785-16027786 on Build GRCh38
rs12753541 Chr.1: 16028177-16028177 on Build GRCh38
rs761492507 Chr.1: 16027782-16027782 on Build GRCh38
rs57016480 Chr.1: 16027786-16027786 on Build GRCh38

View Full Product Details