Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40776105-40776597 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC17
Entrez Gene ID
55192
Gene Name
DnaJ heat shock protein family (Hsp40) member C17
Gene Aliases
-
UniGene
Hs.511069
Gene Chromosome Location
Chr.15: 40767448-40807478 on Build GRCh38
Transcripts
NM_018163.2 XM_017022401.1 XM_017022402.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5940508 c.373G>A p.E125K Chr.15: 40776550-40776550 on Build GRCh38
5021828 c.350A>C p.E117A Chr.15: 40776573-40776573 on Build GRCh38
3936735 c.334A>C p.S112R Chr.15: 40776589-40776589 on Build GRCh38
6204386 c.456G>A p.Q152Q Chr.15: 40776218-40776218 on Build GRCh38
961303 c.419A>G p.E140G Chr.15: 40776255-40776255 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749766305 Chr.15: 40776299-40776299 on Build GRCh38
rs758894674 Chr.15: 40776449-40776449 on Build GRCh38
rs529115513 Chr.15: 40776255-40776255 on Build GRCh38
rs867703010 Chr.15: 40776361-40776361 on Build GRCh38
rs749785446 Chr.15: 40776222-40776222 on Build GRCh38
rs770325330 Chr.15: 40776522-40776522 on Build GRCh38
rs373859480 Chr.15: 40776547-40776547 on Build GRCh38
rs773652605 Chr.15: 40776161-40776161 on Build GRCh38
rs760831818 Chr.15: 40776558-40776558 on Build GRCh38
rs779888515 Chr.15: 40776203-40776203 on Build GRCh38
rs770236915 Chr.15: 40776157-40776157 on Build GRCh38
rs779170943 Chr.15: 40776304-40776304 on Build GRCh38
rs563911364 Chr.15: 40776138-40776138 on Build GRCh38
rs753907400 Chr.15: 40776385-40776385 on Build GRCh38
rs769830132 Chr.15: 40776216-40776216 on Build GRCh38
rs200488800 Chr.15: 40776163-40776163 on Build GRCh38
rs764097170 Chr.15: 40776371-40776371 on Build GRCh38
rs541049943 Chr.15: 40776124-40776124 on Build GRCh38
rs915335564 Chr.15: 40776462-40776462 on Build GRCh38
rs149784969 Chr.15: 40776566-40776566 on Build GRCh38
rs747790802 Chr.15: 40776312-40776312 on Build GRCh38
rs913058221 Chr.15: 40776416-40776416 on Build GRCh38
rs752982849 Chr.15: 40776365-40776365 on Build GRCh38
rs772718887 Chr.15: 40776321-40776321 on Build GRCh38
rs771415170 Chr.15: 40776303-40776303 on Build GRCh38
rs763705700 Chr.15: 40776175-40776175 on Build GRCh38
rs139728903 Chr.15: 40776564-40776564 on Build GRCh38
rs778189158 Chr.15: 40776221-40776221 on Build GRCh38
rs369765784 Chr.15: 40776177-40776177 on Build GRCh38
rs1018571765 Chr.15: 40776130-40776130 on Build GRCh38
rs772625761 Chr.15: 40776505-40776505 on Build GRCh38
rs768789272 Chr.15: 40776146-40776146 on Build GRCh38
rs371807622 Chr.15: 40776166-40776166 on Build GRCh38
rs916555526 Chr.15: 40776225-40776225 on Build GRCh38
rs748865164 Chr.15: 40776511-40776511 on Build GRCh38
rs767302364 Chr.15: 40776544-40776544 on Build GRCh38
rs759375950 Chr.15: 40776343-40776343 on Build GRCh38
rs761869231 Chr.15: 40776178-40776178 on Build GRCh38
rs765221994 Chr.15: 40776567-40776567 on Build GRCh38
rs377498510 Chr.15: 40776563-40776563 on Build GRCh38
rs767285910 Chr.15: 40776345-40776345 on Build GRCh38
rs771129945 Chr.15: 40776445-40776445 on Build GRCh38
rs757736399 Chr.15: 40776426-40776426 on Build GRCh38
rs756260105 Chr.15: 40776370-40776370 on Build GRCh38
rs372426872 Chr.15: 40776173-40776173 on Build GRCh38
rs758178010 Chr.15: 40776188-40776188 on Build GRCh38
rs878857611 Chr.15: 40776131-40776131 on Build GRCh38
rs369297714 Chr.15: 40776305-40776305 on Build GRCh38
rs775061086 Chr.15: 40776167-40776167 on Build GRCh38
rs758094191 Chr.15: 40776408-40776408 on Build GRCh38
rs534789773 Chr.15: 40776286-40776286 on Build GRCh38
rs774237003 Chr.15: 40776342-40776342 on Build GRCh38
rs751041536 Chr.15: 40776568-40776568 on Build GRCh38
rs762879970 Chr.15: 40776265-40776265 on Build GRCh38
rs900151025 Chr.15: 40776466-40776466 on Build GRCh38
rs776311903 Chr.15: 40776235-40776235 on Build GRCh38
rs549170424 Chr.15: 40776264-40776264 on Build GRCh38
rs766100217 Chr.15: 40776275-40776275 on Build GRCh38
rs772534444 Chr.15: 40776306-40776306 on Build GRCh38
rs1015071618 Chr.15: 40776515-40776515 on Build GRCh38
rs747813426 Chr.15: 40776495-40776495 on Build GRCh38
rs762817372 Chr.15: 40776324-40776324 on Build GRCh38
rs1004639350 Chr.15: 40776506-40776506 on Build GRCh38
rs989941646 Chr.15: 40776452-40776452 on Build GRCh38
rs373911227 Chr.15: 40776309-40776309 on Build GRCh38
rs868546616 Chr.15: 40776474-40776474 on Build GRCh38
rs79709714 Chr.15: 40776573-40776573 on Build GRCh38
rs960951302 Chr.15: 40776215-40776215 on Build GRCh38
rs748667057 Chr.15: 40776155-40776155 on Build GRCh38
rs746358632 Chr.15: 40776437-40776437 on Build GRCh38
rs73398528 Chr.15: 40776211-40776211 on Build GRCh38
rs759464104 Chr.15: 40776543-40776543 on Build GRCh38
rs779518441 Chr.15: 40776432-40776432 on Build GRCh38
rs113054374 Chr.15: 40776344-40776344 on Build GRCh38
rs906488927 Chr.15: 40776151-40776151 on Build GRCh38
rs752890995 Chr.15: 40776289-40776289 on Build GRCh38
rs774713452 Chr.15: 40776223-40776223 on Build GRCh38
rs146094788 Chr.15: 40776290-40776290 on Build GRCh38
rs751732790 Chr.15: 40776204-40776204 on Build GRCh38
rs769256544 Chr.15: 40776313-40776313 on Build GRCh38
rs774329211 Chr.15: 40776524-40776524 on Build GRCh38
rs954254702 Chr.15: 40776451-40776451 on Build GRCh38
rs751318823 Chr.15: 40776279-40776279 on Build GRCh38
rs369698216 Chr.15: 40776491-40776491 on Build GRCh38
rs769435879 Chr.15: 40776499-40776499 on Build GRCh38
rs761449037 Chr.15: 40776243-40776243 on Build GRCh38
rs186135784 Chr.15: 40776285-40776285 on Build GRCh38
rs563534178 Chr.15: 40776213-40776213 on Build GRCh38
rs748186644 Chr.15: 40776214-40776214 on Build GRCh38
rs776624067 Chr.15: 40776147-40776147 on Build GRCh38
rs766295841 Chr.15: 40776333-40776333 on Build GRCh38
rs111610422 Chr.15: 40776406-40776406 on Build GRCh38
rs571550532 Chr.15: 40776465-40776465 on Build GRCh38
rs768378470 Chr.15: 40776234-40776234 on Build GRCh38
rs769146353 Chr.15: 40776469-40776469 on Build GRCh38

View Full Product Details