Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 41775089-41775599 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAPKBP1
Entrez Gene ID
23005
Gene Name
mitogen-activated protein kinase binding protein 1
Gene Aliases
JNKBP-1, JNKBP1
UniGene
Hs.513661
Gene Chromosome Location
Chr.15: 41774434-41827855 on Build GRCh38
Transcripts
NM_001128608.1 NM_001265611.1 NM_014994.2 NR_049761.1 NR_049762.1 XM_006720438.1 XM_011521383.1 XM_011521384.2 XM_017022017.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7173699 c.33G>A p.R11R Chr.15: 41775308-41775308 on Build GRCh38
6329323 c.45G>T p.L15L Chr.15: 41775320-41775320 on Build GRCh38
4838535 c.92G>A p.R31Q Chr.15: 41775367-41775367 on Build GRCh38
3747974 c.16T>G p.S6A Chr.15: 41775291-41775291 on Build GRCh38
6318852 c.14G>T p.G5V Chr.15: 41775289-41775289 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs577668822 Chr.15: 41775116-41775116 on Build GRCh38
rs775250612 Chr.15: 41775286-41775286 on Build GRCh38
rs776443247 Chr.15: 41775298-41775298 on Build GRCh38
rs192394045 Chr.15: 41775245-41775245 on Build GRCh38
rs974531994 Chr.15: 41775335-41775335 on Build GRCh38
rs777922669 Chr.15: 41775279-41775279 on Build GRCh38
rs937038124 Chr.15: 41775491-41775491 on Build GRCh38
rs766195217 Chr.15: 41775344-41775344 on Build GRCh38
rs746757530 Chr.15: 41775426-41775426 on Build GRCh38
rs559673133 Chr.15: 41775262-41775262 on Build GRCh38
rs759020925 Chr.15: 41775510-41775510 on Build GRCh38
rs746844515 Chr.15: 41775291-41775291 on Build GRCh38
rs549304918 Chr.15: 41775552-41775552 on Build GRCh38
rs1046896027 Chr.15: 41775175-41775175 on Build GRCh38
rs772893935 Chr.15: 41775307-41775307 on Build GRCh38
rs968939954 Chr.15: 41775169-41775169 on Build GRCh38
rs755499274 Chr.15: 41775254-41775254 on Build GRCh38
rs535365240 Chr.15: 41775522-41775522 on Build GRCh38
rs550583561 Chr.15: 41775490-41775490 on Build GRCh38
rs750119183 Chr.15: 41775227-41775227 on Build GRCh38
rs1013059300 Chr.15: 41775121-41775121 on Build GRCh38
rs753166198 Chr.15: 41775392-41775392 on Build GRCh38
rs376446996 Chr.15: 41775473-41775473 on Build GRCh38
rs762745701 Chr.15: 41775346-41775346 on Build GRCh38
rs563941431 Chr.15: 41775135-41775135 on Build GRCh38
rs766921868 Chr.15: 41775543-41775543 on Build GRCh38
rs148633878 Chr.15: 41775297-41775297 on Build GRCh38
rs1023241380 Chr.15: 41775160-41775160 on Build GRCh38
rs762676287 Chr.15: 41775332-41775332 on Build GRCh38
rs956984232 Chr.15: 41775290-41775290 on Build GRCh38
rs557925691 Chr.15: 41775112-41775112 on Build GRCh38
rs779108698 Chr.15: 41775420-41775420 on Build GRCh38
rs528732214 Chr.15: 41775399-41775399 on Build GRCh38
rs747509179 Chr.15: 41775183-41775183 on Build GRCh38
rs1044569739 Chr.15: 41775468-41775468 on Build GRCh38
rs937009281 Chr.15: 41775115-41775115 on Build GRCh38
rs771191464 Chr.15: 41775283-41775283 on Build GRCh38
rs200914973 Chr.15: 41775255-41775255 on Build GRCh38
rs768011316 Chr.15: 41775380-41775380 on Build GRCh38
rs758800211 Chr.15: 41775236-41775236 on Build GRCh38
rs768354875 Chr.15: 41775439-41775439 on Build GRCh38
rs984565258 Chr.15: 41775352-41775352 on Build GRCh38
rs988757291 Chr.15: 41775295-41775295 on Build GRCh38
rs751971556 Chr.15: 41775345-41775345 on Build GRCh38
rs756163326 Chr.15: 41775273-41775273 on Build GRCh38
rs773766391 Chr.15: 41775485-41775485 on Build GRCh38
rs530768576 Chr.15: 41775488-41775488 on Build GRCh38
rs565048943 Chr.15: 41775417-41775417 on Build GRCh38
rs371094728 Chr.15: 41775359-41775359 on Build GRCh38
rs999892071 Chr.15: 41775212-41775212 on Build GRCh38
rs543443078 Chr.15: 41775120-41775120 on Build GRCh38
rs761334748 Chr.15: 41775303-41775303 on Build GRCh38
rs149158040 Chr.15: 41775521-41775521 on Build GRCh38
rs372913874 Chr.15: 41775438-41775438 on Build GRCh38
rs948889335 Chr.15: 41775449-41775449 on Build GRCh38
rs895878709 Chr.15: 41775428-41775428 on Build GRCh38
rs756684650 Chr.15: 41775396-41775396 on Build GRCh38
rs1057228361 Chr.15: 41775409-41775409 on Build GRCh38
rs373886335 Chr.15: 41775350-41775350 on Build GRCh38
rs929929196 Chr.15: 41775571-41775571 on Build GRCh38
rs764867296 Chr.15: 41775305-41775305 on Build GRCh38
rs888259377 Chr.15: 41775188-41775188 on Build GRCh38
rs76601191 Chr.15: 41775406-41775406 on Build GRCh38
rs781140548 Chr.15: 41775440-41775440 on Build GRCh38
rs200766282 Chr.15: 41775281-41775281 on Build GRCh38
rs1018361031 Chr.15: 41775246-41775246 on Build GRCh38
rs895725356 Chr.15: 41775147-41775147 on Build GRCh38
rs920542581 Chr.15: 41775338-41775338 on Build GRCh38
rs752022715 Chr.15: 41775248-41775248 on Build GRCh38
rs964534101 Chr.15: 41775320-41775320 on Build GRCh38
rs766081628 Chr.15: 41775472-41775472 on Build GRCh38
rs987247011 Chr.15: 41775234-41775234 on Build GRCh38
rs143298817 Chr.15: 41775555-41775555 on Build GRCh38
rs1031332938 Chr.15: 41775216-41775216 on Build GRCh38

View Full Product Details