Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 78565428-78565934 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNA5
Entrez Gene ID
1138
Gene Name
cholinergic receptor nicotinic alpha 5 subunit
Gene Aliases
LNCR2
UniGene
Hs.1614
Gene Chromosome Location
Chr.15: 78565520-78595269 on Build GRCh38
Transcripts
NM_000745.3 NM_001307945.1 XM_005254142.3 XM_017021881.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
226764 c.34C>T p.R12C Chr.15: 78565753-78565753 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1055332693 Chr.15: 78565610-78565613 on Build GRCh38
rs911675549 Chr.15: 78565865-78565865 on Build GRCh38
rs55853698 Chr.15: 78565597-78565597 on Build GRCh38
rs973502683 Chr.15: 78565542-78565542 on Build GRCh38
rs550828280 Chr.15: 78565469-78565469 on Build GRCh38
rs77293642 Chr.15: 78565571-78565571 on Build GRCh38
rs912729299 Chr.15: 78565458-78565458 on Build GRCh38
rs1051625981 Chr.15: 78565462-78565462 on Build GRCh38
rs982494622 Chr.15: 78565807-78565807 on Build GRCh38
rs904476911 Chr.15: 78565568-78565568 on Build GRCh38
rs906355297 Chr.15: 78565530-78565530 on Build GRCh38
rs1020229973 Chr.15: 78565567-78565567 on Build GRCh38
rs1031751646 Chr.15: 78565520-78565520 on Build GRCh38
rs200455274 Chr.15: 78565588-78565588 on Build GRCh38
rs200645739 Chr.15: 78565642-78565642 on Build GRCh38
rs1042266637 Chr.15: 78565909-78565909 on Build GRCh38
rs1000637637 Chr.15: 78565515-78565515 on Build GRCh38
rs904486486 Chr.15: 78565508-78565508 on Build GRCh38
rs1041343164 Chr.15: 78565827-78565827 on Build GRCh38
rs987185198 Chr.15: 78565848-78565848 on Build GRCh38
rs769810134 Chr.15: 78565463-78565463 on Build GRCh38
rs1024406847 Chr.15: 78565527-78565527 on Build GRCh38
rs984360120 Chr.15: 78565746-78565746 on Build GRCh38
rs891814444 Chr.15: 78565576-78565576 on Build GRCh38
rs1015628724 Chr.15: 78565765-78565765 on Build GRCh38
rs773868574 Chr.15: 78565613-78565613 on Build GRCh38
rs1042072107 Chr.15: 78565651-78565651 on Build GRCh38
rs55781567 Chr.15: 78565644-78565644 on Build GRCh38
rs67624739 Chr.15: 78565480-78565501 on Build GRCh38
rs1018715411 Chr.15: 78565689-78565689 on Build GRCh38
rs555177593 Chr.15: 78565812-78565812 on Build GRCh38
rs966047843 Chr.15: 78565459-78565459 on Build GRCh38
rs928078724 Chr.15: 78565811-78565811 on Build GRCh38
rs199898580 Chr.15: 78565522-78565522 on Build GRCh38
rs56182392 Chr.15: 78565537-78565537 on Build GRCh38
rs943179051 Chr.15: 78565871-78565871 on Build GRCh38
rs943749647 Chr.15: 78565735-78565735 on Build GRCh38
rs902311741 Chr.15: 78565565-78565565 on Build GRCh38
rs990709175 Chr.15: 78565828-78565828 on Build GRCh38
rs534031880 Chr.15: 78565773-78565773 on Build GRCh38
rs893727247 Chr.15: 78565630-78565630 on Build GRCh38
rs1024354288 Chr.15: 78565595-78565595 on Build GRCh38
rs1038907690 Chr.15: 78565886-78565886 on Build GRCh38
rs963075141 Chr.15: 78565541-78565541 on Build GRCh38
rs950415191 Chr.15: 78565681-78565681 on Build GRCh38
rs868150762 Chr.15: 78565593-78565593 on Build GRCh38
rs1051527719 Chr.15: 78565551-78565551 on Build GRCh38
rs980289571 Chr.15: 78565536-78565536 on Build GRCh38
rs902173838 Chr.15: 78565660-78565660 on Build GRCh38
rs929018427 Chr.15: 78565546-78565546 on Build GRCh38
rs112992268 Chr.15: 78565708-78565708 on Build GRCh38
rs918999716 Chr.15: 78565545-78565545 on Build GRCh38
rs867794962 Chr.15: 78565649-78565649 on Build GRCh38
rs992887197 Chr.15: 78565526-78565526 on Build GRCh38
rs201545082 Chr.15: 78565601-78565601 on Build GRCh38
rs503464 Chr.15: 78565554-78565554 on Build GRCh38
rs970205240 Chr.15: 78565531-78565531 on Build GRCh38
rs199669075 Chr.15: 78565638-78565638 on Build GRCh38
rs915069927 Chr.15: 78565884-78565884 on Build GRCh38
rs201230605 Chr.15: 78565523-78565523 on Build GRCh38
rs1038959732 Chr.15: 78565564-78565564 on Build GRCh38
rs963320251 Chr.15: 78565639-78565639 on Build GRCh38
rs997031085 Chr.15: 78565736-78565736 on Build GRCh38
rs1017168838 Chr.15: 78565612-78565612 on Build GRCh38
rs549182147 Chr.15: 78565880-78565880 on Build GRCh38
rs931318437 Chr.15: 78565500-78565500 on Build GRCh38
rs1017533890 Chr.15: 78565485-78565485 on Build GRCh38
rs142774214 Chr.15: 78565471-78565492 on Build GRCh38
rs925824155 Chr.15: 78565911-78565911 on Build GRCh38
rs12903524 Chr.15: 78565889-78565889 on Build GRCh38
rs1053501946 Chr.15: 78565574-78565574 on Build GRCh38
rs952978648 Chr.15: 78565745-78565745 on Build GRCh38
rs202210732 Chr.15: 78565533-78565533 on Build GRCh38
rs1014256261 Chr.15: 78565594-78565594 on Build GRCh38
rs375267067 Chr.15: 78565480-78565480 on Build GRCh38
rs961390206 Chr.15: 78565806-78565806 on Build GRCh38
rs1028454004 Chr.15: 78565743-78565743 on Build GRCh38
rs900218915 Chr.15: 78565718-78565718 on Build GRCh38
rs201669774 Chr.15: 78565590-78565590 on Build GRCh38
rs200696239 Chr.15: 78565600-78565600 on Build GRCh38
rs943188936 Chr.15: 78565556-78565556 on Build GRCh38
rs918892716 Chr.15: 78565671-78565671 on Build GRCh38

View Full Product Details