Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 78592919-78593419 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHRNA3
Entrez Gene ID
1136
Gene Name
cholinergic receptor nicotinic alpha 3 subunit
Gene Aliases
LNCR2, NACHRA3, PAOD2
UniGene
Hs.89605
Gene Chromosome Location
Chr.15: 78593052-78621295 on Build GRCh38
Transcripts
NM_001166694.1 NR_046313.1 NM_000745.3 NM_001307945.1 XM_005254142.3 XM_017021881.1

Gene Symbol
CHRNA5
Entrez Gene ID
1138
Gene Name
cholinergic receptor nicotinic alpha 5 subunit
Gene Aliases
LNCR2
UniGene
Hs.1614
Gene Chromosome Location
Chr.15: 78565520-78595269 on Build GRCh38
Transcripts
NM_001166694.1 NR_046313.1 NM_000745.3 NM_001307945.1 XM_005254142.3 XM_017021881.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1135727 c.1303T>C p.F435L Chr.15: 78593149-78593149 on Build GRCh38
6258205 c.1342G>T p.V448F Chr.15: 78593188-78593188 on Build GRCh38
2011172 c.1391G>A p.G464E Chr.15: 78593237-78593237 on Build GRCh38
2011171 c.1285C>T p.R429W Chr.15: 78593131-78593131 on Build GRCh38
5433056 c.1287G>C p.R429R Chr.15: 78593133-78593133 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757656190 Chr.15: 78593076-78593076 on Build GRCh38
rs889893693 Chr.15: 78592960-78592960 on Build GRCh38
rs746886634 Chr.15: 78593203-78593203 on Build GRCh38
rs202044161 Chr.15: 78593252-78593252 on Build GRCh38
rs766695728 Chr.15: 78593236-78593236 on Build GRCh38
rs913460296 Chr.15: 78593094-78593094 on Build GRCh38
rs199796617 Chr.15: 78593259-78593259 on Build GRCh38
rs113619302 Chr.15: 78592961-78592961 on Build GRCh38
rs758933260 Chr.15: 78593049-78593049 on Build GRCh38
rs77773727 Chr.15: 78593171-78593171 on Build GRCh38
rs976095415 Chr.15: 78593291-78593291 on Build GRCh38
rs751943214 Chr.15: 78593070-78593070 on Build GRCh38
rs561791283 Chr.15: 78593004-78593004 on Build GRCh38
rs770782949 Chr.15: 78593224-78593224 on Build GRCh38
rs199932554 Chr.15: 78593382-78593382 on Build GRCh38
rs926158488 Chr.15: 78593051-78593051 on Build GRCh38
rs759148420 Chr.15: 78593297-78593297 on Build GRCh38
rs1025964748 Chr.15: 78592985-78592985 on Build GRCh38
rs763265856 Chr.15: 78593194-78593197 on Build GRCh38
rs751038368 Chr.15: 78593237-78593237 on Build GRCh38
rs763412847 Chr.15: 78593156-78593156 on Build GRCh38
rs775537854 Chr.15: 78593251-78593251 on Build GRCh38
rs1040446529 Chr.15: 78593011-78593011 on Build GRCh38
rs952025523 Chr.15: 78592965-78592965 on Build GRCh38
rs757859948 Chr.15: 78593189-78593189 on Build GRCh38
rs778479885 Chr.15: 78593103-78593103 on Build GRCh38
rs536798652 Chr.15: 78593000-78593000 on Build GRCh38
rs754450688 Chr.15: 78593102-78593102 on Build GRCh38
rs540559200 Chr.15: 78592956-78592956 on Build GRCh38
rs202136309 Chr.15: 78593157-78593157 on Build GRCh38
rs150329151 Chr.15: 78593122-78593122 on Build GRCh38
rs3830438 Chr.15: 78593277-78593278 on Build GRCh38
rs201714288 Chr.15: 78593327-78593327 on Build GRCh38
rs368612437 Chr.15: 78593132-78593132 on Build GRCh38
rs113264952 Chr.15: 78593388-78593388 on Build GRCh38
rs201184771 Chr.15: 78593350-78593350 on Build GRCh38
rs199559512 Chr.15: 78593172-78593172 on Build GRCh38
rs777707103 Chr.15: 78593199-78593199 on Build GRCh38
rs959165006 Chr.15: 78593198-78593198 on Build GRCh38
rs367546696 Chr.15: 78593158-78593158 on Build GRCh38
rs754643899 Chr.15: 78593165-78593165 on Build GRCh38
rs199881121 Chr.15: 78593335-78593335 on Build GRCh38
rs770027654 Chr.15: 78593243-78593243 on Build GRCh38
rs934230114 Chr.15: 78593107-78593107 on Build GRCh38
rs202175642 Chr.15: 78593385-78593385 on Build GRCh38
rs372562923 Chr.15: 78593001-78593001 on Build GRCh38
rs748171498 Chr.15: 78593086-78593086 on Build GRCh38
rs960433490 Chr.15: 78592989-78592989 on Build GRCh38
rs972714913 Chr.15: 78592980-78592980 on Build GRCh38
rs762405020 Chr.15: 78593277-78593277 on Build GRCh38
rs202052590 Chr.15: 78593120-78593120 on Build GRCh38
rs983433002 Chr.15: 78592971-78592971 on Build GRCh38
rs920478161 Chr.15: 78592943-78592943 on Build GRCh38
rs148376112 Chr.15: 78593170-78593170 on Build GRCh38
rs779875936 Chr.15: 78593084-78593084 on Build GRCh38
rs1053394384 Chr.15: 78593067-78593067 on Build GRCh38
rs772051214 Chr.15: 78593074-78593076 on Build GRCh38
rs201177341 Chr.15: 78593229-78593229 on Build GRCh38
rs144352852 Chr.15: 78593151-78593151 on Build GRCh38
rs72534078 Chr.15: 78593276-78593277 on Build GRCh38
rs746129356 Chr.15: 78593234-78593234 on Build GRCh38
rs764603712 Chr.15: 78593058-78593058 on Build GRCh38
rs76071148 Chr.15: 78593232-78593232 on Build GRCh38
rs144182521 Chr.15: 78593273-78593274 on Build GRCh38
rs67529014 Chr.15: 78593027-78593027 on Build GRCh38
rs200921958 Chr.15: 78593383-78593383 on Build GRCh38
rs770658199 Chr.15: 78593136-78593136 on Build GRCh38
rs202066018 Chr.15: 78593153-78593153 on Build GRCh38
rs532896027 Chr.15: 78593183-78593183 on Build GRCh38
rs965993931 Chr.15: 78593278-78593278 on Build GRCh38
rs200919591 Chr.15: 78593273-78593273 on Build GRCh38
rs936162811 Chr.15: 78593066-78593066 on Build GRCh38
rs768051249 Chr.15: 78593284-78593284 on Build GRCh38
rs200384194 Chr.15: 78593174-78593174 on Build GRCh38
rs200454752 Chr.15: 78593246-78593246 on Build GRCh38
rs138253116 Chr.15: 78593131-78593131 on Build GRCh38
rs770154759 Chr.15: 78593147-78593151 on Build GRCh38
rs201776618 Chr.15: 78593359-78593359 on Build GRCh38
rs768799141 Chr.15: 78593261-78593261 on Build GRCh38
rs759802002 Chr.15: 78593150-78593150 on Build GRCh38
rs200357731 Chr.15: 78593164-78593164 on Build GRCh38
rs553608776 Chr.15: 78592943-78592943 on Build GRCh38
rs776726741 Chr.15: 78593143-78593143 on Build GRCh38
rs1038461611 Chr.15: 78593146-78593146 on Build GRCh38
rs922012434 Chr.15: 78593017-78593017 on Build GRCh38
rs372674046 Chr.15: 78593293-78593293 on Build GRCh38
rs200911665 Chr.15: 78593272-78593272 on Build GRCh38
rs748912077 Chr.15: 78593095-78593095 on Build GRCh38
rs773626999 Chr.15: 78593154-78593158 on Build GRCh38

View Full Product Details