Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 12663023-12663544 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPPED1
Entrez Gene ID
55313
Gene Name
calcineurin like phosphoesterase domain containing 1
Gene Aliases
CSTP1
UniGene
Hs.460002
Gene Chromosome Location
Chr.16: 12659799-12803887 on Build GRCh38
Transcripts
NM_001099455.1 NM_018340.2

SNP Detail

SNP ID Chromosome Location
rs944943152 Chr.16: 12663062-12663062 on Build GRCh38
rs897744541 Chr.16: 12663460-12663460 on Build GRCh38
rs906924042 Chr.16: 12663231-12663231 on Build GRCh38
rs907826844 Chr.16: 12663394-12663394 on Build GRCh38
rs937069962 Chr.16: 12663383-12663383 on Build GRCh38
rs146778582 Chr.16: 12663183-12663183 on Build GRCh38
rs186167576 Chr.16: 12663331-12663331 on Build GRCh38
rs59120394 Chr.16: 12663062-12663063 on Build GRCh38
rs895644234 Chr.16: 12663212-12663212 on Build GRCh38
rs565007409 Chr.16: 12663215-12663215 on Build GRCh38
rs1055935470 Chr.16: 12663243-12663243 on Build GRCh38
rs1052650284 Chr.16: 12663063-12663063 on Build GRCh38
rs140486372 Chr.16: 12663266-12663266 on Build GRCh38
rs988260069 Chr.16: 12663498-12663498 on Build GRCh38
rs560828089 Chr.16: 12663326-12663326 on Build GRCh38
rs903170482 Chr.16: 12663302-12663302 on Build GRCh38
rs770940296 Chr.16: 12663063-12663064 on Build GRCh38
rs796934470 Chr.16: 12663245-12663245 on Build GRCh38
rs200461098 Chr.16: 12663096-12663096 on Build GRCh38
rs538863388 Chr.16: 12663359-12663359 on Build GRCh38
rs537116469 Chr.16: 12663384-12663384 on Build GRCh38
rs1042732305 Chr.16: 12663104-12663104 on Build GRCh38
rs761050057 Chr.16: 12663515-12663515 on Build GRCh38
rs745555005 Chr.16: 12663062-12663064 on Build GRCh38
rs994761962 Chr.16: 12663465-12663465 on Build GRCh38
rs984655008 Chr.16: 12663386-12663386 on Build GRCh38
rs937106831 Chr.16: 12663232-12663232 on Build GRCh38
rs889129308 Chr.16: 12663476-12663476 on Build GRCh38
rs1027623810 Chr.16: 12663469-12663469 on Build GRCh38
rs397974969 Chr.16: 12663079-12663080 on Build GRCh38
rs3097190 Chr.16: 12663352-12663352 on Build GRCh38
rs79448375 Chr.16: 12663075-12663075 on Build GRCh38
rs3074348 Chr.16: 12663078-12663079 on Build GRCh38
rs552106611 Chr.16: 12663198-12663198 on Build GRCh38
rs1045878748 Chr.16: 12663220-12663220 on Build GRCh38
rs550598904 Chr.16: 12663269-12663269 on Build GRCh38
rs974578002 Chr.16: 12663148-12663148 on Build GRCh38
rs145170969 Chr.16: 12663150-12663150 on Build GRCh38
rs746637741 Chr.16: 12663124-12663124 on Build GRCh38
rs964120668 Chr.16: 12663340-12663340 on Build GRCh38
rs930527166 Chr.16: 12663155-12663155 on Build GRCh38
rs987269327 Chr.16: 12663113-12663113 on Build GRCh38
rs925612719 Chr.16: 12663371-12663371 on Build GRCh38
rs763203701 Chr.16: 12663206-12663206 on Build GRCh38
rs145655717 Chr.16: 12663341-12663341 on Build GRCh38
rs372123199 Chr.16: 12663426-12663426 on Build GRCh38
rs375717118 Chr.16: 12663195-12663195 on Build GRCh38
rs141075724 Chr.16: 12663102-12663102 on Build GRCh38
rs1055590519 Chr.16: 12663496-12663496 on Build GRCh38
rs879179497 Chr.16: 12663062-12663062 on Build GRCh38
rs554468049 Chr.16: 12663408-12663408 on Build GRCh38
rs367922487 Chr.16: 12663103-12663103 on Build GRCh38
rs912035814 Chr.16: 12663327-12663327 on Build GRCh38
rs962284540 Chr.16: 12663395-12663395 on Build GRCh38
rs557021671 Chr.16: 12663483-12663483 on Build GRCh38
rs571408244 Chr.16: 12663365-12663365 on Build GRCh38
rs769457327 Chr.16: 12663520-12663520 on Build GRCh38
rs953451577 Chr.16: 12663307-12663307 on Build GRCh38
rs768526982 Chr.16: 12663146-12663146 on Build GRCh38
rs907546410 Chr.16: 12663185-12663185 on Build GRCh38
rs1019429466 Chr.16: 12663495-12663495 on Build GRCh38
rs1019723206 Chr.16: 12663115-12663115 on Build GRCh38
rs1001729937 Chr.16: 12663167-12663167 on Build GRCh38
rs1020569892 Chr.16: 12663285-12663285 on Build GRCh38
rs552054820 Chr.16: 12663364-12663364 on Build GRCh38
rs994833181 Chr.16: 12663272-12663272 on Build GRCh38
rs912877321 Chr.16: 12663086-12663086 on Build GRCh38
rs981109511 Chr.16: 12663407-12663407 on Build GRCh38
rs367736942 Chr.16: 12663252-12663252 on Build GRCh38

View Full Product Details