Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 15604090-15604601 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA0430
Entrez Gene ID
9665
Gene Name
KIAA0430
Gene Aliases
LKAP, MARF1, PPP1R34
UniGene
Hs.173524
Gene Chromosome Location
Chr.16: 15594369-15643166 on Build GRCh38
Transcripts
NM_001184998.1 NM_001184999.1 NM_014647.3 XM_005255704.1 XM_005255706.1 XM_005255707.1 XM_017023900.1 XM_017023901.1 XM_017023902.1 XM_017023903.1 XM_017023904.1 XM_017023905.1 XM_017023906.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3817264 c.3080C>A p.S1027Y Chr.16: 15604307-15604307 on Build GRCh38
2126826 c.4399C>T p.P1467S Chr.16: 15604182-15604182 on Build GRCh38
5028082 c.4325A>G p.E1442G Chr.16: 15604256-15604256 on Build GRCh38
6489355 c.2989G>A p.V997I Chr.16: 15604398-15604398 on Build GRCh38
6693304 c.3043C>T p.R1015* Chr.16: 15604344-15604344 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs568500227 Chr.16: 15604277-15604277 on Build GRCh38
rs527881323 Chr.16: 15604149-15604149 on Build GRCh38
rs369367491 Chr.16: 15604208-15604208 on Build GRCh38
rs781778542 Chr.16: 15604125-15604125 on Build GRCh38
rs549312748 Chr.16: 15604156-15604156 on Build GRCh38
rs910971035 Chr.16: 15604318-15604318 on Build GRCh38
rs542321365 Chr.16: 15604499-15604499 on Build GRCh38
rs374820287 Chr.16: 15604388-15604388 on Build GRCh38
rs557628641 Chr.16: 15604390-15604390 on Build GRCh38
rs765744884 Chr.16: 15604121-15604121 on Build GRCh38
rs754294853 Chr.16: 15604256-15604256 on Build GRCh38
rs1045062243 Chr.16: 15604528-15604528 on Build GRCh38
rs751739331 Chr.16: 15604346-15604347 on Build GRCh38
rs760963901 Chr.16: 15604120-15604120 on Build GRCh38
rs370510789 Chr.16: 15604140-15604140 on Build GRCh38
rs772881542 Chr.16: 15604304-15604304 on Build GRCh38
rs941364094 Chr.16: 15604526-15604526 on Build GRCh38
rs771454240 Chr.16: 15604509-15604509 on Build GRCh38
rs1044095358 Chr.16: 15604512-15604512 on Build GRCh38
rs747162284 Chr.16: 15604381-15604381 on Build GRCh38
rs772096894 Chr.16: 15604276-15604276 on Build GRCh38
rs757677863 Chr.16: 15604357-15604357 on Build GRCh38
rs892650736 Chr.16: 15604558-15604558 on Build GRCh38
rs146524775 Chr.16: 15604124-15604124 on Build GRCh38
rs757171070 Chr.16: 15604294-15604294 on Build GRCh38
rs750006732 Chr.16: 15604118-15604118 on Build GRCh38
rs1031458637 Chr.16: 15604543-15604543 on Build GRCh38
rs770470455 Chr.16: 15604314-15604314 on Build GRCh38
rs764719534 Chr.16: 15604338-15604338 on Build GRCh38
rs778230527 Chr.16: 15604129-15604129 on Build GRCh38
rs758628280 Chr.16: 15604375-15604375 on Build GRCh38
rs371389972 Chr.16: 15604302-15604302 on Build GRCh38
rs777933126 Chr.16: 15604248-15604248 on Build GRCh38
rs762449561 Chr.16: 15604185-15604185 on Build GRCh38
rs539088364 Chr.16: 15604297-15604297 on Build GRCh38
rs757406736 Chr.16: 15604257-15604257 on Build GRCh38
rs750192331 Chr.16: 15604557-15604557 on Build GRCh38
rs139962572 Chr.16: 15604258-15604258 on Build GRCh38
rs764475769 Chr.16: 15604444-15604444 on Build GRCh38
rs771916744 Chr.16: 15604425-15604425 on Build GRCh38
rs765681821 Chr.16: 15604343-15604343 on Build GRCh38
rs756585649 Chr.16: 15604128-15604128 on Build GRCh38
rs762172787 Chr.16: 15604141-15604141 on Build GRCh38
rs748500355 Chr.16: 15604126-15604126 on Build GRCh38
rs777153390 Chr.16: 15604445-15604445 on Build GRCh38
rs745460512 Chr.16: 15604417-15604417 on Build GRCh38
rs942446405 Chr.16: 15604347-15604347 on Build GRCh38
rs1003609111 Chr.16: 15604554-15604554 on Build GRCh38
rs767088126 Chr.16: 15604327-15604327 on Build GRCh38
rs201602388 Chr.16: 15604201-15604201 on Build GRCh38
rs368216867 Chr.16: 15604179-15604179 on Build GRCh38
rs752268378 Chr.16: 15604328-15604328 on Build GRCh38
rs888348848 Chr.16: 15604519-15604519 on Build GRCh38
rs749422760 Chr.16: 15604133-15604133 on Build GRCh38
rs939576567 Chr.16: 15604410-15604410 on Build GRCh38
rs190872760 Chr.16: 15604531-15604531 on Build GRCh38
rs760280057 Chr.16: 15604438-15604438 on Build GRCh38
rs745914446 Chr.16: 15604260-15604260 on Build GRCh38
rs568115023 Chr.16: 15604200-15604200 on Build GRCh38
rs754129515 Chr.16: 15604351-15604351 on Build GRCh38
rs761258455 Chr.16: 15604333-15604333 on Build GRCh38
rs759294926 Chr.16: 15604316-15604316 on Build GRCh38
rs375289823 Chr.16: 15604159-15604159 on Build GRCh38
rs761767509 Chr.16: 15604160-15604160 on Build GRCh38
rs768556384 Chr.16: 15604153-15604153 on Build GRCh38
rs773077616 Chr.16: 15604180-15604180 on Build GRCh38
rs368968955 Chr.16: 15604434-15604434 on Build GRCh38
rs770720625 Chr.16: 15604409-15604409 on Build GRCh38
rs750590815 Chr.16: 15604360-15604360 on Build GRCh38
rs779173015 Chr.16: 15604359-15604359 on Build GRCh38
rs895422269 Chr.16: 15604451-15604451 on Build GRCh38
rs528902067 Chr.16: 15604321-15604321 on Build GRCh38
rs948342295 Chr.16: 15604456-15604456 on Build GRCh38
rs774188740 Chr.16: 15604315-15604315 on Build GRCh38
rs751081670 Chr.16: 15604123-15604123 on Build GRCh38
rs769525367 Chr.16: 15604174-15604174 on Build GRCh38
rs753216531 Chr.16: 15604231-15604231 on Build GRCh38
rs766125540 Chr.16: 15604187-15604187 on Build GRCh38
rs762246827 Chr.16: 15604447-15604447 on Build GRCh38
rs780046934 Chr.16: 15604379-15604379 on Build GRCh38
rs748017051 Chr.16: 15604295-15604295 on Build GRCh38
rs749007150 Chr.16: 15604392-15604392 on Build GRCh38
rs756712115 Chr.16: 15604238-15604238 on Build GRCh38
rs762867615 Chr.16: 15604308-15604308 on Build GRCh38

View Full Product Details