Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 20784859-20785404 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACSM3
Entrez Gene ID
6296
Gene Name
acyl-CoA synthetase medium-chain family member 3
Gene Aliases
SA, SAH
UniGene
Hs.706754
Gene Chromosome Location
Chr.16: 20763990-20797157 on Build GRCh38
Transcripts
NM_005622.3 NM_202000.2 XM_011545911.1 XM_017023523.1 XM_017023524.1 NM_080663.2

Gene Symbol
ERI2
Entrez Gene ID
112479
Gene Name
ERI1 exoribonuclease family member 2
Gene Aliases
EXOD1, ZGRF5
UniGene
Hs.248437
Gene Chromosome Location
Chr.16: 20780193-20806473 on Build GRCh38
Transcripts
NM_005622.3 NM_202000.2 XM_011545911.1 XM_017023523.1 XM_017023524.1 NM_080663.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4419512 c.1101G>C p.K367N Chr.16: 20785065-20785065 on Build GRCh38
6442939 c.1034G>A p.S345N Chr.16: 20784998-20784998 on Build GRCh38
2130173 c.1029_1030insA p.S345fs*15 Chr.16: 20784993-20784994 on Build GRCh38
4059007 c.1103C>T p.T368M Chr.16: 20785067-20785067 on Build GRCh38
4419513 c.1101G>C p.K367N Chr.16: 20785065-20785065 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752645636 Chr.16: 20784964-20784964 on Build GRCh38
rs779279874 Chr.16: 20785033-20785033 on Build GRCh38
rs370252816 Chr.16: 20785340-20785340 on Build GRCh38
rs1001163441 Chr.16: 20785031-20785031 on Build GRCh38
rs745783736 Chr.16: 20784910-20784910 on Build GRCh38
rs748181119 Chr.16: 20785067-20785067 on Build GRCh38
rs1057127766 Chr.16: 20784894-20784894 on Build GRCh38
rs992826530 Chr.16: 20785206-20785206 on Build GRCh38
rs750153921 Chr.16: 20784986-20784986 on Build GRCh38
rs374096382 Chr.16: 20785105-20785105 on Build GRCh38
rs147257547 Chr.16: 20785016-20785016 on Build GRCh38
rs749349259 Chr.16: 20785020-20785020 on Build GRCh38
rs772910375 Chr.16: 20785063-20785063 on Build GRCh38
rs181255157 Chr.16: 20785311-20785311 on Build GRCh38
rs950479891 Chr.16: 20785069-20785069 on Build GRCh38
rs934470163 Chr.16: 20785336-20785336 on Build GRCh38
rs1053305775 Chr.16: 20785359-20785359 on Build GRCh38
rs978648869 Chr.16: 20785241-20785241 on Build GRCh38
rs776152834 Chr.16: 20785042-20785042 on Build GRCh38
rs1038958090 Chr.16: 20785330-20785330 on Build GRCh38
rs748388350 Chr.16: 20785346-20785346 on Build GRCh38
rs749158142 Chr.16: 20785293-20785293 on Build GRCh38
rs780518315 Chr.16: 20785117-20785117 on Build GRCh38
rs372476385 Chr.16: 20785034-20785034 on Build GRCh38
rs8058407 Chr.16: 20784967-20784967 on Build GRCh38
rs1051395112 Chr.16: 20785375-20785375 on Build GRCh38
rs201962964 Chr.16: 20785047-20785047 on Build GRCh38
rs997181499 Chr.16: 20785356-20785356 on Build GRCh38
rs936543191 Chr.16: 20785328-20785328 on Build GRCh38
rs772616069 Chr.16: 20785036-20785036 on Build GRCh38
rs145743388 Chr.16: 20785092-20785092 on Build GRCh38
rs8058758 Chr.16: 20785186-20785186 on Build GRCh38
rs1038989118 Chr.16: 20785035-20785035 on Build GRCh38
rs757346849 Chr.16: 20785104-20785104 on Build GRCh38
rs766197968 Chr.16: 20785068-20785068 on Build GRCh38
rs758724882 Chr.16: 20785107-20785107 on Build GRCh38
rs199851319 Chr.16: 20785073-20785073 on Build GRCh38
rs144411258 Chr.16: 20785140-20785140 on Build GRCh38
rs774785835 Chr.16: 20785320-20785320 on Build GRCh38
rs537932144 Chr.16: 20785050-20785050 on Build GRCh38
rs749376809 Chr.16: 20785066-20785066 on Build GRCh38
rs538407351 Chr.16: 20785084-20785084 on Build GRCh38
rs1016815589 Chr.16: 20785093-20785093 on Build GRCh38
rs758260429 Chr.16: 20784987-20784987 on Build GRCh38
rs779951052 Chr.16: 20784993-20784993 on Build GRCh38
rs879220574 Chr.16: 20785112-20785112 on Build GRCh38
rs554041477 Chr.16: 20785106-20785106 on Build GRCh38
rs139667199 Chr.16: 20785041-20785041 on Build GRCh38
rs747351317 Chr.16: 20785132-20785132 on Build GRCh38
rs373248833 Chr.16: 20785098-20785098 on Build GRCh38
rs202155170 Chr.16: 20785083-20785083 on Build GRCh38
rs5716 Chr.16: 20785065-20785065 on Build GRCh38
rs781063792 Chr.16: 20784943-20784944 on Build GRCh38
rs756112490 Chr.16: 20785014-20785014 on Build GRCh38
rs751545090 Chr.16: 20785000-20785000 on Build GRCh38
rs1005726383 Chr.16: 20785006-20785006 on Build GRCh38
rs913240126 Chr.16: 20785208-20785208 on Build GRCh38
rs548962433 Chr.16: 20785037-20785037 on Build GRCh38
rs369017155 Chr.16: 20785030-20785030 on Build GRCh38
rs997845979 Chr.16: 20784901-20784901 on Build GRCh38
rs761481939 Chr.16: 20784952-20784952 on Build GRCh38

View Full Product Details