Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67201383-67201882 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ELMO3
Entrez Gene ID
79767
Gene Name
engulfment and cell motility 3
Gene Aliases
CED-12, CED12, ELMO-3
UniGene
Hs.377416
Gene Chromosome Location
Chr.16: 67199125-67204024 on Build GRCh38
Transcripts
NM_024712.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1205211 c.1169G>A p.R390H Chr.16: 67201833-67201833 on Build GRCh38
4579156 c.1008C>T p.Y336Y Chr.16: 67201573-67201573 on Build GRCh38
460502 c.1186G>C p.E396Q Chr.16: 67201850-67201850 on Build GRCh38
3510955 c.919C>T p.L307F Chr.16: 67201400-67201400 on Build GRCh38
972343 c.1115C>G p.A372G Chr.16: 67201779-67201779 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773884075 Chr.16: 67201589-67201589 on Build GRCh38
rs776266737 Chr.16: 67201821-67201821 on Build GRCh38
rs777708052 Chr.16: 67201617-67201617 on Build GRCh38
rs778678997 Chr.16: 67201723-67201723 on Build GRCh38
rs767605719 Chr.16: 67201800-67201800 on Build GRCh38
rs199860236 Chr.16: 67201601-67201601 on Build GRCh38
rs765904297 Chr.16: 67201656-67201656 on Build GRCh38
rs745529398 Chr.16: 67201826-67201826 on Build GRCh38
rs375408698 Chr.16: 67201609-67201609 on Build GRCh38
rs33948247 Chr.16: 67201769-67201769 on Build GRCh38
rs779511488 Chr.16: 67201610-67201610 on Build GRCh38
rs377314519 Chr.16: 67201414-67201414 on Build GRCh38
rs762380682 Chr.16: 67201835-67201835 on Build GRCh38
rs368459803 Chr.16: 67201734-67201734 on Build GRCh38
rs756553568 Chr.16: 67201541-67201541 on Build GRCh38
rs751012893 Chr.16: 67201504-67201504 on Build GRCh38
rs746931182 Chr.16: 67201817-67201817 on Build GRCh38
rs201654935 Chr.16: 67201480-67201480 on Build GRCh38
rs375040978 Chr.16: 67201750-67201750 on Build GRCh38
rs944539765 Chr.16: 67201537-67201537 on Build GRCh38
rs747875589 Chr.16: 67201804-67201804 on Build GRCh38
rs751683195 Chr.16: 67201783-67201783 on Build GRCh38
rs766535773 Chr.16: 67201857-67201857 on Build GRCh38
rs569287419 Chr.16: 67201549-67201549 on Build GRCh38
rs201796351 Chr.16: 67201806-67201806 on Build GRCh38
rs927810942 Chr.16: 67201671-67201671 on Build GRCh38
rs763179548 Chr.16: 67201830-67201830 on Build GRCh38
rs748145779 Chr.16: 67201724-67201724 on Build GRCh38
rs200639831 Chr.16: 67201803-67201803 on Build GRCh38
rs768605277 Chr.16: 67201628-67201628 on Build GRCh38
rs973161117 Chr.16: 67201619-67201619 on Build GRCh38
rs183146864 Chr.16: 67201574-67201574 on Build GRCh38
rs1023892455 Chr.16: 67201464-67201464 on Build GRCh38
rs747167131 Chr.16: 67201562-67201562 on Build GRCh38
rs750777098 Chr.16: 67201528-67201528 on Build GRCh38
rs757973944 Chr.16: 67201431-67201431 on Build GRCh38
rs200309966 Chr.16: 67201720-67201720 on Build GRCh38
rs532593956 Chr.16: 67201827-67201827 on Build GRCh38
rs946624385 Chr.16: 67201784-67201784 on Build GRCh38
rs79736950 Chr.16: 67201748-67201748 on Build GRCh38
rs774765515 Chr.16: 67201405-67201405 on Build GRCh38
rs763563032 Chr.16: 67201433-67201433 on Build GRCh38
rs751447505 Chr.16: 67201796-67201796 on Build GRCh38
rs765179389 Chr.16: 67201418-67201418 on Build GRCh38
rs761722485 Chr.16: 67201570-67201573 on Build GRCh38
rs375594879 Chr.16: 67201481-67201481 on Build GRCh38
rs756891097 Chr.16: 67201448-67201448 on Build GRCh38
rs151223840 Chr.16: 67201555-67201555 on Build GRCh38
rs906403275 Chr.16: 67201536-67201536 on Build GRCh38
rs765664148 Chr.16: 67201674-67201674 on Build GRCh38
rs368732433 Chr.16: 67201697-67201697 on Build GRCh38
rs190200837 Chr.16: 67201490-67201490 on Build GRCh38
rs201261849 Chr.16: 67201573-67201573 on Build GRCh38
rs757530514 Chr.16: 67201712-67201712 on Build GRCh38
rs879141454 Chr.16: 67201735-67201735 on Build GRCh38
rs375654241 Chr.16: 67201522-67201522 on Build GRCh38
rs779904982 Chr.16: 67201621-67201621 on Build GRCh38
rs377324421 Chr.16: 67201833-67201833 on Build GRCh38
rs557316702 Chr.16: 67201613-67201613 on Build GRCh38
rs1027293285 Chr.16: 67201698-67201698 on Build GRCh38
rs757174577 Chr.16: 67201848-67201848 on Build GRCh38
rs1052344385 Chr.16: 67201679-67201679 on Build GRCh38
rs770450376 Chr.16: 67201749-67201749 on Build GRCh38
rs750448202 Chr.16: 67201847-67201847 on Build GRCh38
rs775535583 Chr.16: 67201500-67201500 on Build GRCh38
rs542665221 Chr.16: 67201632-67201632 on Build GRCh38
rs765489952 Chr.16: 67201586-67201586 on Build GRCh38
rs960558590 Chr.16: 67201837-67201837 on Build GRCh38
rs764917023 Chr.16: 67201491-67201491 on Build GRCh38
rs766828979 Chr.16: 67201608-67201608 on Build GRCh38
rs753320498 Chr.16: 67201692-67201692 on Build GRCh38
rs765753879 Chr.16: 67201751-67201751 on Build GRCh38
rs775939061 Chr.16: 67201765-67201765 on Build GRCh38
rs370132419 Chr.16: 67201624-67201624 on Build GRCh38
rs140352129 Chr.16: 67201590-67201590 on Build GRCh38
rs777617654 Chr.16: 67201810-67201810 on Build GRCh38
rs748723781 Chr.16: 67201498-67201499 on Build GRCh38
rs369916018 Chr.16: 67201419-67201419 on Build GRCh38
rs779134617 Chr.16: 67201550-67201550 on Build GRCh38
rs762961932 Chr.16: 67201582-67201582 on Build GRCh38
rs61735429 Chr.16: 67201802-67201802 on Build GRCh38
rs746179145 Chr.16: 67201454-67201454 on Build GRCh38
rs911750078 Chr.16: 67201520-67201520 on Build GRCh38
rs767005065 Chr.16: 67201856-67201856 on Build GRCh38
rs370005444 Chr.16: 67201797-67201797 on Build GRCh38
rs374898676 Chr.16: 67201645-67201645 on Build GRCh38
rs761460312 Chr.16: 67201511-67201511 on Build GRCh38
rs780564467 Chr.16: 67201469-67201469 on Build GRCh38
rs200012079 Chr.16: 67201620-67201620 on Build GRCh38
rs776596771 Chr.16: 67201487-67201487 on Build GRCh38
rs970164034 Chr.16: 67201468-67201468 on Build GRCh38
rs1004898980 Chr.16: 67201762-67201762 on Build GRCh38
rs751279301 Chr.16: 67201443-67201443 on Build GRCh38
rs572778974 Chr.16: 67201829-67201829 on Build GRCh38
rs760244384 Chr.16: 67201603-67201603 on Build GRCh38
rs187434892 Chr.16: 67201721-67201721 on Build GRCh38
rs369902689 Chr.16: 67201726-67201726 on Build GRCh38
rs767645603 Chr.16: 67201845-67201845 on Build GRCh38
rs751527760 Chr.16: 67201704-67201704 on Build GRCh38
rs939304459 Chr.16: 67201676-67201676 on Build GRCh38
rs764468014 Chr.16: 67201771-67201771 on Build GRCh38
rs754439840 Chr.16: 67201547-67201547 on Build GRCh38
rs762794419 Chr.16: 67201503-67201503 on Build GRCh38
rs372317058 Chr.16: 67201470-67201470 on Build GRCh38
rs760025178 Chr.16: 67201662-67201662 on Build GRCh38
rs367683482 Chr.16: 67201611-67201611 on Build GRCh38
rs376387864 Chr.16: 67201700-67201700 on Build GRCh38
rs765003559 Chr.16: 67201789-67201795 on Build GRCh38
rs371124873 Chr.16: 67201569-67201569 on Build GRCh38
rs372685567 Chr.16: 67201719-67201719 on Build GRCh38
rs756611393 Chr.16: 67201460-67201460 on Build GRCh38
rs762100224 Chr.16: 67201413-67201413 on Build GRCh38
rs374988066 Chr.16: 67201777-67201777 on Build GRCh38
rs147009177 Chr.16: 67201832-67201832 on Build GRCh38
rs369051966 Chr.16: 67201553-67201553 on Build GRCh38
rs775233857 Chr.16: 67201659-67201659 on Build GRCh38
rs748194145 Chr.16: 67201552-67201552 on Build GRCh38
rs988478157 Chr.16: 67201850-67201850 on Build GRCh38
rs760392627 Chr.16: 67201651-67201651 on Build GRCh38
rs759394106 Chr.16: 67201417-67201417 on Build GRCh38
rs565522399 Chr.16: 67201449-67201449 on Build GRCh38
rs772588917 Chr.16: 67201649-67201649 on Build GRCh38
rs770411417 Chr.16: 67201818-67201818 on Build GRCh38
rs929048380 Chr.16: 67201488-67201488 on Build GRCh38
rs774323005 Chr.16: 67201631-67201631 on Build GRCh38
rs780253107 Chr.16: 67201544-67201544 on Build GRCh38

View Full Product Details