Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 68230749-68231253 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESRP2
Entrez Gene ID
80004
Gene Name
epithelial splicing regulatory protein 2
Gene Aliases
RBM35B
UniGene
Hs.436585
Gene Chromosome Location
Chr.16: 68228547-68236233 on Build GRCh38
Transcripts
NM_024939.2 XM_005256153.4 XM_006721273.3 XM_011523338.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4883651 c.1736C>A p.T579N Chr.16: 68231003-68231003 on Build GRCh38
4829101 c.1676G>A p.R559H Chr.16: 68231213-68231213 on Build GRCh38
5715823 c.1727C>T p.P576L Chr.16: 68231192-68231192 on Build GRCh38
471978 c.1733C>G p.T578S Chr.16: 68231006-68231006 on Build GRCh38
1379080 c.1773G>A p.T591T Chr.16: 68230966-68230966 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs913729915 Chr.16: 68231180-68231180 on Build GRCh38
rs754700540 Chr.16: 68230903-68230904 on Build GRCh38
rs1008336783 Chr.16: 68231225-68231225 on Build GRCh38
rs749273721 Chr.16: 68231170-68231170 on Build GRCh38
rs760300022 Chr.16: 68231212-68231212 on Build GRCh38
rs556537582 Chr.16: 68230967-68230967 on Build GRCh38
rs747339669 Chr.16: 68230844-68230844 on Build GRCh38
rs117260313 Chr.16: 68230951-68230951 on Build GRCh38
rs749792127 Chr.16: 68230972-68230972 on Build GRCh38
rs766467821 Chr.16: 68230824-68230824 on Build GRCh38
rs139416619 Chr.16: 68230858-68230858 on Build GRCh38
rs765673788 Chr.16: 68231229-68231229 on Build GRCh38
rs373486598 Chr.16: 68230834-68230834 on Build GRCh38
rs200279617 Chr.16: 68230912-68230912 on Build GRCh38
rs986045866 Chr.16: 68230827-68230827 on Build GRCh38
rs748358961 Chr.16: 68230985-68230985 on Build GRCh38
rs965935315 Chr.16: 68230869-68230869 on Build GRCh38
rs763196838 Chr.16: 68230790-68230790 on Build GRCh38
rs199808836 Chr.16: 68231030-68231030 on Build GRCh38
rs758120670 Chr.16: 68231104-68231104 on Build GRCh38
rs201160955 Chr.16: 68230820-68230820 on Build GRCh38
rs199906536 Chr.16: 68231172-68231172 on Build GRCh38
rs774429319 Chr.16: 68230803-68230803 on Build GRCh38
rs756780323 Chr.16: 68230856-68230856 on Build GRCh38
rs761396911 Chr.16: 68231034-68231034 on Build GRCh38
rs781395192 Chr.16: 68231161-68231161 on Build GRCh38
rs368193048 Chr.16: 68231140-68231140 on Build GRCh38
rs557967621 Chr.16: 68231107-68231107 on Build GRCh38
rs36054935 Chr.16: 68230890-68230890 on Build GRCh38
rs562625559 Chr.16: 68230778-68230778 on Build GRCh38
rs758560628 Chr.16: 68231129-68231129 on Build GRCh38
rs763972371 Chr.16: 68230938-68230938 on Build GRCh38
rs752499069 Chr.16: 68230832-68230832 on Build GRCh38
rs756906275 Chr.16: 68230948-68230948 on Build GRCh38
rs1046606817 Chr.16: 68230989-68230989 on Build GRCh38
rs762966945 Chr.16: 68231020-68231020 on Build GRCh38
rs768871334 Chr.16: 68230885-68230885 on Build GRCh38
rs184799308 Chr.16: 68231201-68231201 on Build GRCh38
rs759422467 Chr.16: 68230909-68230909 on Build GRCh38
rs755446587 Chr.16: 68231058-68231058 on Build GRCh38
rs762766253 Chr.16: 68230928-68230928 on Build GRCh38
rs779280493 Chr.16: 68231059-68231059 on Build GRCh38
rs537836223 Chr.16: 68230954-68230954 on Build GRCh38
rs577951281 Chr.16: 68231109-68231109 on Build GRCh38
rs755393677 Chr.16: 68230973-68230973 on Build GRCh38
rs760057049 Chr.16: 68231219-68231219 on Build GRCh38
rs951860481 Chr.16: 68230891-68230891 on Build GRCh38
rs552251668 Chr.16: 68230941-68230941 on Build GRCh38
rs753055981 Chr.16: 68231063-68231063 on Build GRCh38
rs564952164 Chr.16: 68231087-68231087 on Build GRCh38
rs192768082 Chr.16: 68231044-68231044 on Build GRCh38
rs200796602 Chr.16: 68231148-68231148 on Build GRCh38
rs777506617 Chr.16: 68230837-68230837 on Build GRCh38
rs751558791 Chr.16: 68231189-68231189 on Build GRCh38
rs954057531 Chr.16: 68231121-68231121 on Build GRCh38
rs117732754 Chr.16: 68230774-68230774 on Build GRCh38
rs756594868 Chr.16: 68230966-68230966 on Build GRCh38
rs768819927 Chr.16: 68230796-68230796 on Build GRCh38
rs370690332 Chr.16: 68231214-68231214 on Build GRCh38
rs577046779 Chr.16: 68231103-68231103 on Build GRCh38
rs111882305 Chr.16: 68231081-68231081 on Build GRCh38
rs759495566 Chr.16: 68230821-68230821 on Build GRCh38
rs751115440 Chr.16: 68230850-68230850 on Build GRCh38
rs756606781 Chr.16: 68230876-68230876 on Build GRCh38
rs987900470 Chr.16: 68231158-68231158 on Build GRCh38
rs377474438 Chr.16: 68230979-68230979 on Build GRCh38
rs775436384 Chr.16: 68230920-68230920 on Build GRCh38
rs888716792 Chr.16: 68231043-68231043 on Build GRCh38
rs773185656 Chr.16: 68230995-68230995 on Build GRCh38
rs749599581 Chr.16: 68230877-68230877 on Build GRCh38
rs769473702 Chr.16: 68231178-68231178 on Build GRCh38
rs557228511 Chr.16: 68231149-68231149 on Build GRCh38
rs201276474 Chr.16: 68230805-68230805 on Build GRCh38
rs559554269 Chr.16: 68230787-68230787 on Build GRCh38
rs145134851 Chr.16: 68231003-68231003 on Build GRCh38
rs773310955 Chr.16: 68230905-68230905 on Build GRCh38
rs372642622 Chr.16: 68230793-68230793 on Build GRCh38
rs965453588 Chr.16: 68230782-68230782 on Build GRCh38
rs748388857 Chr.16: 68230888-68230888 on Build GRCh38
rs771762193 Chr.16: 68231190-68231190 on Build GRCh38
rs1046130892 Chr.16: 68231222-68231222 on Build GRCh38
rs1028848272 Chr.16: 68231120-68231120 on Build GRCh38
rs772296415 Chr.16: 68230988-68230988 on Build GRCh38
rs755718291 Chr.16: 68231041-68231041 on Build GRCh38
rs201684281 Chr.16: 68231035-68231035 on Build GRCh38
rs375171498 Chr.16: 68231036-68231036 on Build GRCh38
rs925416130 Chr.16: 68231094-68231094 on Build GRCh38
rs979184348 Chr.16: 68231127-68231127 on Build GRCh38
rs764980660 Chr.16: 68230825-68230825 on Build GRCh38
rs774032420 Chr.16: 68231185-68231185 on Build GRCh38
rs775632478 Chr.16: 68231010-68231010 on Build GRCh38
rs754392299 Chr.16: 68231049-68231049 on Build GRCh38
rs774252203 Chr.16: 68231033-68231033 on Build GRCh38
rs200397589 Chr.16: 68231008-68231008 on Build GRCh38
rs949284278 Chr.16: 68231213-68231213 on Build GRCh38
rs530015102 Chr.16: 68230779-68230779 on Build GRCh38
rs772977411 Chr.16: 68231204-68231204 on Build GRCh38
rs752592064 Chr.16: 68231032-68231032 on Build GRCh38

View Full Product Details