Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 70454489-70454993 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FUK
Entrez Gene ID
197258
Gene Name
fucokinase
Gene Aliases
1110046B12Rik
UniGene
Hs.7907
Gene Chromosome Location
Chr.16: 70454421-70480274 on Build GRCh38
Transcripts
NM_145059.2 XM_006721162.1 XM_011522928.1 XM_011522929.1 XM_011522930.2 XM_011522931.2 XM_017023013.1

SNP Detail

SNP ID Chromosome Location
rs920396417 Chr.16: 70454577-70454577 on Build GRCh38
rs940419434 Chr.16: 70454561-70454561 on Build GRCh38
rs910409066 Chr.16: 70454707-70454707 on Build GRCh38
rs544269919 Chr.16: 70454529-70454530 on Build GRCh38
rs1021118316 Chr.16: 70454787-70454787 on Build GRCh38
rs1050265907 Chr.16: 70454595-70454595 on Build GRCh38
rs1002371761 Chr.16: 70454651-70454651 on Build GRCh38
rs549213810 Chr.16: 70454658-70454666 on Build GRCh38
rs968580483 Chr.16: 70454643-70454643 on Build GRCh38
rs1040280156 Chr.16: 70454717-70454717 on Build GRCh38
rs17880103 Chr.16: 70454709-70454709 on Build GRCh38
rs563659303 Chr.16: 70454626-70454626 on Build GRCh38
rs1036134392 Chr.16: 70454562-70454562 on Build GRCh38
rs564010822 Chr.16: 70454645-70454645 on Build GRCh38
rs961976535 Chr.16: 70454796-70454796 on Build GRCh38
rs553557044 Chr.16: 70454896-70454896 on Build GRCh38
rs1002315902 Chr.16: 70454732-70454732 on Build GRCh38
rs892426958 Chr.16: 70454718-70454718 on Build GRCh38
rs926863506 Chr.16: 70454536-70454536 on Build GRCh38
rs918627646 Chr.16: 70454902-70454902 on Build GRCh38
rs530800941 Chr.16: 70454636-70454636 on Build GRCh38
rs975931174 Chr.16: 70454900-70454900 on Build GRCh38
rs975662531 Chr.16: 70454557-70454557 on Build GRCh38
rs61610761 Chr.16: 70454925-70454925 on Build GRCh38
rs368626133 Chr.16: 70454683-70454691 on Build GRCh38
rs1037111421 Chr.16: 70454899-70454899 on Build GRCh38
rs565858524 Chr.16: 70454716-70454717 on Build GRCh38
rs938056923 Chr.16: 70454944-70454944 on Build GRCh38
rs909908814 Chr.16: 70454882-70454882 on Build GRCh38
rs541616644 Chr.16: 70454532-70454532 on Build GRCh38
rs11865500 Chr.16: 70454674-70454674 on Build GRCh38
rs1010690331 Chr.16: 70454775-70454775 on Build GRCh38
rs535249228 Chr.16: 70454861-70454861 on Build GRCh38
rs1033879620 Chr.16: 70454653-70454653 on Build GRCh38
rs567944727 Chr.16: 70454857-70454857 on Build GRCh38
rs913834109 Chr.16: 70454511-70454511 on Build GRCh38
rs916950260 Chr.16: 70454555-70454555 on Build GRCh38
rs565662155 Chr.16: 70454725-70454725 on Build GRCh38
rs1007173672 Chr.16: 70454960-70454960 on Build GRCh38
rs945317979 Chr.16: 70454921-70454921 on Build GRCh38
rs533000917 Chr.16: 70454771-70454771 on Build GRCh38
rs981395415 Chr.16: 70454533-70454533 on Build GRCh38
rs549710033 Chr.16: 70454681-70454681 on Build GRCh38
rs945291386 Chr.16: 70454518-70454518 on Build GRCh38
rs1033832141 Chr.16: 70454763-70454763 on Build GRCh38
rs8058124 Chr.16: 70454663-70454663 on Build GRCh38
rs938026417 Chr.16: 70454652-70454652 on Build GRCh38
rs913815569 Chr.16: 70454910-70454910 on Build GRCh38
rs578024710 Chr.16: 70454609-70454609 on Build GRCh38
rs992436975 Chr.16: 70454662-70454662 on Build GRCh38
rs189516298 Chr.16: 70454966-70454966 on Build GRCh38
rs545076822 Chr.16: 70454622-70454622 on Build GRCh38
rs574332001 Chr.16: 70454530-70454530 on Build GRCh38
rs959251187 Chr.16: 70454856-70454856 on Build GRCh38
rs1055230221 Chr.16: 70454957-70454957 on Build GRCh38
rs1046727831 Chr.16: 70454523-70454523 on Build GRCh38
rs903926620 Chr.16: 70454731-70454731 on Build GRCh38
rs7201442 Chr.16: 70454683-70454683 on Build GRCh38
rs556087846 Chr.16: 70454510-70454510 on Build GRCh38
rs1017257668 Chr.16: 70454730-70454730 on Build GRCh38
rs971755332 Chr.16: 70454824-70454824 on Build GRCh38
rs960885512 Chr.16: 70454770-70454770 on Build GRCh38
rs1022385030 Chr.16: 70454641-70454641 on Build GRCh38
rs903878520 Chr.16: 70454941-70454941 on Build GRCh38
rs569559493 Chr.16: 70454683-70454684 on Build GRCh38
rs754245405 Chr.16: 70454845-70454845 on Build GRCh38
rs927908914 Chr.16: 70454849-70454849 on Build GRCh38
rs559921935 Chr.16: 70454594-70454594 on Build GRCh38
rs537041244 Chr.16: 70454808-70454808 on Build GRCh38
rs1044155178 Chr.16: 70454729-70454729 on Build GRCh38
rs985754936 Chr.16: 70454574-70454574 on Build GRCh38
rs890062746 Chr.16: 70454940-70454940 on Build GRCh38
rs373019623 Chr.16: 70454631-70454631 on Build GRCh38
rs893512869 Chr.16: 70454965-70454965 on Build GRCh38
rs796685570 Chr.16: 70454526-70454526 on Build GRCh38
rs868420380 Chr.16: 70454587-70454587 on Build GRCh38
rs35528850 Chr.16: 70454549-70454550 on Build GRCh38
rs11865564 Chr.16: 70454829-70454829 on Build GRCh38
rs960936329 Chr.16: 70454659-70454659 on Build GRCh38
rs796797147 Chr.16: 70454657-70454658 on Build GRCh38
rs569676042 Chr.16: 70454793-70454793 on Build GRCh38
rs1030313743 Chr.16: 70454556-70454556 on Build GRCh38
rs1013817886 Chr.16: 70454779-70454779 on Build GRCh38
rs1018940957 Chr.16: 70454515-70454515 on Build GRCh38
rs745720046 Chr.16: 70454666-70454667 on Build GRCh38
rs1009571210 Chr.16: 70454726-70454726 on Build GRCh38
rs913762067 Chr.16: 70454606-70454606 on Build GRCh38
rs772498764 Chr.16: 70454625-70454625 on Build GRCh38
rs547153198 Chr.16: 70454719-70454719 on Build GRCh38
rs1050212506 Chr.16: 70454686-70454686 on Build GRCh38
rs960984704 Chr.16: 70454537-70454537 on Build GRCh38
rs957747081 Chr.16: 70454595-70454595 on Build GRCh38
rs532768281 Chr.16: 70454677-70454677 on Build GRCh38
rs564970869 Chr.16: 70454678-70454679 on Build GRCh38
rs34232217 Chr.16: 70454715-70454716 on Build GRCh38
rs1022860849 Chr.16: 70454531-70454531 on Build GRCh38
rs992484091 Chr.16: 70454538-70454538 on Build GRCh38
rs889029630 Chr.16: 70454613-70454613 on Build GRCh38
rs1008798376 Chr.16: 70454616-70454616 on Build GRCh38
rs551100289 Chr.16: 70454788-70454788 on Build GRCh38
rs1054701973 Chr.16: 70454961-70454961 on Build GRCh38

View Full Product Details