Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 70854942-70855476 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HYDIN
Entrez Gene ID
54768
Gene Name
HYDIN, axonemal central pair apparatus protein
Gene Aliases
CILD5, HYDIN1, HYDIN2, PPP1R31
UniGene
Hs.351914
Gene Chromosome Location
Chr.16: 70802084-71230722 on Build GRCh38
Transcripts
NM_001270974.2 XM_006721206.2 XM_011523146.2 XM_011523147.1 XM_011523148.1 XM_011523151.1 XM_011523152.1 XM_011523155.2 XM_017023346.1 XM_017023347.1 XM_017023348.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
435674 c.12152G>A p.R4051K Chr.16: 70855272-70855272 on Build GRCh38
6145172 c.12202G>C p.D4068H Chr.16: 70855222-70855222 on Build GRCh38
5048302 c.12157G>T p.A4053S Chr.16: 70855267-70855267 on Build GRCh38
7371694 c.12152G>C p.R4051T Chr.16: 70855272-70855272 on Build GRCh38
3511456 c.12190G>A p.E4064K Chr.16: 70855234-70855234 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377745805 Chr.16: 70855170-70855170 on Build GRCh38
rs953527757 Chr.16: 70855336-70855336 on Build GRCh38
rs201418753 Chr.16: 70855189-70855189 on Build GRCh38
rs773442477 Chr.16: 70855156-70855156 on Build GRCh38
rs1024127185 Chr.16: 70855098-70855098 on Build GRCh38
rs760935875 Chr.16: 70855247-70855247 on Build GRCh38
rs557014682 Chr.16: 70855265-70855265 on Build GRCh38
rs71401841 Chr.16: 70855009-70855009 on Build GRCh38
rs781244433 Chr.16: 70855117-70855117 on Build GRCh38
rs370085141 Chr.16: 70855137-70855137 on Build GRCh38
rs1001284796 Chr.16: 70855125-70855125 on Build GRCh38
rs751248140 Chr.16: 70855327-70855327 on Build GRCh38
rs748124214 Chr.16: 70855223-70855223 on Build GRCh38
rs528818285 Chr.16: 70855197-70855197 on Build GRCh38
rs374719281 Chr.16: 70855154-70855154 on Build GRCh38
rs765614220 Chr.16: 70855274-70855274 on Build GRCh38
rs2502698 Chr.16: 70855244-70855244 on Build GRCh38
rs554345447 Chr.16: 70855446-70855446 on Build GRCh38
rs781195433 Chr.16: 70855208-70855208 on Build GRCh38
rs563680162 Chr.16: 70855034-70855034 on Build GRCh38
rs770882136 Chr.16: 70855111-70855111 on Build GRCh38
rs745845573 Chr.16: 70855122-70855122 on Build GRCh38
rs754347937 Chr.16: 70855166-70855166 on Build GRCh38
rs752080404 Chr.16: 70855192-70855192 on Build GRCh38
rs368579966 Chr.16: 70855222-70855222 on Build GRCh38
rs935735527 Chr.16: 70855445-70855445 on Build GRCh38
rs758051334 Chr.16: 70855084-70855084 on Build GRCh38
rs773387381 Chr.16: 70855236-70855236 on Build GRCh38
rs768771427 Chr.16: 70855127-70855127 on Build GRCh38
rs545465566 Chr.16: 70855019-70855019 on Build GRCh38
rs796491426 Chr.16: 70855038-70855038 on Build GRCh38
rs533232453 Chr.16: 70855012-70855012 on Build GRCh38
rs535999740 Chr.16: 70855398-70855398 on Build GRCh38
rs979570327 Chr.16: 70855405-70855409 on Build GRCh38
rs377099414 Chr.16: 70855064-70855064 on Build GRCh38
rs976730509 Chr.16: 70855227-70855227 on Build GRCh38
rs751297522 Chr.16: 70855202-70855202 on Build GRCh38
rs763746683 Chr.16: 70855303-70855303 on Build GRCh38
rs375384248 Chr.16: 70855211-70855211 on Build GRCh38
rs762521550 Chr.16: 70855302-70855302 on Build GRCh38
rs759947189 Chr.16: 70855266-70855266 on Build GRCh38
rs540979088 Chr.16: 70854975-70854975 on Build GRCh38
rs140981761 Chr.16: 70855294-70855295 on Build GRCh38
rs772166531 Chr.16: 70855228-70855228 on Build GRCh38
rs568900176 Chr.16: 70855295-70855295 on Build GRCh38
rs372442147 Chr.16: 70855395-70855395 on Build GRCh38
rs1006062 Chr.16: 70855013-70855013 on Build GRCh38
rs531128027 Chr.16: 70855067-70855067 on Build GRCh38
rs777584123 Chr.16: 70855086-70855086 on Build GRCh38
rs771271181 Chr.16: 70855256-70855256 on Build GRCh38
rs891739349 Chr.16: 70855062-70855062 on Build GRCh38
rs761960214 Chr.16: 70855133-70855133 on Build GRCh38
rs546586902 Chr.16: 70855205-70855205 on Build GRCh38
rs774365209 Chr.16: 70855130-70855130 on Build GRCh38
rs200926550 Chr.16: 70855090-70855090 on Build GRCh38
rs371361556 Chr.16: 70855209-70855209 on Build GRCh38
rs567912825 Chr.16: 70855158-70855158 on Build GRCh38
rs905713867 Chr.16: 70855120-70855120 on Build GRCh38
rs368490097 Chr.16: 70855296-70855296 on Build GRCh38
rs925705504 Chr.16: 70855406-70855406 on Build GRCh38
rs760019380 Chr.16: 70855169-70855169 on Build GRCh38
rs148094332 Chr.16: 70855295-70855296 on Build GRCh38

View Full Product Details