Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 7576970-7577476 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF4A1
Entrez Gene ID
1973
Gene Name
eukaryotic translation initiation factor 4A1
Gene Aliases
DDX2A, EIF-4A, EIF4A, eIF-4A-I, eIF4A-I
UniGene
Hs.129673
Gene Chromosome Location
Chr.17: 7572706-7579006 on Build GRCh38
Transcripts
NM_001204510.1 NM_001416.3 NR_037926.1

Gene Symbol
SENP3-EIF4A1
Entrez Gene ID
100533955
Gene Name
SENP3-EIF4A1 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.129673
Gene Chromosome Location
Chr.17: 7561992-7579006 on Build GRCh38
Transcripts
NM_001204510.1 NM_001416.3 NR_037926.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4849866 c.638C>T p.S213L Chr.17: 7577357-7577357 on Build GRCh38
160678 c.664G>C p.E222Q Chr.17: 7577383-7577383 on Build GRCh38
4069943 c.621C>A p.T207T Chr.17: 7577162-7577162 on Build GRCh38
1630406 c.526A>G p.I176V Chr.17: 7577067-7577067 on Build GRCh38
7237026 c.638C>A p.S213* Chr.17: 7577357-7577357 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779945406 Chr.17: 7577198-7577198 on Build GRCh38
rs763186463 Chr.17: 7577013-7577013 on Build GRCh38
rs142972972 Chr.17: 7577007-7577007 on Build GRCh38
rs761118765 Chr.17: 7577424-7577424 on Build GRCh38
rs753238326 Chr.17: 7577144-7577144 on Build GRCh38
rs780179034 Chr.17: 7577047-7577047 on Build GRCh38
rs941398465 Chr.17: 7577030-7577030 on Build GRCh38
rs750013227 Chr.17: 7577330-7577330 on Build GRCh38
rs1053109896 Chr.17: 7577025-7577025 on Build GRCh38
rs950159666 Chr.17: 7577022-7577022 on Build GRCh38
rs184646196 Chr.17: 7577330-7577330 on Build GRCh38
rs978635014 Chr.17: 7577310-7577310 on Build GRCh38
rs904452626 Chr.17: 7577015-7577015 on Build GRCh38
rs757302772 Chr.17: 7577181-7577182 on Build GRCh38
rs189527959 Chr.17: 7577332-7577332 on Build GRCh38
rs759641276 Chr.17: 7577307-7577307 on Build GRCh38
rs545937171 Chr.17: 7577358-7577358 on Build GRCh38
rs932753506 Chr.17: 7577087-7577087 on Build GRCh38
rs865904208 Chr.17: 7577416-7577416 on Build GRCh38
rs373835260 Chr.17: 7577370-7577370 on Build GRCh38
rs575267818 Chr.17: 7577353-7577353 on Build GRCh38
rs373643163 Chr.17: 7577319-7577320 on Build GRCh38
rs756704876 Chr.17: 7577179-7577179 on Build GRCh38
rs567749350 Chr.17: 7577273-7577273 on Build GRCh38
rs746903969 Chr.17: 7577052-7577052 on Build GRCh38
rs775553075 Chr.17: 7577006-7577006 on Build GRCh38
rs890965445 Chr.17: 7577126-7577126 on Build GRCh38
rs780000726 Chr.17: 7577369-7577369 on Build GRCh38
rs557992096 Chr.17: 7577361-7577361 on Build GRCh38
rs774133809 Chr.17: 7577002-7577002 on Build GRCh38
rs761424714 Chr.17: 7577339-7577339 on Build GRCh38
rs756863159 Chr.17: 7577033-7577033 on Build GRCh38
rs766667120 Chr.17: 7577017-7577017 on Build GRCh38
rs75223220 Chr.17: 7576998-7576998 on Build GRCh38
rs900690182 Chr.17: 7577123-7577123 on Build GRCh38
rs773598477 Chr.17: 7577010-7577010 on Build GRCh38
rs948443811 Chr.17: 7577296-7577296 on Build GRCh38
rs777229292 Chr.17: 7577029-7577029 on Build GRCh38
rs767840551 Chr.17: 7577108-7577108 on Build GRCh38
rs773367543 Chr.17: 7577057-7577057 on Build GRCh38
rs763821510 Chr.17: 7577050-7577052 on Build GRCh38
rs750010636 Chr.17: 7577184-7577184 on Build GRCh38
rs772452235 Chr.17: 7577311-7577311 on Build GRCh38
rs143864020 Chr.17: 7577096-7577096 on Build GRCh38
rs775760123 Chr.17: 7577321-7577321 on Build GRCh38
rs754808974 Chr.17: 7577379-7577379 on Build GRCh38
rs2270341 Chr.17: 7577229-7577229 on Build GRCh38
rs879194979 Chr.17: 7577316-7577316 on Build GRCh38
rs774453689 Chr.17: 7576999-7577000 on Build GRCh38
rs539986945 Chr.17: 7577177-7577177 on Build GRCh38
rs538141104 Chr.17: 7577008-7577008 on Build GRCh38
rs760482417 Chr.17: 7577046-7577047 on Build GRCh38
rs770016770 Chr.17: 7577053-7577053 on Build GRCh38
rs368852353 Chr.17: 7577181-7577181 on Build GRCh38
rs1038356916 Chr.17: 7577037-7577037 on Build GRCh38
rs1021327452 Chr.17: 7577195-7577195 on Build GRCh38
rs552548590 Chr.17: 7577269-7577269 on Build GRCh38
rs753794876 Chr.17: 7577168-7577169 on Build GRCh38
rs761019570 Chr.17: 7577328-7577328 on Build GRCh38
rs746957247 Chr.17: 7577199-7577199 on Build GRCh38
rs755665027 Chr.17: 7577028-7577028 on Build GRCh38
rs748912261 Chr.17: 7577000-7577000 on Build GRCh38
rs772110555 Chr.17: 7577005-7577005 on Build GRCh38
rs764836748 Chr.17: 7577318-7577319 on Build GRCh38
rs748949296 Chr.17: 7577032-7577032 on Build GRCh38
rs781272890 Chr.17: 7577210-7577210 on Build GRCh38
rs1043393797 Chr.17: 7577009-7577009 on Build GRCh38
rs372790135 Chr.17: 7577035-7577035 on Build GRCh38
rs767835311 Chr.17: 7577038-7577038 on Build GRCh38
rs368324276 Chr.17: 7577004-7577004 on Build GRCh38
rs745731353 Chr.17: 7577046-7577046 on Build GRCh38
rs200590129 Chr.17: 7577202-7577202 on Build GRCh38
rs373429772 Chr.17: 7577196-7577196 on Build GRCh38
rs774676775 Chr.17: 7577302-7577302 on Build GRCh38
rs778814483 Chr.17: 7577440-7577440 on Build GRCh38
rs775584957 Chr.17: 7577037-7577038 on Build GRCh38
rs768131535 Chr.17: 7577024-7577024 on Build GRCh38
rs774354420 Chr.17: 7577021-7577022 on Build GRCh38
rs770895700 Chr.17: 7577293-7577293 on Build GRCh38
rs371767370 Chr.17: 7577081-7577081 on Build GRCh38
rs746103175 Chr.17: 7577446-7577446 on Build GRCh38
rs368089729 Chr.17: 7577020-7577020 on Build GRCh38
rs368882645 Chr.17: 7577189-7577189 on Build GRCh38
rs748007558 Chr.17: 7577390-7577390 on Build GRCh38
rs993027605 Chr.17: 7577279-7577279 on Build GRCh38
rs959728013 Chr.17: 7577232-7577232 on Build GRCh38
rs554303388 Chr.17: 7577337-7577337 on Build GRCh38
rs781329958 Chr.17: 7577381-7577381 on Build GRCh38
rs761109374 Chr.17: 7577168-7577168 on Build GRCh38
rs995004606 Chr.17: 7577155-7577155 on Build GRCh38
rs753183356 Chr.17: 7577027-7577027 on Build GRCh38
rs11552639 Chr.17: 7577393-7577393 on Build GRCh38
rs569334601 Chr.17: 7577069-7577069 on Build GRCh38

View Full Product Details