Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 8162481-8162979 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
VAMP2
Entrez Gene ID
6844
Gene Name
vesicle associated membrane protein 2
Gene Aliases
SYB2, VAMP-2
UniGene
Hs.25348
Gene Chromosome Location
Chr.17: 8159147-8162975 on Build GRCh38
Transcripts
NM_014232.2 XM_005256775.4

SNP Detail

SNP ID Chromosome Location
rs8079444 Chr.17: 8162612-8162612 on Build GRCh38
rs570597684 Chr.17: 8162732-8162732 on Build GRCh38
rs1057504877 Chr.17: 8162569-8162569 on Build GRCh38
rs1039998729 Chr.17: 8162956-8162956 on Build GRCh38
rs1002560384 Chr.17: 8162656-8162656 on Build GRCh38
rs996306417 Chr.17: 8162812-8162812 on Build GRCh38
rs1053631594 Chr.17: 8162807-8162807 on Build GRCh38
rs964490934 Chr.17: 8162722-8162722 on Build GRCh38
rs1008562176 Chr.17: 8162804-8162804 on Build GRCh38
rs766338464 Chr.17: 8162887-8162887 on Build GRCh38
rs916190700 Chr.17: 8162780-8162780 on Build GRCh38
rs982527991 Chr.17: 8162898-8162898 on Build GRCh38
rs931083251 Chr.17: 8162748-8162748 on Build GRCh38
rs1018737138 Chr.17: 8162505-8162505 on Build GRCh38
rs534237719 Chr.17: 8162537-8162537 on Build GRCh38
rs908668709 Chr.17: 8162754-8162754 on Build GRCh38
rs938865343 Chr.17: 8162767-8162767 on Build GRCh38
rs1057354584 Chr.17: 8162773-8162773 on Build GRCh38
rs986712658 Chr.17: 8162703-8162703 on Build GRCh38
rs930522473 Chr.17: 8162553-8162554 on Build GRCh38
rs1052654268 Chr.17: 8162911-8162911 on Build GRCh38
rs951952800 Chr.17: 8162822-8162822 on Build GRCh38
rs529052595 Chr.17: 8162835-8162835 on Build GRCh38
rs939353103 Chr.17: 8162766-8162767 on Build GRCh38
rs998912649 Chr.17: 8162802-8162802 on Build GRCh38
rs1001101161 Chr.17: 8162659-8162659 on Build GRCh38
rs931449451 Chr.17: 8162502-8162502 on Build GRCh38
rs1044246681 Chr.17: 8162599-8162599 on Build GRCh38
rs907001723 Chr.17: 8162571-8162571 on Build GRCh38
rs1056453737 Chr.17: 8162574-8162574 on Build GRCh38
rs948215019 Chr.17: 8162594-8162594 on Build GRCh38
rs545763294 Chr.17: 8162770-8162770 on Build GRCh38
rs374021351 Chr.17: 8162501-8162501 on Build GRCh38
rs551715275 Chr.17: 8162927-8162927 on Build GRCh38
rs944232506 Chr.17: 8162928-8162928 on Build GRCh38
rs1043833951 Chr.17: 8162778-8162778 on Build GRCh38
rs1047265507 Chr.17: 8162518-8162518 on Build GRCh38
rs996505929 Chr.17: 8162884-8162884 on Build GRCh38
rs1004757873 Chr.17: 8162824-8162824 on Build GRCh38
rs751694318 Chr.17: 8162600-8162600 on Build GRCh38
rs1021648827 Chr.17: 8162621-8162621 on Build GRCh38
rs938911704 Chr.17: 8162561-8162561 on Build GRCh38
rs114050157 Chr.17: 8162652-8162652 on Build GRCh38
rs907807634 Chr.17: 8162760-8162760 on Build GRCh38
rs963694200 Chr.17: 8162723-8162723 on Build GRCh38
rs550403327 Chr.17: 8162843-8162843 on Build GRCh38
rs1041526357 Chr.17: 8162553-8162553 on Build GRCh38
rs575280028 Chr.17: 8162741-8162741 on Build GRCh38
rs982945713 Chr.17: 8162753-8162753 on Build GRCh38
rs894455881 Chr.17: 8162591-8162591 on Build GRCh38
rs956750373 Chr.17: 8162698-8162698 on Build GRCh38
rs935496813 Chr.17: 8162909-8162909 on Build GRCh38
rs948097627 Chr.17: 8162777-8162777 on Build GRCh38
rs539569127 Chr.17: 8162892-8162892 on Build GRCh38
rs964523572 Chr.17: 8162525-8162525 on Build GRCh38
rs2304909 Chr.17: 8162789-8162789 on Build GRCh38
rs200515650 Chr.17: 8162554-8162554 on Build GRCh38
rs960577750 Chr.17: 8162858-8162858 on Build GRCh38
rs112118752 Chr.17: 8162866-8162866 on Build GRCh38
rs368261829 Chr.17: 8162576-8162576 on Build GRCh38
rs188854454 Chr.17: 8162885-8162885 on Build GRCh38
rs1018706047 Chr.17: 8162702-8162702 on Build GRCh38
rs916586393 Chr.17: 8162772-8162772 on Build GRCh38
rs142488375 Chr.17: 8162660-8162660 on Build GRCh38
rs992038091 Chr.17: 8162875-8162875 on Build GRCh38
rs974498210 Chr.17: 8162726-8162726 on Build GRCh38
rs1008507617 Chr.17: 8162818-8162818 on Build GRCh38
rs907844923 Chr.17: 8162562-8162562 on Build GRCh38
rs563948088 Chr.17: 8162774-8162774 on Build GRCh38
rs920515471 Chr.17: 8162548-8162548 on Build GRCh38
rs895273744 Chr.17: 8162590-8162590 on Build GRCh38
rs963994813 Chr.17: 8162867-8162867 on Build GRCh38
rs1019556538 Chr.17: 8162705-8162705 on Build GRCh38
rs561502498 Chr.17: 8162811-8162811 on Build GRCh38
rs566761247 Chr.17: 8162937-8162937 on Build GRCh38
rs1053098300 Chr.17: 8162795-8162795 on Build GRCh38
rs983760198 Chr.17: 8162742-8162742 on Build GRCh38
rs916555137 Chr.17: 8162883-8162883 on Build GRCh38
rs377276190 Chr.17: 8162680-8162680 on Build GRCh38
rs546589683 Chr.17: 8162799-8162799 on Build GRCh38
rs1008593151 Chr.17: 8162696-8162696 on Build GRCh38
rs887348500 Chr.17: 8162560-8162560 on Build GRCh38
rs754782997 Chr.17: 8162897-8162897 on Build GRCh38
rs919671838 Chr.17: 8162733-8162733 on Build GRCh38
rs904541674 Chr.17: 8162801-8162801 on Build GRCh38

View Full Product Details