Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 8287651-8288151 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RANGRF
Entrez Gene ID
29098
Gene Name
RAN guanine nucleotide release factor
Gene Aliases
HSPC165, HSPC236, MOG1, RANGNRF
UniGene
Hs.408233
Gene Chromosome Location
Chr.17: 8287150-8290091 on Build GRCh38
Transcripts
XM_005256618.4 NM_001320871.1 NM_201520.2

Gene Symbol
SLC25A35
Entrez Gene ID
399512
Gene Name
solute carrier family 25 member 35
Gene Aliases
-
UniGene
Hs.118918
Gene Chromosome Location
Chr.17: 8287763-8295694 on Build GRCh38
Transcripts
XM_005256618.4 NM_001320871.1 NM_201520.2

SNP Detail

SNP ID Chromosome Location
rs1055396061 Chr.17: 8288034-8288034 on Build GRCh38
rs536190753 Chr.17: 8287676-8287676 on Build GRCh38
rs988103993 Chr.17: 8287942-8287942 on Build GRCh38
rs1020020402 Chr.17: 8288078-8288078 on Build GRCh38
rs554736692 Chr.17: 8287680-8287680 on Build GRCh38
rs899682707 Chr.17: 8287746-8287746 on Build GRCh38
rs764915101 Chr.17: 8287675-8287675 on Build GRCh38
rs576235274 Chr.17: 8287811-8287811 on Build GRCh38
rs960291598 Chr.17: 8287863-8287863 on Build GRCh38
rs1030874698 Chr.17: 8288112-8288112 on Build GRCh38
rs750181243 Chr.17: 8287840-8287840 on Build GRCh38
rs758014220 Chr.17: 8287899-8287899 on Build GRCh38
rs576791555 Chr.17: 8287916-8287916 on Build GRCh38
rs937220808 Chr.17: 8288016-8288016 on Build GRCh38
rs986131698 Chr.17: 8287838-8287838 on Build GRCh38
rs576202002 Chr.17: 8287740-8287740 on Build GRCh38
rs976642321 Chr.17: 8287788-8287788 on Build GRCh38
rs1004152798 Chr.17: 8288092-8288092 on Build GRCh38
rs146646084 Chr.17: 8288088-8288088 on Build GRCh38
rs912242478 Chr.17: 8287983-8287983 on Build GRCh38
rs537098363 Chr.17: 8287836-8287836 on Build GRCh38
rs1011178032 Chr.17: 8288058-8288058 on Build GRCh38
rs972121058 Chr.17: 8287853-8287853 on Build GRCh38
rs558633066 Chr.17: 8287878-8287878 on Build GRCh38
rs943666205 Chr.17: 8287984-8287984 on Build GRCh38
rs907157772 Chr.17: 8287973-8287973 on Build GRCh38
rs1052093786 Chr.17: 8287681-8287681 on Build GRCh38
rs956298341 Chr.17: 8287929-8287930 on Build GRCh38
rs968877694 Chr.17: 8287896-8287896 on Build GRCh38
rs909176878 Chr.17: 8287839-8287839 on Build GRCh38
rs1030614885 Chr.17: 8287828-8287828 on Build GRCh38
rs892780298 Chr.17: 8288057-8288057 on Build GRCh38
rs902923869 Chr.17: 8288082-8288082 on Build GRCh38
rs1023022485 Chr.17: 8287886-8287886 on Build GRCh38
rs1035678845 Chr.17: 8287861-8287861 on Build GRCh38
rs1039830340 Chr.17: 8287717-8287717 on Build GRCh38
rs890892551 Chr.17: 8287697-8287697 on Build GRCh38
rs943819918 Chr.17: 8287698-8287698 on Build GRCh38
rs924885647 Chr.17: 8287921-8287921 on Build GRCh38

View Full Product Details