Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 8474115-8474609 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYH10
Entrez Gene ID
4628
Gene Name
myosin, heavy chain 10, non-muscle
Gene Aliases
NMMHC-IIB, NMMHCB
UniGene
Hs.16355
Gene Chromosome Location
Chr.17: 8474205-8631369 on Build GRCh38
Transcripts
NM_001256012.1 NM_001256095.1 NM_005964.3 XM_011523875.2 XM_011523878.2 XM_011523879.2 XM_011523880.2 XM_017024677.1 XM_017024678.1 XM_017024679.1 XM_017024680.1 XM_017024681.1 XM_017024682.1

SNP Detail

SNP ID Chromosome Location
rs138136961 Chr.17: 8474502-8474502 on Build GRCh38
rs928150171 Chr.17: 8474406-8474406 on Build GRCh38
rs934593427 Chr.17: 8474518-8474518 on Build GRCh38
rs959836895 Chr.17: 8474430-8474430 on Build GRCh38
rs777450568 Chr.17: 8474278-8474278 on Build GRCh38
rs947060952 Chr.17: 8474256-8474256 on Build GRCh38
rs1055990111 Chr.17: 8474250-8474250 on Build GRCh38
rs550377014 Chr.17: 8474508-8474508 on Build GRCh38
rs571845631 Chr.17: 8474510-8474510 on Build GRCh38
rs561532386 Chr.17: 8474443-8474443 on Build GRCh38
rs947135922 Chr.17: 8474449-8474449 on Build GRCh38
rs1007634696 Chr.17: 8474293-8474293 on Build GRCh38
rs1004863689 Chr.17: 8474271-8474271 on Build GRCh38
rs1030593334 Chr.17: 8474152-8474152 on Build GRCh38
rs998678560 Chr.17: 8474149-8474149 on Build GRCh38
rs978573730 Chr.17: 8474345-8474345 on Build GRCh38
rs1049277511 Chr.17: 8474498-8474498 on Build GRCh38
rs915717275 Chr.17: 8474170-8474170 on Build GRCh38
rs963380343 Chr.17: 8474193-8474193 on Build GRCh38
rs925706713 Chr.17: 8474191-8474191 on Build GRCh38
rs919533413 Chr.17: 8474222-8474222 on Build GRCh38
rs143610111 Chr.17: 8474542-8474542 on Build GRCh38
rs995008602 Chr.17: 8474240-8474240 on Build GRCh38
rs960769524 Chr.17: 8474307-8474307 on Build GRCh38
rs934427278 Chr.17: 8474377-8474377 on Build GRCh38
rs1027709209 Chr.17: 8474247-8474247 on Build GRCh38
rs929565720 Chr.17: 8474226-8474226 on Build GRCh38
rs954782859 Chr.17: 8474153-8474153 on Build GRCh38
rs1042765427 Chr.17: 8474275-8474275 on Build GRCh38
rs1042783780 Chr.17: 8474181-8474181 on Build GRCh38
rs950793115 Chr.17: 8474255-8474255 on Build GRCh38
rs991220240 Chr.17: 8474434-8474434 on Build GRCh38
rs982772365 Chr.17: 8474381-8474383 on Build GRCh38
rs566834866 Chr.17: 8474211-8474211 on Build GRCh38
rs578015985 Chr.17: 8474210-8474210 on Build GRCh38
rs571313923 Chr.17: 8474369-8474369 on Build GRCh38
rs34231620 Chr.17: 8474459-8474459 on Build GRCh38
rs1026283820 Chr.17: 8474361-8474361 on Build GRCh38
rs534277744 Chr.17: 8474246-8474246 on Build GRCh38
rs986624145 Chr.17: 8474157-8474157 on Build GRCh38
rs1035414187 Chr.17: 8474289-8474289 on Build GRCh38
rs890687811 Chr.17: 8474207-8474207 on Build GRCh38
rs900667228 Chr.17: 8474223-8474223 on Build GRCh38
rs532877715 Chr.17: 8474534-8474534 on Build GRCh38
rs1012251258 Chr.17: 8474539-8474539 on Build GRCh38
rs1035145586 Chr.17: 8474530-8474530 on Build GRCh38
rs969823783 Chr.17: 8474334-8474334 on Build GRCh38
rs1052823924 Chr.17: 8474392-8474392 on Build GRCh38
rs1039655702 Chr.17: 8474452-8474452 on Build GRCh38
rs928127839 Chr.17: 8474245-8474245 on Build GRCh38
rs747924049 Chr.17: 8474192-8474192 on Build GRCh38
rs993146927 Chr.17: 8474328-8474328 on Build GRCh38
rs911533356 Chr.17: 8474429-8474429 on Build GRCh38
rs903076005 Chr.17: 8474279-8474279 on Build GRCh38
rs560285120 Chr.17: 8474332-8474332 on Build GRCh38
rs1017556650 Chr.17: 8474181-8474187 on Build GRCh38
rs147002459 Chr.17: 8474288-8474288 on Build GRCh38
rs982271473 Chr.17: 8474227-8474227 on Build GRCh38
rs543055824 Chr.17: 8474420-8474420 on Build GRCh38
rs527640767 Chr.17: 8474370-8474370 on Build GRCh38
rs973352790 Chr.17: 8474339-8474339 on Build GRCh38
rs1007827330 Chr.17: 8474541-8474541 on Build GRCh38
rs770343423 Chr.17: 8474407-8474407 on Build GRCh38
rs994977910 Chr.17: 8474476-8474476 on Build GRCh38
rs781652111 Chr.17: 8474280-8474281 on Build GRCh38
rs1023269846 Chr.17: 8474540-8474540 on Build GRCh38
rs1052730418 Chr.17: 8474201-8474201 on Build GRCh38
rs948614864 Chr.17: 8474180-8474180 on Build GRCh38

View Full Product Details