Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 9628247-9628747 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP52
Entrez Gene ID
146845
Gene Name
cilia and flagella associated protein 52
Gene Aliases
WDR16, WDRPUH
UniGene
Hs.232270
Gene Chromosome Location
Chr.17: 9576627-9643459 on Build GRCh38
Transcripts
NM_001080556.1 NM_145054.4 XM_017024227.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6585299 c.1057G>A p.G353S Chr.17: 9628673-9628673 on Build GRCh38
4130902 c.1065T>C p.D355D Chr.17: 9628711-9628711 on Build GRCh38
4649936 c.1063G>A p.A355T Chr.17: 9628679-9628679 on Build GRCh38
3524097 c.1096G>A p.E366K Chr.17: 9628742-9628742 on Build GRCh38
4273828 c.1107G>A p.W369* Chr.17: 9628723-9628723 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367791419 Chr.17: 9628717-9628717 on Build GRCh38
rs148803926 Chr.17: 9628565-9628565 on Build GRCh38
rs1007350506 Chr.17: 9628459-9628459 on Build GRCh38
rs947598191 Chr.17: 9628586-9628586 on Build GRCh38
rs780107266 Chr.17: 9628469-9628469 on Build GRCh38
rs993100102 Chr.17: 9628495-9628495 on Build GRCh38
rs574869940 Chr.17: 9628427-9628427 on Build GRCh38
rs911572288 Chr.17: 9628294-9628294 on Build GRCh38
rs141229371 Chr.17: 9628677-9628677 on Build GRCh38
rs766413263 Chr.17: 9628623-9628623 on Build GRCh38
rs57960962 Chr.17: 9628711-9628711 on Build GRCh38
rs368310731 Chr.17: 9628632-9628632 on Build GRCh38
rs565658366 Chr.17: 9628569-9628569 on Build GRCh38
rs895840668 Chr.17: 9628515-9628515 on Build GRCh38
rs537397294 Chr.17: 9628285-9628285 on Build GRCh38
rs1012916906 Chr.17: 9628536-9628536 on Build GRCh38
rs138276250 Chr.17: 9628375-9628375 on Build GRCh38
rs529781256 Chr.17: 9628611-9628611 on Build GRCh38
rs562254713 Chr.17: 9628482-9628482 on Build GRCh38
rs748343871 Chr.17: 9628617-9628617 on Build GRCh38
rs987162450 Chr.17: 9628286-9628287 on Build GRCh38
rs937528659 Chr.17: 9628320-9628320 on Build GRCh38
rs1051890177 Chr.17: 9628430-9628430 on Build GRCh38
rs776242745 Chr.17: 9628566-9628566 on Build GRCh38
rs749427640 Chr.17: 9628478-9628478 on Build GRCh38
rs542285947 Chr.17: 9628440-9628440 on Build GRCh38
rs756901981 Chr.17: 9628696-9628696 on Build GRCh38
rs1026383306 Chr.17: 9628503-9628503 on Build GRCh38
rs933386957 Chr.17: 9628426-9628426 on Build GRCh38
rs1017417880 Chr.17: 9628298-9628298 on Build GRCh38
rs61617597 Chr.17: 9628297-9628297 on Build GRCh38
rs541046658 Chr.17: 9628382-9628382 on Build GRCh38
rs1052241941 Chr.17: 9628704-9628704 on Build GRCh38
rs778381409 Chr.17: 9628702-9628702 on Build GRCh38
rs978973838 Chr.17: 9628579-9628579 on Build GRCh38
rs759559976 Chr.17: 9628661-9628661 on Build GRCh38
rs570211567 Chr.17: 9628278-9628278 on Build GRCh38
rs374648753 Chr.17: 9628674-9628674 on Build GRCh38
rs113576151 Chr.17: 9628693-9628693 on Build GRCh38
rs186226984 Chr.17: 9628568-9628568 on Build GRCh38
rs1023059243 Chr.17: 9628546-9628546 on Build GRCh38
rs755094177 Chr.17: 9628486-9628486 on Build GRCh38
rs920396220 Chr.17: 9628673-9628673 on Build GRCh38
rs560970743 Chr.17: 9628441-9628441 on Build GRCh38
rs75671790 Chr.17: 9628669-9628669 on Build GRCh38
rs147509941 Chr.17: 9628306-9628306 on Build GRCh38
rs144401450 Chr.17: 9628514-9628514 on Build GRCh38
rs970449384 Chr.17: 9628513-9628513 on Build GRCh38
rs886382085 Chr.17: 9628488-9628488 on Build GRCh38
rs756073774 Chr.17: 9628419-9628419 on Build GRCh38
rs934299798 Chr.17: 9628282-9628282 on Build GRCh38
rs1056058120 Chr.17: 9628358-9628358 on Build GRCh38
rs751688968 Chr.17: 9628645-9628645 on Build GRCh38
rs149630996 Chr.17: 9628383-9628383 on Build GRCh38
rs889213613 Chr.17: 9628710-9628710 on Build GRCh38
rs1033475867 Chr.17: 9628540-9628540 on Build GRCh38
rs543603225 Chr.17: 9628529-9628529 on Build GRCh38
rs769230477 Chr.17: 9628606-9628606 on Build GRCh38
rs373711395 Chr.17: 9628567-9628567 on Build GRCh38
rs540718330 Chr.17: 9628468-9628468 on Build GRCh38
rs867819243 Chr.17: 9628708-9628708 on Build GRCh38
rs956245705 Chr.17: 9628614-9628614 on Build GRCh38

View Full Product Details